Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 178852550-178853091 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANGPTL1
Entrez Gene ID
9068
Gene Name
angiopoietin like 1
Gene Aliases
ANG3, ANGPT3, ARP1, AngY, UNQ162, dJ595C2.2
UniGene
Hs.591474
Gene Chromosome Location
Chr.1: 178849535-178871353 on Build GRCh38
Transcripts
NM_004673.3 XM_005245577.2 NM_001286247.1 NM_152663.4 XM_006711410.3 XM_006711411.3 XM_011509688.1 XM_017001591.1

Gene Symbol
RALGPS2
Entrez Gene ID
55103
Gene Name
Ral GEF with PH domain and SH3 binding motif 2
Gene Aliases
dJ595C2.1
UniGene
Hs.632485
Gene Chromosome Location
Chr.1: 178725147-178921842 on Build GRCh38
Transcripts
NM_004673.3 XM_005245577.2 NM_001286247.1 NM_152663.4 XM_006711410.3 XM_006711411.3 XM_011509688.1 XM_017001591.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6599635 c.1048T>C p.W350R Chr.1: 178852923-178852923 on Build GRCh38
6310521 c.1018-3C>A p.? Chr.1: 178852956-178852956 on Build GRCh38
4521359 c.1108G>A p.E370K Chr.1: 178852863-178852863 on Build GRCh38
5480330 c.1148A>G p.Y383C Chr.1: 178852823-178852823 on Build GRCh38
5126351 c.1039G>A p.G347R Chr.1: 178852932-178852932 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs150839113 Chr.1: 178852968-178852968 on Build GRCh38
rs544426338 Chr.1: 178852648-178852648 on Build GRCh38
rs779355325 Chr.1: 178852806-178852806 on Build GRCh38
rs753970255 Chr.1: 178852889-178852889 on Build GRCh38
rs979717900 Chr.1: 178852607-178852607 on Build GRCh38
rs114222276 Chr.1: 178853033-178853033 on Build GRCh38
rs767995957 Chr.1: 178852983-178852983 on Build GRCh38
rs776238636 Chr.1: 178852637-178852637 on Build GRCh38
rs980044720 Chr.1: 178852777-178852777 on Build GRCh38
rs372465278 Chr.1: 178852655-178852655 on Build GRCh38
rs34332039 Chr.1: 178852786-178852786 on Build GRCh38
rs375369347 Chr.1: 178852882-178852882 on Build GRCh38
rs1045604208 Chr.1: 178852803-178852803 on Build GRCh38
rs183816858 Chr.1: 178853038-178853038 on Build GRCh38
rs763830126 Chr.1: 178852754-178852754 on Build GRCh38
rs759326599 Chr.1: 178852641-178852641 on Build GRCh38
rs146890252 Chr.1: 178852933-178852933 on Build GRCh38
rs756202340 Chr.1: 178852683-178852683 on Build GRCh38
rs539906053 Chr.1: 178852590-178852590 on Build GRCh38
rs747508264 Chr.1: 178852928-178852929 on Build GRCh38
rs575486511 Chr.1: 178852634-178852634 on Build GRCh38
rs138264170 Chr.1: 178852926-178852926 on Build GRCh38
rs761587922 Chr.1: 178852848-178852848 on Build GRCh38
rs201219967 Chr.1: 178852989-178852989 on Build GRCh38
rs138794873 Chr.1: 178852602-178852602 on Build GRCh38
rs765449284 Chr.1: 178852990-178852990 on Build GRCh38
rs751357250 Chr.1: 178852765-178852765 on Build GRCh38
rs745952913 Chr.1: 178852749-178852749 on Build GRCh38
rs779151567 Chr.1: 178852680-178852681 on Build GRCh38
rs188281805 Chr.1: 178852616-178852616 on Build GRCh38
rs777052895 Chr.1: 178852986-178852986 on Build GRCh38
rs763920096 Chr.1: 178852658-178852658 on Build GRCh38
rs779478516 Chr.1: 178852932-178852932 on Build GRCh38
rs755337121 Chr.1: 178852710-178852710 on Build GRCh38
rs202098083 Chr.1: 178852817-178852817 on Build GRCh38
rs34483360 Chr.1: 178852705-178852705 on Build GRCh38
rs765937339 Chr.1: 178852636-178852636 on Build GRCh38
rs566764958 Chr.1: 178852741-178852741 on Build GRCh38
rs189235675 Chr.1: 178853040-178853040 on Build GRCh38
rs1006754094 Chr.1: 178852793-178852793 on Build GRCh38
rs772128964 Chr.1: 178852832-178852833 on Build GRCh38
rs780893309 Chr.1: 178852809-178852809 on Build GRCh38
rs755571333 Chr.1: 178852798-178852798 on Build GRCh38
rs555651437 Chr.1: 178852631-178852631 on Build GRCh38
rs200482313 Chr.1: 178852714-178852714 on Build GRCh38
rs767763090 Chr.1: 178852775-178852775 on Build GRCh38
rs780046482 Chr.1: 178852780-178852780 on Build GRCh38
rs780502740 Chr.1: 178852729-178852729 on Build GRCh38
rs751052965 Chr.1: 178852865-178852865 on Build GRCh38
rs951921095 Chr.1: 178852645-178852645 on Build GRCh38
rs778052392 Chr.1: 178852917-178852917 on Build GRCh38
rs573187175 Chr.1: 178852610-178852610 on Build GRCh38
rs551634791 Chr.1: 178852952-178852952 on Build GRCh38
rs368824919 Chr.1: 178852923-178852923 on Build GRCh38
rs372846517 Chr.1: 178853018-178853018 on Build GRCh38
rs374600280 Chr.1: 178852811-178852811 on Build GRCh38
rs753324412 Chr.1: 178852912-178852912 on Build GRCh38
rs775471958 Chr.1: 178852732-178852732 on Build GRCh38
rs768591296 Chr.1: 178852821-178852821 on Build GRCh38
rs760452268 Chr.1: 178852635-178852635 on Build GRCh38
rs570797031 Chr.1: 178852778-178852778 on Build GRCh38
rs991041322 Chr.1: 178852885-178852885 on Build GRCh38
rs751762108 Chr.1: 178852665-178852665 on Build GRCh38
rs775978569 Chr.1: 178852836-178852837 on Build GRCh38
rs765234857 Chr.1: 178852643-178852643 on Build GRCh38
rs762930538 Chr.1: 178852738-178852738 on Build GRCh38
rs949407144 Chr.1: 178852801-178852801 on Build GRCh38
rs749799101 Chr.1: 178852690-178852690 on Build GRCh38
rs766440885 Chr.1: 178852886-178852886 on Build GRCh38
rs370931833 Chr.1: 178852807-178852807 on Build GRCh38
rs372163774 Chr.1: 178852954-178852954 on Build GRCh38
rs979711211 Chr.1: 178853031-178853031 on Build GRCh38
rs767252550 Chr.1: 178852863-178852863 on Build GRCh38
rs191109199 Chr.1: 178853042-178853042 on Build GRCh38
rs141473288 Chr.1: 178852934-178852934 on Build GRCh38
rs745479614 Chr.1: 178852920-178852920 on Build GRCh38
rs760883969 Chr.1: 178852978-178852978 on Build GRCh38
rs758614879 Chr.1: 178852782-178852782 on Build GRCh38
rs766524285 Chr.1: 178852982-178852982 on Build GRCh38
rs367915584 Chr.1: 178852907-178852907 on Build GRCh38
rs905680530 Chr.1: 178852577-178852577 on Build GRCh38
rs745970245 Chr.1: 178852682-178852682 on Build GRCh38
rs762840189 Chr.1: 178852820-178852820 on Build GRCh38
rs548949964 Chr.1: 178852915-178852915 on Build GRCh38
rs769787196 Chr.1: 178852819-178852819 on Build GRCh38
rs759074163 Chr.1: 178852717-178852717 on Build GRCh38
rs755486642 Chr.1: 178852898-178852898 on Build GRCh38
rs757495599 Chr.1: 178852674-178852674 on Build GRCh38
rs1045638009 Chr.1: 178852576-178852576 on Build GRCh38
rs753237115 Chr.1: 178852991-178852991 on Build GRCh38
rs931201405 Chr.1: 178852767-178852767 on Build GRCh38
rs140806007 Chr.1: 178852779-178852779 on Build GRCh38
rs771850663 Chr.1: 178852964-178852964 on Build GRCh38
rs570971426 Chr.1: 178852580-178852580 on Build GRCh38
rs370767726 Chr.1: 178852824-178852824 on Build GRCh38
rs377271356 Chr.1: 178852816-178852816 on Build GRCh38
rs780602915 Chr.1: 178852685-178852685 on Build GRCh38
rs564307595 Chr.1: 178852675-178852675 on Build GRCh38
rs976301717 Chr.1: 178852720-178852720 on Build GRCh38
rs200844291 Chr.1: 178852697-178852697 on Build GRCh38

View Full Product Details