Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 178870549-178871049 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANGPTL1
Entrez Gene ID
9068
Gene Name
angiopoietin like 1
Gene Aliases
ANG3, ANGPT3, ARP1, AngY, UNQ162, dJ595C2.2
UniGene
Hs.591474
Gene Chromosome Location
Chr.1: 178849535-178871353 on Build GRCh38
Transcripts
NM_004673.3 XM_005245577.2 NM_001286247.1 NM_152663.4 XM_006711410.3 XM_006711411.3 XM_011509688.1 XM_017001591.1

Gene Symbol
RALGPS2
Entrez Gene ID
55103
Gene Name
Ral GEF with PH domain and SH3 binding motif 2
Gene Aliases
dJ595C2.1
UniGene
Hs.632485
Gene Chromosome Location
Chr.1: 178725147-178921842 on Build GRCh38
Transcripts
NM_004673.3 XM_005245577.2 NM_001286247.1 NM_152663.4 XM_006711410.3 XM_006711411.3 XM_011509688.1 XM_017001591.1

SNP Detail

SNP ID Chromosome Location
rs201639859 Chr.1: 178870853-178870853 on Build GRCh38
rs965670406 Chr.1: 178870654-178870655 on Build GRCh38
rs184109146 Chr.1: 178870823-178870823 on Build GRCh38
rs536806387 Chr.1: 178871017-178871017 on Build GRCh38
rs1028737495 Chr.1: 178870598-178870599 on Build GRCh38
rs903742364 Chr.1: 178870575-178870575 on Build GRCh38
rs1026622919 Chr.1: 178870782-178870782 on Build GRCh38
rs369479007 Chr.1: 178870924-178870924 on Build GRCh38
rs951611060 Chr.1: 178870805-178870805 on Build GRCh38
rs554506610 Chr.1: 178870979-178870979 on Build GRCh38
rs896308635 Chr.1: 178871003-178871003 on Build GRCh38
rs937392812 Chr.1: 178870798-178870798 on Build GRCh38
rs924630791 Chr.1: 178870954-178870954 on Build GRCh38
rs141668952 Chr.1: 178870901-178870901 on Build GRCh38
rs1013384474 Chr.1: 178871009-178871009 on Build GRCh38
rs953123580 Chr.1: 178870604-178870604 on Build GRCh38
rs772830724 Chr.1: 178870646-178870646 on Build GRCh38
rs1023424025 Chr.1: 178870731-178870731 on Build GRCh38
rs771751285 Chr.1: 178870620-178870620 on Build GRCh38
rs529795702 Chr.1: 178870586-178870586 on Build GRCh38
rs549848797 Chr.1: 178870690-178870690 on Build GRCh38
rs995103129 Chr.1: 178870742-178870742 on Build GRCh38
rs113881816 Chr.1: 178870912-178870912 on Build GRCh38
rs965612183 Chr.1: 178870862-178870862 on Build GRCh38
rs913403503 Chr.1: 178870701-178870701 on Build GRCh38
rs969547790 Chr.1: 178870741-178870741 on Build GRCh38
rs947466019 Chr.1: 178870902-178870902 on Build GRCh38
rs375812635 Chr.1: 178870851-178870851 on Build GRCh38
rs896350032 Chr.1: 178870661-178870661 on Build GRCh38
rs983022062 Chr.1: 178870819-178870819 on Build GRCh38
rs994661650 Chr.1: 178870956-178870956 on Build GRCh38
rs969333611 Chr.1: 178870727-178870727 on Build GRCh38
rs531992775 Chr.1: 178870841-178870841 on Build GRCh38
rs147053319 Chr.1: 178871000-178871000 on Build GRCh38
rs1008546497 Chr.1: 178870624-178870625 on Build GRCh38
rs565665503 Chr.1: 178870946-178870946 on Build GRCh38
rs1013745142 Chr.1: 178870664-178870664 on Build GRCh38
rs766990046 Chr.1: 178870980-178870980 on Build GRCh38
rs796225902 Chr.1: 178870922-178870922 on Build GRCh38
rs975989204 Chr.1: 178870896-178870896 on Build GRCh38
rs1036648463 Chr.1: 178870934-178870934 on Build GRCh38
rs190202179 Chr.1: 178870951-178870951 on Build GRCh38
rs978919097 Chr.1: 178870732-178870732 on Build GRCh38
rs771585526 Chr.1: 178871003-178871005 on Build GRCh38

View Full Product Details