Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 179900109-179900657 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TOR1AIP1
Entrez Gene ID
26092
Gene Name
torsin 1A interacting protein 1
Gene Aliases
LAP1, LAP1B
UniGene
Hs.496459
Gene Chromosome Location
Chr.1: 179882042-179920077 on Build GRCh38
Transcripts
NM_001267578.1 NM_015602.3 XM_011509403.1 XM_011509404.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6296803 c.620G>T p.S207I Chr.1: 179900135-179900135 on Build GRCh38
1336741 c.648A>C p.E216D Chr.1: 179900163-179900163 on Build GRCh38
3360522 c.638A>G p.N213S Chr.1: 179900153-179900153 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs905966426 Chr.1: 179900265-179900265 on Build GRCh38
rs768288848 Chr.1: 179900212-179900212 on Build GRCh38
rs74263853 Chr.1: 179900576-179900579 on Build GRCh38
rs73050384 Chr.1: 179900487-179900487 on Build GRCh38
rs189281110 Chr.1: 179900471-179900471 on Build GRCh38
rs761178376 Chr.1: 179900209-179900210 on Build GRCh38
rs554933247 Chr.1: 179900411-179900411 on Build GRCh38
rs771353312 Chr.1: 179900449-179900449 on Build GRCh38
rs921889753 Chr.1: 179900522-179900522 on Build GRCh38
rs932179034 Chr.1: 179900368-179900368 on Build GRCh38
rs925407658 Chr.1: 179900375-179900375 on Build GRCh38
rs764529950 Chr.1: 179900215-179900216 on Build GRCh38
rs980349623 Chr.1: 179900633-179900633 on Build GRCh38
rs925461437 Chr.1: 179900574-179900574 on Build GRCh38
rs148254748 Chr.1: 179900539-179900539 on Build GRCh38
rs1001955066 Chr.1: 179900268-179900268 on Build GRCh38
rs953772369 Chr.1: 179900383-179900384 on Build GRCh38
rs758417719 Chr.1: 179900148-179900148 on Build GRCh38
rs778326858 Chr.1: 179900161-179900161 on Build GRCh38
rs896181526 Chr.1: 179900416-179900416 on Build GRCh38
rs1044999737 Chr.1: 179900429-179900429 on Build GRCh38
rs186038215 Chr.1: 179900444-179900444 on Build GRCh38
rs397982119 Chr.1: 179900578-179900579 on Build GRCh38
rs542190943 Chr.1: 179900571-179900571 on Build GRCh38
rs1019380660 Chr.1: 179900577-179900580 on Build GRCh38
rs1027982695 Chr.1: 179900540-179900540 on Build GRCh38
rs989002327 Chr.1: 179900593-179900593 on Build GRCh38
rs759179638 Chr.1: 179900477-179900477 on Build GRCh38
rs114697736 Chr.1: 179900453-179900453 on Build GRCh38
rs966424462 Chr.1: 179900300-179900300 on Build GRCh38
rs776133315 Chr.1: 179900201-179900201 on Build GRCh38
rs200946499 Chr.1: 179900567-179900567 on Build GRCh38
rs912710595 Chr.1: 179900410-179900410 on Build GRCh38
rs752124781 Chr.1: 179900173-179900173 on Build GRCh38
rs200495464 Chr.1: 179900153-179900153 on Build GRCh38
rs778444673 Chr.1: 179900213-179900213 on Build GRCh38
rs914771290 Chr.1: 179900598-179900598 on Build GRCh38
rs1043920693 Chr.1: 179900373-179900373 on Build GRCh38
rs746253295 Chr.1: 179900196-179900196 on Build GRCh38
rs112435086 Chr.1: 179900281-179900281 on Build GRCh38
rs768139213 Chr.1: 179900198-179900198 on Build GRCh38
rs776262904 Chr.1: 179900467-179900467 on Build GRCh38
rs956376301 Chr.1: 179900579-179900579 on Build GRCh38
rs960600169 Chr.1: 179900350-179900351 on Build GRCh38
rs907886454 Chr.1: 179900282-179900282 on Build GRCh38
rs898416244 Chr.1: 179900530-179900530 on Build GRCh38
rs905121085 Chr.1: 179900439-179900439 on Build GRCh38
rs66464156 Chr.1: 179900566-179900567 on Build GRCh38
rs765405746 Chr.1: 179900136-179900136 on Build GRCh38
rs575588949 Chr.1: 179900505-179900505 on Build GRCh38
rs966106827 Chr.1: 179900495-179900495 on Build GRCh38
rs774316316 Chr.1: 179900155-179900157 on Build GRCh38
rs558201282 Chr.1: 179900605-179900605 on Build GRCh38
rs535457954 Chr.1: 179900359-179900359 on Build GRCh38
rs935110219 Chr.1: 179900402-179900402 on Build GRCh38
rs986938100 Chr.1: 179900576-179900576 on Build GRCh38
rs935483759 Chr.1: 179900580-179900580 on Build GRCh38
rs749339374 Chr.1: 179900345-179900345 on Build GRCh38
rs575063885 Chr.1: 179900417-179900417 on Build GRCh38
rs141252138 Chr.1: 179900631-179900631 on Build GRCh38
rs757586064 Chr.1: 179900176-179900176 on Build GRCh38
rs987941821 Chr.1: 179900615-179900615 on Build GRCh38
rs781584462 Chr.1: 179900195-179900195 on Build GRCh38
rs759786680 Chr.1: 179900134-179900134 on Build GRCh38
rs181454553 Chr.1: 179900228-179900228 on Build GRCh38
rs759630373 Chr.1: 179900183-179900187 on Build GRCh38
rs952136339 Chr.1: 179900220-179900220 on Build GRCh38
rs922173240 Chr.1: 179900366-179900366 on Build GRCh38
rs752751720 Chr.1: 179900142-179900142 on Build GRCh38

View Full Product Details