Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 180166384-180166878 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
QSOX1
Entrez Gene ID
5768
Gene Name
quiescin sulfhydryl oxidase 1
Gene Aliases
Q6, QSCN6
UniGene
Hs.744925
Gene Chromosome Location
Chr.1: 180154833-180198034 on Build GRCh38
Transcripts
NM_001004128.2 NM_002826.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6059852 c.347C>T p.P116L Chr.1: 180166572-180166572 on Build GRCh38
677775 c.269G>T p.W90L Chr.1: 180166494-180166494 on Build GRCh38
4025675 c.292G>A p.A98T Chr.1: 180166517-180166517 on Build GRCh38
399848 c.298G>C p.D100H Chr.1: 180166523-180166523 on Build GRCh38
5473255 c.275C>T p.P92L Chr.1: 180166500-180166500 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs189033021 Chr.1: 180166745-180166745 on Build GRCh38
rs777327659 Chr.1: 180166439-180166439 on Build GRCh38
rs1048312302 Chr.1: 180166827-180166827 on Build GRCh38
rs148397885 Chr.1: 180166538-180166538 on Build GRCh38
rs966185207 Chr.1: 180166423-180166423 on Build GRCh38
rs772042431 Chr.1: 180166548-180166548 on Build GRCh38
rs767119330 Chr.1: 180166519-180166519 on Build GRCh38
rs750310533 Chr.1: 180166520-180166520 on Build GRCh38
rs938910397 Chr.1: 180166501-180166501 on Build GRCh38
rs1045162605 Chr.1: 180166808-180166808 on Build GRCh38
rs376417038 Chr.1: 180166564-180166564 on Build GRCh38
rs748983674 Chr.1: 180166435-180166435 on Build GRCh38
rs748443918 Chr.1: 180166544-180166544 on Build GRCh38
rs375285118 Chr.1: 180166523-180166523 on Build GRCh38
rs955705755 Chr.1: 180166590-180166590 on Build GRCh38
rs532669855 Chr.1: 180166469-180166469 on Build GRCh38
rs772245000 Chr.1: 180166449-180166449 on Build GRCh38
rs759933269 Chr.1: 180166582-180166582 on Build GRCh38
rs373887690 Chr.1: 180166765-180166765 on Build GRCh38
rs912277315 Chr.1: 180166407-180166407 on Build GRCh38
rs940720515 Chr.1: 180166761-180166761 on Build GRCh38
rs905007015 Chr.1: 180166546-180166546 on Build GRCh38
rs201825048 Chr.1: 180166517-180166517 on Build GRCh38
rs947559266 Chr.1: 180166436-180166436 on Build GRCh38
rs372037235 Chr.1: 180166505-180166505 on Build GRCh38
rs1008576333 Chr.1: 180166833-180166833 on Build GRCh38
rs779969740 Chr.1: 180166532-180166532 on Build GRCh38
rs914310935 Chr.1: 180166430-180166430 on Build GRCh38
rs3894211 Chr.1: 180166566-180166566 on Build GRCh38
rs746532674 Chr.1: 180166500-180166500 on Build GRCh38
rs778980035 Chr.1: 180166542-180166542 on Build GRCh38
rs535096387 Chr.1: 180166673-180166673 on Build GRCh38
rs10715769 Chr.1: 180166623-180166623 on Build GRCh38
rs975131923 Chr.1: 180166735-180166735 on Build GRCh38
rs947112974 Chr.1: 180166624-180166624 on Build GRCh38
rs906141564 Chr.1: 180166467-180166467 on Build GRCh38
rs988579268 Chr.1: 180166796-180166796 on Build GRCh38
rs922096354 Chr.1: 180166474-180166474 on Build GRCh38
rs931112746 Chr.1: 180166816-180166816 on Build GRCh38
rs905041443 Chr.1: 180166814-180166814 on Build GRCh38
rs369738822 Chr.1: 180166609-180166609 on Build GRCh38
rs774585941 Chr.1: 180166444-180166444 on Build GRCh38
rs367678301 Chr.1: 180166516-180166516 on Build GRCh38
rs980273000 Chr.1: 180166667-180166667 on Build GRCh38
rs979725878 Chr.1: 180166747-180166747 on Build GRCh38
rs935353592 Chr.1: 180166785-180166785 on Build GRCh38
rs1019369138 Chr.1: 180166654-180166654 on Build GRCh38
rs1004249475 Chr.1: 180166653-180166653 on Build GRCh38
rs949470214 Chr.1: 180166804-180166804 on Build GRCh38
rs996184022 Chr.1: 180166782-180166782 on Build GRCh38
rs917920230 Chr.1: 180166798-180166798 on Build GRCh38
rs761312198 Chr.1: 180166470-180166470 on Build GRCh38
rs114307591 Chr.1: 180166737-180166737 on Build GRCh38
rs1057441838 Chr.1: 180166509-180166509 on Build GRCh38
rs773399208 Chr.1: 180166550-180166550 on Build GRCh38
rs777314092 Chr.1: 180166567-180166567 on Build GRCh38
rs375407780 Chr.1: 180166514-180166514 on Build GRCh38
rs886679030 Chr.1: 180166752-180166752 on Build GRCh38
rs899291779 Chr.1: 180166774-180166774 on Build GRCh38
rs927442829 Chr.1: 180166706-180166706 on Build GRCh38
rs773563910 Chr.1: 180166463-180166463 on Build GRCh38
rs149269903 Chr.1: 180166537-180166537 on Build GRCh38
rs757094993 Chr.1: 180166685-180166686 on Build GRCh38
rs761939311 Chr.1: 180166570-180166570 on Build GRCh38

View Full Product Details