Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 92404393-92404892 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CENPP
Entrez Gene ID
401541
Gene Name
centromere protein P
Gene Aliases
CENP-P
UniGene
Hs.713775
Gene Chromosome Location
Chr.9: 92325457-92615164 on Build GRCh38
Transcripts
NM_001012267.2 NM_001286969.1 XM_011518689.1 NM_014057.4 NM_024416.4 NM_033014.3

Gene Symbol
OGN
Entrez Gene ID
4969
Gene Name
osteoglycin
Gene Aliases
OG, OIF, SLRR3A
UniGene
Hs.109439
Gene Chromosome Location
Chr.9: 92383271-92404699 on Build GRCh38
Transcripts
NM_001012267.2 NM_001286969.1 XM_011518689.1 NM_014057.4 NM_024416.4 NM_033014.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6359080 c.145A>G p.R49G Chr.9: 92404531-92404531 on Build GRCh38
6842331 c.103C>A p.P35T Chr.9: 92404573-92404573 on Build GRCh38
5573321 c.46C>T p.L16L Chr.9: 92404630-92404630 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs374524047 Chr.9: 92404454-92404454 on Build GRCh38
rs368437420 Chr.9: 92404497-92404497 on Build GRCh38
rs145618993 Chr.9: 92404575-92404575 on Build GRCh38
rs745538020 Chr.9: 92404490-92404490 on Build GRCh38
rs770685642 Chr.9: 92404763-92404763 on Build GRCh38
rs777788718 Chr.9: 92404715-92404715 on Build GRCh38
rs900730096 Chr.9: 92404775-92404775 on Build GRCh38
rs771461826 Chr.9: 92404496-92404496 on Build GRCh38
rs905187007 Chr.9: 92404426-92404426 on Build GRCh38
rs570038698 Chr.9: 92404694-92404694 on Build GRCh38
rs571426340 Chr.9: 92404477-92404477 on Build GRCh38
rs371854947 Chr.9: 92404472-92404472 on Build GRCh38
rs532655835 Chr.9: 92404521-92404521 on Build GRCh38
rs750617350 Chr.9: 92404584-92404584 on Build GRCh38
rs1030463507 Chr.9: 92404515-92404515 on Build GRCh38
rs550016368 Chr.9: 92404448-92404448 on Build GRCh38
rs200929817 Chr.9: 92404644-92404644 on Build GRCh38
rs1041656558 Chr.9: 92404812-92404812 on Build GRCh38
rs764602148 Chr.9: 92404528-92404528 on Build GRCh38
rs1005916238 Chr.9: 92404443-92404443 on Build GRCh38
rs966890633 Chr.9: 92404434-92404434 on Build GRCh38
rs751723917 Chr.9: 92404674-92404674 on Build GRCh38
rs972315288 Chr.9: 92404499-92404499 on Build GRCh38
rs372381648 Chr.9: 92404506-92404506 on Build GRCh38
rs773731872 Chr.9: 92404476-92404476 on Build GRCh38
rs918406060 Chr.9: 92404559-92404559 on Build GRCh38
rs1039792484 Chr.9: 92404771-92404771 on Build GRCh38
rs1055532807 Chr.9: 92404830-92404830 on Build GRCh38
rs181807087 Chr.9: 92404535-92404535 on Build GRCh38
rs1052043711 Chr.9: 92404827-92404827 on Build GRCh38
rs780227176 Chr.9: 92404447-92404447 on Build GRCh38
rs554929551 Chr.9: 92404652-92404652 on Build GRCh38
rs375718621 Chr.9: 92404593-92404593 on Build GRCh38
rs902673284 Chr.9: 92404832-92404832 on Build GRCh38
rs1032873898 Chr.9: 92404450-92404450 on Build GRCh38
rs749271546 Chr.9: 92404721-92404721 on Build GRCh38
rs566156456 Chr.9: 92404567-92404567 on Build GRCh38
rs746576779 Chr.9: 92404576-92404576 on Build GRCh38
rs938545098 Chr.9: 92404820-92404820 on Build GRCh38
rs537316851 Chr.9: 92404700-92404700 on Build GRCh38
rs997657859 Chr.9: 92404421-92404421 on Build GRCh38
rs368163044 Chr.9: 92404432-92404432 on Build GRCh38
rs866408105 Chr.9: 92404543-92404543 on Build GRCh38
rs907030273 Chr.9: 92404814-92404814 on Build GRCh38
rs1000140589 Chr.9: 92404846-92404846 on Build GRCh38
rs771726745 Chr.9: 92404699-92404699 on Build GRCh38
rs561053033 Chr.9: 92404418-92404418 on Build GRCh38
rs997747534 Chr.9: 92404803-92404803 on Build GRCh38
rs761073661 Chr.9: 92404524-92404524 on Build GRCh38

View Full Product Details