Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 93659428-93659932 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PHF2
Entrez Gene ID
5253
Gene Name
PHD finger protein 2
Gene Aliases
CENP-35, GRC5, JHDM1E
UniGene
Hs.211441
Gene Chromosome Location
Chr.9: 93576627-93679587 on Build GRCh38
Transcripts
NM_005392.3 XM_005252051.2 XM_006717143.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1686590 c.1329+1G>A p.? Chr.9: 93659601-93659601 on Build GRCh38
5788374 c.1291C>G p.L431V Chr.9: 93659562-93659562 on Build GRCh38
6184396 c.1327G>T p.E443* Chr.9: 93659598-93659598 on Build GRCh38
2732601 c.1271A>T p.E424V Chr.9: 93659542-93659542 on Build GRCh38
7088987 c.1269G>T p.P423P Chr.9: 93659540-93659540 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs936021772 Chr.9: 93659466-93659467 on Build GRCh38
rs201365575 Chr.9: 93659497-93659497 on Build GRCh38
rs564406055 Chr.9: 93659608-93659608 on Build GRCh38
rs576390467 Chr.9: 93659608-93659609 on Build GRCh38
rs10992835 Chr.9: 93659746-93659746 on Build GRCh38
rs1001891586 Chr.9: 93659646-93659646 on Build GRCh38
rs779475781 Chr.9: 93659543-93659543 on Build GRCh38
rs75817819 Chr.9: 93659760-93659760 on Build GRCh38
rs968784210 Chr.9: 93659835-93659835 on Build GRCh38
rs935971992 Chr.9: 93659870-93659870 on Build GRCh38
rs767315339 Chr.9: 93659851-93659851 on Build GRCh38
rs778647391 Chr.9: 93659531-93659531 on Build GRCh38
rs766602223 Chr.9: 93659652-93659652 on Build GRCh38
rs1004557358 Chr.9: 93659741-93659741 on Build GRCh38
rs78254907 Chr.9: 93659549-93659549 on Build GRCh38
rs763124523 Chr.9: 93659617-93659617 on Build GRCh38
rs1046654017 Chr.9: 93659474-93659474 on Build GRCh38
rs1045565663 Chr.9: 93659552-93659552 on Build GRCh38
rs768062608 Chr.9: 93659721-93659721 on Build GRCh38
rs756982986 Chr.9: 93659528-93659528 on Build GRCh38
rs893883976 Chr.9: 93659655-93659655 on Build GRCh38
rs777210002 Chr.9: 93659579-93659579 on Build GRCh38
rs759357196 Chr.9: 93659481-93659481 on Build GRCh38
rs373310830 Chr.9: 93659492-93659492 on Build GRCh38
rs965304344 Chr.9: 93659828-93659828 on Build GRCh38
rs566594128 Chr.9: 93659551-93659551 on Build GRCh38
rs751408198 Chr.9: 93659469-93659469 on Build GRCh38
rs755659409 Chr.9: 93659720-93659720 on Build GRCh38
rs1019529564 Chr.9: 93659808-93659808 on Build GRCh38
rs75271573 Chr.9: 93659768-93659768 on Build GRCh38
rs764009924 Chr.9: 93659496-93659496 on Build GRCh38
rs758033401 Chr.9: 93659537-93659537 on Build GRCh38
rs1053144670 Chr.9: 93659884-93659884 on Build GRCh38
rs3750357 Chr.9: 93659837-93659837 on Build GRCh38
rs368004083 Chr.9: 93659705-93659705 on Build GRCh38
rs954319338 Chr.9: 93659803-93659803 on Build GRCh38
rs745468234 Chr.9: 93659535-93659535 on Build GRCh38
rs922801072 Chr.9: 93659775-93659775 on Build GRCh38
rs769353039 Chr.9: 93659554-93659554 on Build GRCh38
rs551556463 Chr.9: 93659454-93659454 on Build GRCh38
rs1033716056 Chr.9: 93659581-93659581 on Build GRCh38
rs751627896 Chr.9: 93659485-93659485 on Build GRCh38
rs951762009 Chr.9: 93659735-93659735 on Build GRCh38
rs1027319176 Chr.9: 93659689-93659689 on Build GRCh38
rs748535292 Chr.9: 93659590-93659590 on Build GRCh38
rs926234086 Chr.9: 93659847-93659847 on Build GRCh38
rs766331357 Chr.9: 93659483-93659483 on Build GRCh38
rs767552905 Chr.9: 93659506-93659506 on Build GRCh38
rs371223458 Chr.9: 93659747-93659747 on Build GRCh38
rs752131988 Chr.9: 93659609-93659609 on Build GRCh38
rs773248774 Chr.9: 93659468-93659468 on Build GRCh38

View Full Product Details