Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 109137109-109137635 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRRS1L
Entrez Gene ID
23732
Gene Name
ferric chelate reductase 1 like
Gene Aliases
C9orf4, CG-6, CG6, EIEE37
UniGene
Hs.347537
Gene Chromosome Location
Chr.9: 109136188-109167291 on Build GRCh38
Transcripts
NM_014334.2 XM_011518453.2 XM_011518454.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6048377 c.1032delC p.*345fs? Chr.9: 109137458-109137458 on Build GRCh38
3652721 c.898G>A p.A300T Chr.9: 109137592-109137592 on Build GRCh38
6182105 c.950C>G p.S317* Chr.9: 109137540-109137540 on Build GRCh38
3432753 c.904G>A p.E302K Chr.9: 109137586-109137586 on Build GRCh38
5893366 c.956C>T p.A319V Chr.9: 109137534-109137534 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781543300 Chr.9: 109137596-109137596 on Build GRCh38
rs369193985 Chr.9: 109137582-109137582 on Build GRCh38
rs1052187481 Chr.9: 109137384-109137384 on Build GRCh38
rs918636357 Chr.9: 109137223-109137224 on Build GRCh38
rs762363721 Chr.9: 109137411-109137411 on Build GRCh38
rs769371724 Chr.9: 109137520-109137520 on Build GRCh38
rs772724279 Chr.9: 109137525-109137525 on Build GRCh38
rs878853280 Chr.9: 109137529-109137529 on Build GRCh38
rs907869687 Chr.9: 109137562-109137562 on Build GRCh38
rs774372717 Chr.9: 109137542-109137542 on Build GRCh38
rs947952161 Chr.9: 109137338-109137338 on Build GRCh38
rs747814314 Chr.9: 109137519-109137519 on Build GRCh38
rs760544545 Chr.9: 109137568-109137568 on Build GRCh38
rs751429704 Chr.9: 109137420-109137420 on Build GRCh38
rs957136038 Chr.9: 109137252-109137252 on Build GRCh38
rs779958643 Chr.9: 109137583-109137583 on Build GRCh38
rs1018262344 Chr.9: 109137543-109137543 on Build GRCh38
rs930729318 Chr.9: 109137473-109137473 on Build GRCh38
rs781488129 Chr.9: 109137508-109137508 on Build GRCh38
rs766395815 Chr.9: 109137415-109137415 on Build GRCh38
rs753180568 Chr.9: 109137561-109137561 on Build GRCh38
rs563101375 Chr.9: 109137556-109137556 on Build GRCh38
rs146108071 Chr.9: 109137339-109137339 on Build GRCh38
rs766816520 Chr.9: 109137366-109137366 on Build GRCh38
rs777520519 Chr.9: 109137606-109137606 on Build GRCh38
rs759443940 Chr.9: 109137428-109137428 on Build GRCh38
rs962805249 Chr.9: 109137200-109137200 on Build GRCh38
rs377132738 Chr.9: 109137464-109137464 on Build GRCh38
rs777150168 Chr.9: 109137258-109137258 on Build GRCh38
rs765555791 Chr.9: 109137577-109137577 on Build GRCh38
rs778859840 Chr.9: 109137454-109137454 on Build GRCh38
rs982813211 Chr.9: 109137348-109137348 on Build GRCh38
rs750592242 Chr.9: 109137580-109137580 on Build GRCh38
rs537102049 Chr.9: 109137212-109137212 on Build GRCh38
rs185774761 Chr.9: 109137223-109137223 on Build GRCh38
rs990969750 Chr.9: 109137400-109137400 on Build GRCh38
rs746604659 Chr.9: 109137357-109137357 on Build GRCh38
rs759702689 Chr.9: 109137594-109137594 on Build GRCh38
rs374679996 Chr.9: 109137442-109137442 on Build GRCh38
rs751914199 Chr.9: 109137522-109137522 on Build GRCh38
rs750586857 Chr.9: 109137461-109137461 on Build GRCh38
rs757494229 Chr.9: 109137453-109137453 on Build GRCh38
rs917854339 Chr.9: 109137226-109137226 on Build GRCh38
rs76521520 Chr.9: 109137586-109137586 on Build GRCh38
rs900408626 Chr.9: 109137584-109137584 on Build GRCh38
rs369428108 Chr.9: 109137605-109137605 on Build GRCh38
rs201919050 Chr.9: 109137467-109137467 on Build GRCh38
rs576180163 Chr.9: 109137169-109137169 on Build GRCh38
rs772722857 Chr.9: 109137407-109137407 on Build GRCh38
rs936289277 Chr.9: 109137381-109137381 on Build GRCh38
rs77696081 Chr.9: 109137341-109137341 on Build GRCh38
rs200221527 Chr.9: 109137554-109137554 on Build GRCh38
rs372303605 Chr.9: 109137436-109137436 on Build GRCh38
rs371581339 Chr.9: 109137588-109137588 on Build GRCh38
rs752083667 Chr.9: 109137593-109137593 on Build GRCh38
rs201416713 Chr.9: 109137553-109137553 on Build GRCh38
rs959558899 Chr.9: 109137393-109137393 on Build GRCh38
rs199731037 Chr.9: 109137597-109137597 on Build GRCh38
rs79316984 Chr.9: 109137174-109137174 on Build GRCh38
rs118149760 Chr.9: 109137191-109137191 on Build GRCh38
rs992377440 Chr.9: 109137224-109137224 on Build GRCh38
rs748851583 Chr.9: 109137609-109137609 on Build GRCh38
rs1047774492 Chr.9: 109137499-109137499 on Build GRCh38
rs867249635 Chr.9: 109137573-109137573 on Build GRCh38
rs374121699 Chr.9: 109137610-109137610 on Build GRCh38
rs773837729 Chr.9: 109137138-109137138 on Build GRCh38
rs570300260 Chr.9: 109137476-109137476 on Build GRCh38
rs551792076 Chr.9: 109137458-109137458 on Build GRCh38
rs573580811 Chr.9: 109137284-109137284 on Build GRCh38
rs550427304 Chr.9: 109137160-109137160 on Build GRCh38
rs769040844 Chr.9: 109137477-109137477 on Build GRCh38

View Full Product Details