Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 70277220-70277731 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMC5
Entrez Gene ID
23137
Gene Name
structural maintenance of chromosomes 5
Gene Aliases
SMC5L1
UniGene
Hs.534189
Gene Chromosome Location
Chr.9: 70258962-70354873 on Build GRCh38
Transcripts
NM_015110.3 XM_005251837.1 XM_005251838.1 XM_005251839.1 XM_017014507.1 XM_017014508.1 XM_017014509.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1109761 c.436C>T p.Q146* Chr.9: 70277365-70277365 on Build GRCh38
2779063 c.400C>T p.L134F Chr.9: 70277329-70277329 on Build GRCh38
1674520 c.539C>A p.P180H Chr.9: 70277468-70277468 on Build GRCh38
487525 c.541C>T p.Q181* Chr.9: 70277470-70277470 on Build GRCh38
6785243 c.451A>G p.I151V Chr.9: 70277380-70277380 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748717368 Chr.9: 70277379-70277379 on Build GRCh38
rs370425906 Chr.9: 70277452-70277452 on Build GRCh38
rs781411396 Chr.9: 70277363-70277363 on Build GRCh38
rs768925921 Chr.9: 70277360-70277360 on Build GRCh38
rs181453254 Chr.9: 70277466-70277466 on Build GRCh38
rs933671020 Chr.9: 70277575-70277575 on Build GRCh38
rs1005824245 Chr.9: 70277337-70277337 on Build GRCh38
rs751891954 Chr.9: 70277490-70277490 on Build GRCh38
rs778612425 Chr.9: 70277341-70277341 on Build GRCh38
rs116486194 Chr.9: 70277335-70277335 on Build GRCh38
rs747986719 Chr.9: 70277358-70277404 on Build GRCh38
rs143361396 Chr.9: 70277545-70277545 on Build GRCh38
rs773106566 Chr.9: 70277604-70277604 on Build GRCh38
rs185892897 Chr.9: 70277665-70277665 on Build GRCh38
rs1016767897 Chr.9: 70277362-70277362 on Build GRCh38
rs117986796 Chr.9: 70277628-70277628 on Build GRCh38
rs994064412 Chr.9: 70277656-70277656 on Build GRCh38
rs200628909 Chr.9: 70277489-70277489 on Build GRCh38
rs775058406 Chr.9: 70277437-70277437 on Build GRCh38
rs753829381 Chr.9: 70277332-70277332 on Build GRCh38
rs552208885 Chr.9: 70277267-70277267 on Build GRCh38
rs199803332 Chr.9: 70277510-70277510 on Build GRCh38
rs759152253 Chr.9: 70277280-70277280 on Build GRCh38
rs889575439 Chr.9: 70277585-70277585 on Build GRCh38
rs879130183 Chr.9: 70277382-70277382 on Build GRCh38
rs913364225 Chr.9: 70277513-70277513 on Build GRCh38
rs758566983 Chr.9: 70277344-70277344 on Build GRCh38
rs377344787 Chr.9: 70277348-70277348 on Build GRCh38
rs562157189 Chr.9: 70277698-70277698 on Build GRCh38
rs1024148167 Chr.9: 70277667-70277667 on Build GRCh38
rs1049402905 Chr.9: 70277577-70277577 on Build GRCh38
rs1042082954 Chr.9: 70277268-70277268 on Build GRCh38
rs763588735 Chr.9: 70277327-70277327 on Build GRCh38
rs994013522 Chr.9: 70277394-70277394 on Build GRCh38
rs760306031 Chr.9: 70277446-70277446 on Build GRCh38
rs781466644 Chr.9: 70277511-70277511 on Build GRCh38
rs758143564 Chr.9: 70277488-70277488 on Build GRCh38
rs763641956 Chr.9: 70277451-70277451 on Build GRCh38
rs770307620 Chr.9: 70277392-70277392 on Build GRCh38
rs554882406 Chr.9: 70277458-70277458 on Build GRCh38
rs972228456 Chr.9: 70277609-70277609 on Build GRCh38
rs763328847 Chr.9: 70277408-70277408 on Build GRCh38
rs73647452 Chr.9: 70277425-70277425 on Build GRCh38
rs189966832 Chr.9: 70277700-70277700 on Build GRCh38
rs760122316 Chr.9: 70277325-70277325 on Build GRCh38
rs922274596 Chr.9: 70277550-70277550 on Build GRCh38
rs141041598 Chr.9: 70277534-70277534 on Build GRCh38
rs997951844 Chr.9: 70277315-70277315 on Build GRCh38
rs747088047 Chr.9: 70277358-70277358 on Build GRCh38
rs560332260 Chr.9: 70277624-70277624 on Build GRCh38
rs773480446 Chr.9: 70277401-70277401 on Build GRCh38
rs376102908 Chr.9: 70277518-70277518 on Build GRCh38
rs941112830 Chr.9: 70277586-70277586 on Build GRCh38
rs752228614 Chr.9: 70277297-70277297 on Build GRCh38
rs370543010 Chr.9: 70277548-70277548 on Build GRCh38
rs201393554 Chr.9: 70277295-70277295 on Build GRCh38
rs376489062 Chr.9: 70277286-70277286 on Build GRCh38

View Full Product Details