Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 74722995-74723501 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRPM6
Entrez Gene ID
140803
Gene Name
transient receptor potential cation channel subfamily M member 6
Gene Aliases
CHAK2, HMGX, HOMG, HOMG1, HSH
UniGene
Hs.272225
Gene Chromosome Location
Chr.9: 74722495-74888094 on Build GRCh38
Transcripts
NM_001177310.1 NM_001177311.1 NM_017662.4

SNP Detail

SNP ID Chromosome Location
rs765315092 Chr.9: 74723302-74723302 on Build GRCh38
rs768075146 Chr.9: 74723086-74723086 on Build GRCh38
rs773269259 Chr.9: 74723310-74723310 on Build GRCh38
rs563649179 Chr.9: 74723453-74723453 on Build GRCh38
rs867921317 Chr.9: 74723097-74723097 on Build GRCh38
rs34062320 Chr.9: 74723041-74723042 on Build GRCh38
rs1028918074 Chr.9: 74723334-74723334 on Build GRCh38
rs555877648 Chr.9: 74723291-74723291 on Build GRCh38
rs1008722269 Chr.9: 74723370-74723370 on Build GRCh38
rs1057515629 Chr.9: 74723124-74723124 on Build GRCh38
rs923954574 Chr.9: 74723222-74723222 on Build GRCh38
rs190131362 Chr.9: 74723391-74723391 on Build GRCh38
rs776169040 Chr.9: 74723214-74723214 on Build GRCh38
rs766139590 Chr.9: 74723318-74723318 on Build GRCh38
rs549594058 Chr.9: 74723183-74723183 on Build GRCh38
rs978099441 Chr.9: 74723164-74723164 on Build GRCh38
rs1005065343 Chr.9: 74723317-74723317 on Build GRCh38
rs752883751 Chr.9: 74723368-74723368 on Build GRCh38
rs138021809 Chr.9: 74723386-74723386 on Build GRCh38
rs912641987 Chr.9: 74723140-74723140 on Build GRCh38
rs553610449 Chr.9: 74723215-74723215 on Build GRCh38
rs542368501 Chr.9: 74723262-74723262 on Build GRCh38
rs953201414 Chr.9: 74723124-74723124 on Build GRCh38
rs1028798294 Chr.9: 74723076-74723076 on Build GRCh38
rs762845077 Chr.9: 74723316-74723316 on Build GRCh38
rs150358241 Chr.9: 74723252-74723252 on Build GRCh38
rs994731404 Chr.9: 74723333-74723333 on Build GRCh38
rs987340856 Chr.9: 74723131-74723131 on Build GRCh38
rs898840033 Chr.9: 74723319-74723319 on Build GRCh38
rs761376874 Chr.9: 74723233-74723233 on Build GRCh38
rs978592283 Chr.9: 74723407-74723407 on Build GRCh38
rs944132805 Chr.9: 74723158-74723158 on Build GRCh38

View Full Product Details