Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 83000078-83000578 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RASEF
Entrez Gene ID
158158
Gene Name
RAS and EF-hand domain containing
Gene Aliases
RAB45
UniGene
Hs.129136
Gene Chromosome Location
Chr.9: 82979585-83063177 on Build GRCh38
Transcripts
NM_152573.3 XM_005251730.3 XM_005251731.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4468625 c.1553C>T p.P518L Chr.9: 83000455-83000455 on Build GRCh38
5835795 c.1693_1695delGAA p.E565delE Chr.9: 83000197-83000199 on Build GRCh38
5831152 c.1576-3delC p.? Chr.9: 83000319-83000319 on Build GRCh38
6574851 c.1547G>C p.R516T Chr.9: 83000461-83000461 on Build GRCh38
2153127 c.1666A>G p.S556G Chr.9: 83000226-83000226 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759408626 Chr.9: 83000192-83000192 on Build GRCh38
rs775469651 Chr.9: 83000440-83000440 on Build GRCh38
rs754185855 Chr.9: 83000208-83000208 on Build GRCh38
rs933978393 Chr.9: 83000467-83000467 on Build GRCh38
rs149786197 Chr.9: 83000244-83000244 on Build GRCh38
rs892046140 Chr.9: 83000205-83000205 on Build GRCh38
rs755043038 Chr.9: 83000409-83000409 on Build GRCh38
rs747178297 Chr.9: 83000162-83000162 on Build GRCh38
rs368221419 Chr.9: 83000238-83000238 on Build GRCh38
rs767586579 Chr.9: 83000413-83000413 on Build GRCh38
rs60997315 Chr.9: 83000534-83000534 on Build GRCh38
rs748356184 Chr.9: 83000419-83000419 on Build GRCh38
rs551669693 Chr.9: 83000159-83000159 on Build GRCh38
rs1003636480 Chr.9: 83000289-83000289 on Build GRCh38
rs761384258 Chr.9: 83000351-83000351 on Build GRCh38
rs1040462595 Chr.9: 83000480-83000480 on Build GRCh38
rs375201668 Chr.9: 83000226-83000226 on Build GRCh38
rs745668150 Chr.9: 83000507-83000507 on Build GRCh38
rs541159158 Chr.9: 83000483-83000483 on Build GRCh38
rs756332858 Chr.9: 83000501-83000501 on Build GRCh38
rs749689537 Chr.9: 83000129-83000129 on Build GRCh38
rs185100986 Chr.9: 83000296-83000296 on Build GRCh38
rs760234455 Chr.9: 83000341-83000341 on Build GRCh38
rs759442725 Chr.9: 83000232-83000233 on Build GRCh38
rs746898942 Chr.9: 83000521-83000521 on Build GRCh38
rs768844396 Chr.9: 83000319-83000319 on Build GRCh38
rs939676748 Chr.9: 83000105-83000107 on Build GRCh38
rs774280574 Chr.9: 83000184-83000184 on Build GRCh38
rs765588382 Chr.9: 83000353-83000353 on Build GRCh38
rs767597750 Chr.9: 83000482-83000482 on Build GRCh38
rs569521838 Chr.9: 83000163-83000163 on Build GRCh38
rs573773265 Chr.9: 83000474-83000474 on Build GRCh38
rs761839418 Chr.9: 83000347-83000347 on Build GRCh38
rs761415285 Chr.9: 83000476-83000476 on Build GRCh38
rs1020615901 Chr.9: 83000213-83000213 on Build GRCh38
rs917002886 Chr.9: 83000150-83000150 on Build GRCh38
rs559147700 Chr.9: 83000143-83000143 on Build GRCh38
rs747135098 Chr.9: 83000262-83000262 on Build GRCh38
rs781630047 Chr.9: 83000272-83000272 on Build GRCh38
rs748326357 Chr.9: 83000291-83000291 on Build GRCh38
rs558434027 Chr.9: 83000369-83000369 on Build GRCh38
rs374305065 Chr.9: 83000429-83000429 on Build GRCh38
rs199630496 Chr.9: 83000401-83000401 on Build GRCh38
rs77070682 Chr.9: 83000447-83000447 on Build GRCh38
rs778068076 Chr.9: 83000128-83000128 on Build GRCh38
rs748535689 Chr.9: 83000102-83000102 on Build GRCh38
rs534738577 Chr.9: 83000252-83000252 on Build GRCh38
rs927534480 Chr.9: 83000436-83000436 on Build GRCh38
rs770704145 Chr.9: 83000295-83000295 on Build GRCh38
rs911277203 Chr.9: 83000526-83000526 on Build GRCh38
rs769944554 Chr.9: 83000420-83000420 on Build GRCh38
rs777910876 Chr.9: 83000503-83000503 on Build GRCh38
rs772033799 Chr.9: 83000323-83000323 on Build GRCh38
rs753462861 Chr.9: 83000348-83000348 on Build GRCh38
rs760608840 Chr.9: 83000441-83000441 on Build GRCh38
rs369961860 Chr.9: 83000414-83000414 on Build GRCh38
rs761486019 Chr.9: 83000449-83000449 on Build GRCh38
rs200343613 Chr.9: 83000206-83000206 on Build GRCh38
rs114266938 Chr.9: 83000176-83000176 on Build GRCh38
rs202211776 Chr.9: 83000179-83000179 on Build GRCh38
rs144744360 Chr.9: 83000347-83000347 on Build GRCh38
rs773119570 Chr.9: 83000171-83000171 on Build GRCh38
rs768095672 Chr.9: 83000445-83000445 on Build GRCh38
rs745929363 Chr.9: 83000152-83000152 on Build GRCh38
rs372119351 Chr.9: 83000217-83000217 on Build GRCh38
rs779836440 Chr.9: 83000520-83000520 on Build GRCh38
rs961271838 Chr.9: 83000325-83000325 on Build GRCh38
rs766969065 Chr.9: 83000199-83000199 on Build GRCh38
rs983934371 Chr.9: 83000431-83000431 on Build GRCh38
rs979752853 Chr.9: 83000432-83000432 on Build GRCh38
rs539719248 Chr.9: 83000354-83000354 on Build GRCh38
rs780765472 Chr.9: 83000405-83000405 on Build GRCh38
rs1055041928 Chr.9: 83000165-83000165 on Build GRCh38
rs752231088 Chr.9: 83000203-83000203 on Build GRCh38
rs577622126 Chr.9: 83000151-83000151 on Build GRCh38
rs533038973 Chr.9: 83000149-83000149 on Build GRCh38
rs775724799 Chr.9: 83000160-83000160 on Build GRCh38
rs745694312 Chr.9: 83000430-83000430 on Build GRCh38
rs758119764 Chr.9: 83000519-83000519 on Build GRCh38
rs766225247 Chr.9: 83000178-83000178 on Build GRCh38
rs771982315 Chr.9: 83000439-83000439 on Build GRCh38
rs751687989 Chr.9: 83000408-83000408 on Build GRCh38
rs147216417 Chr.9: 83000462-83000462 on Build GRCh38
rs768611266 Chr.9: 83000527-83000527 on Build GRCh38
rs139664099 Chr.9: 83000470-83000470 on Build GRCh38
rs752057655 Chr.9: 83000473-83000473 on Build GRCh38
rs147383713 Chr.9: 83000212-83000212 on Build GRCh38
rs1036658725 Chr.9: 83000108-83000108 on Build GRCh38
rs759481478 Chr.9: 83000310-83000310 on Build GRCh38
rs774577938 Chr.9: 83000318-83000319 on Build GRCh38

View Full Product Details