Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 136009178-136009679 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NACC2
Entrez Gene ID
138151
Gene Name
NACC family member 2
Gene Aliases
BEND9, BTBD14, BTBD14A, BTBD31, NAC-2, RBB
UniGene
Hs.112895
Gene Chromosome Location
Chr.9: 136006537-136095285 on Build GRCh38
Transcripts
NM_144653.4 XM_011518221.2

SNP Detail

SNP ID Chromosome Location
rs573156054 Chr.9: 136009583-136009583 on Build GRCh38
rs955553476 Chr.9: 136009564-136009564 on Build GRCh38
rs935685709 Chr.9: 136009395-136009395 on Build GRCh38
rs537512352 Chr.9: 136009457-136009457 on Build GRCh38
rs932733570 Chr.9: 136009633-136009633 on Build GRCh38
rs774416331 Chr.9: 136009264-136009264 on Build GRCh38
rs7848231 Chr.9: 136009473-136009473 on Build GRCh38
rs887215361 Chr.9: 136009294-136009294 on Build GRCh38
rs922343904 Chr.9: 136009355-136009355 on Build GRCh38
rs368014583 Chr.9: 136009626-136009626 on Build GRCh38
rs984745725 Chr.9: 136009319-136009319 on Build GRCh38
rs964483940 Chr.9: 136009628-136009628 on Build GRCh38
rs553399380 Chr.9: 136009528-136009528 on Build GRCh38
rs565302442 Chr.9: 136009627-136009627 on Build GRCh38
rs758074494 Chr.9: 136009496-136009496 on Build GRCh38
rs907470404 Chr.9: 136009562-136009562 on Build GRCh38
rs866644332 Chr.9: 136009548-136009548 on Build GRCh38
rs910974971 Chr.9: 136009599-136009599 on Build GRCh38
rs12376616 Chr.9: 136009203-136009203 on Build GRCh38
rs530095657 Chr.9: 136009292-136009292 on Build GRCh38
rs188412228 Chr.9: 136009244-136009244 on Build GRCh38
rs1013369047 Chr.9: 136009546-136009546 on Build GRCh38
rs559394262 Chr.9: 136009517-136009517 on Build GRCh38
rs146384679 Chr.9: 136009458-136009458 on Build GRCh38
rs1048052265 Chr.9: 136009482-136009482 on Build GRCh38
rs891749149 Chr.9: 136009436-136009436 on Build GRCh38
rs956223692 Chr.9: 136009618-136009618 on Build GRCh38
rs117654243 Chr.9: 136009330-136009330 on Build GRCh38
rs10776868 Chr.9: 136009424-136009424 on Build GRCh38
rs1012883233 Chr.9: 136009330-136009331 on Build GRCh38
rs776085496 Chr.9: 136009559-136009559 on Build GRCh38
rs943032801 Chr.9: 136009442-136009442 on Build GRCh38
rs1017189864 Chr.9: 136009322-136009322 on Build GRCh38
rs945968804 Chr.9: 136009647-136009647 on Build GRCh38
rs770214275 Chr.9: 136009646-136009646 on Build GRCh38
rs1010074422 Chr.9: 136009263-136009263 on Build GRCh38
rs898519389 Chr.9: 136009328-136009328 on Build GRCh38
rs935660535 Chr.9: 136009606-136009606 on Build GRCh38
rs972910541 Chr.9: 136009342-136009342 on Build GRCh38
rs766231038 Chr.9: 136009374-136009374 on Build GRCh38
rs1034119456 Chr.9: 136009293-136009293 on Build GRCh38
rs959419203 Chr.9: 136009298-136009298 on Build GRCh38
rs113709326 Chr.9: 136009409-136009409 on Build GRCh38
rs753878775 Chr.9: 136009404-136009404 on Build GRCh38
rs895100118 Chr.9: 136009537-136009537 on Build GRCh38
rs55939904 Chr.9: 136009550-136009550 on Build GRCh38
rs76353438 Chr.9: 136009368-136009368 on Build GRCh38
rs1017597092 Chr.9: 136009543-136009543 on Build GRCh38
rs754430292 Chr.9: 136009405-136009405 on Build GRCh38
rs1029763835 Chr.9: 136009563-136009563 on Build GRCh38
rs1031853366 Chr.9: 136009579-136009579 on Build GRCh38
rs141544649 Chr.9: 136009291-136009291 on Build GRCh38
rs961635946 Chr.9: 136009555-136009555 on Build GRCh38
rs985144516 Chr.9: 136009585-136009585 on Build GRCh38
rs907722779 Chr.9: 136009489-136009489 on Build GRCh38
rs558426383 Chr.9: 136009417-136009417 on Build GRCh38
rs1005590114 Chr.9: 136009542-136009542 on Build GRCh38
rs566768771 Chr.9: 136009367-136009367 on Build GRCh38
rs906596769 Chr.9: 136009474-136009474 on Build GRCh38
rs557057276 Chr.9: 136009419-136009419 on Build GRCh38

View Full Product Details