Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 136805346-136805844 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC183
Entrez Gene ID
84960
Gene Name
coiled-coil domain containing 183
Gene Aliases
KIAA1984, PARF, bA216L13.7
UniGene
Hs.728796
Gene Chromosome Location
Chr.9: 136796338-136807741 on Build GRCh38
Transcripts
NM_001039374.4 NR_024580.1

Gene Symbol
CCDC183-AS1
Entrez Gene ID
100131193
Gene Name
CCDC183 antisense RNA 1
Gene Aliases
KIAA1984-AS1
UniGene
Hs.734812
Gene Chromosome Location
Chr.9: 136803927-136808848 on Build GRCh38
Transcripts
NM_001039374.4 NR_024580.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6694514 c.864C>T p.T288T Chr.9: 136805373-136805373 on Build GRCh38
7242907 c.859G>C p.E287Q Chr.9: 136805368-136805368 on Build GRCh38
6694515 c.871G>A p.A291T Chr.9: 136805380-136805380 on Build GRCh38
1292822 c.948+2T>A p.? Chr.9: 136805459-136805459 on Build GRCh38
3905834 c.894C>T p.G298G Chr.9: 136805403-136805403 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763257000 Chr.9: 136805441-136805441 on Build GRCh38
rs968469287 Chr.9: 136805544-136805544 on Build GRCh38
rs558564967 Chr.9: 136805606-136805606 on Build GRCh38
rs754778017 Chr.9: 136805484-136805484 on Build GRCh38
rs546753937 Chr.9: 136805511-136805511 on Build GRCh38
rs755852432 Chr.9: 136805416-136805416 on Build GRCh38
rs920566075 Chr.9: 136805779-136805779 on Build GRCh38
rs193251202 Chr.9: 136805746-136805746 on Build GRCh38
rs143427692 Chr.9: 136805472-136805472 on Build GRCh38
rs777364183 Chr.9: 136805418-136805418 on Build GRCh38
rs370604674 Chr.9: 136805696-136805696 on Build GRCh38
rs1045557793 Chr.9: 136805673-136805673 on Build GRCh38
rs933149508 Chr.9: 136805619-136805619 on Build GRCh38
rs750015879 Chr.9: 136805471-136805471 on Build GRCh38
rs751995218 Chr.9: 136805449-136805449 on Build GRCh38
rs1038912023 Chr.9: 136805531-136805531 on Build GRCh38
rs114312295 Chr.9: 136805372-136805372 on Build GRCh38
rs370377081 Chr.9: 136805411-136805411 on Build GRCh38
rs1028268580 Chr.9: 136805762-136805762 on Build GRCh38
rs2811793 Chr.9: 136805512-136805512 on Build GRCh38
rs758363905 Chr.9: 136805477-136805477 on Build GRCh38
rs574525599 Chr.9: 136805636-136805636 on Build GRCh38
rs935091701 Chr.9: 136805734-136805734 on Build GRCh38
rs543602452 Chr.9: 136805388-136805388 on Build GRCh38
rs950878429 Chr.9: 136805681-136805681 on Build GRCh38
rs1056170854 Chr.9: 136805616-136805616 on Build GRCh38
rs191696565 Chr.9: 136805387-136805387 on Build GRCh38
rs995417830 Chr.9: 136805653-136805653 on Build GRCh38
rs1056223659 Chr.9: 136805735-136805735 on Build GRCh38
rs1043526215 Chr.9: 136805575-136805575 on Build GRCh38
rs781010319 Chr.9: 136805596-136805596 on Build GRCh38
rs538873126 Chr.9: 136805548-136805548 on Build GRCh38
rs186801997 Chr.9: 136805683-136805683 on Build GRCh38
rs906664358 Chr.9: 136805796-136805796 on Build GRCh38
rs765537117 Chr.9: 136805437-136805437 on Build GRCh38
rs756717959 Chr.9: 136805406-136805406 on Build GRCh38
rs776833927 Chr.9: 136805432-136805432 on Build GRCh38
rs988461828 Chr.9: 136805513-136805513 on Build GRCh38
rs371399315 Chr.9: 136805467-136805467 on Build GRCh38
rs753176746 Chr.9: 136805399-136805399 on Build GRCh38
rs538953467 Chr.9: 136805774-136805774 on Build GRCh38
rs923783515 Chr.9: 136805732-136805732 on Build GRCh38
rs148853646 Chr.9: 136805451-136805451 on Build GRCh38
rs999246233 Chr.9: 136805612-136805612 on Build GRCh38
rs569572297 Chr.9: 136805415-136805415 on Build GRCh38
rs774379290 Chr.9: 136805426-136805426 on Build GRCh38
rs879061777 Chr.9: 136805706-136805706 on Build GRCh38
rs553568735 Chr.9: 136805609-136805609 on Build GRCh38
rs553210227 Chr.9: 136805715-136805715 on Build GRCh38
rs1052525869 Chr.9: 136805814-136805814 on Build GRCh38
rs946629355 Chr.9: 136805719-136805719 on Build GRCh38
rs116796202 Chr.9: 136805791-136805791 on Build GRCh38
rs911918830 Chr.9: 136805687-136805687 on Build GRCh38
rs768688768 Chr.9: 136805425-136805425 on Build GRCh38
rs945388 Chr.9: 136805652-136805652 on Build GRCh38
rs145489052 Chr.9: 136805404-136805404 on Build GRCh38
rs748103938 Chr.9: 136805427-136805427 on Build GRCh38
rs973547972 Chr.9: 136805465-136805465 on Build GRCh38
rs532593881 Chr.9: 136805403-136805403 on Build GRCh38
rs566369632 Chr.9: 136805570-136805570 on Build GRCh38
rs183486038 Chr.9: 136805627-136805627 on Build GRCh38
rs764476364 Chr.9: 136805442-136805442 on Build GRCh38
rs778072949 Chr.9: 136805510-136805510 on Build GRCh38
rs970628643 Chr.9: 136805462-136805462 on Build GRCh38
rs770861059 Chr.9: 136805429-136805429 on Build GRCh38
rs780756372 Chr.9: 136805408-136805408 on Build GRCh38
rs765509416 Chr.9: 136805396-136805396 on Build GRCh38
rs966007266 Chr.9: 136805689-136805689 on Build GRCh38
rs1045145665 Chr.9: 136805795-136805795 on Build GRCh38
rs759677985 Chr.9: 136805436-136805436 on Build GRCh38
rs1044697717 Chr.9: 136805747-136805747 on Build GRCh38
rs746541542 Chr.9: 136805432-136805441 on Build GRCh38
rs77293543 Chr.9: 136805697-136805697 on Build GRCh38
rs946548240 Chr.9: 136805573-136805573 on Build GRCh38
rs779802702 Chr.9: 136805474-136805474 on Build GRCh38
rs898515704 Chr.9: 136805495-136805495 on Build GRCh38
rs530106553 Chr.9: 136805375-136805375 on Build GRCh38
rs1004965301 Chr.9: 136805771-136805771 on Build GRCh38
rs995073130 Chr.9: 136805753-136805753 on Build GRCh38
rs142618065 Chr.9: 136805621-136805621 on Build GRCh38
rs914295856 Chr.9: 136805523-136805523 on Build GRCh38
rs146913980 Chr.9: 136805660-136805660 on Build GRCh38
rs752856020 Chr.9: 136805440-136805440 on Build GRCh38
rs200555293 Chr.9: 136805419-136805419 on Build GRCh38
rs886592724 Chr.9: 136805770-136805770 on Build GRCh38
rs979687110 Chr.9: 136805731-136805731 on Build GRCh38
rs371991716 Chr.9: 136805661-136805661 on Build GRCh38
rs2811794 Chr.9: 136805742-136805742 on Build GRCh38
rs567294486 Chr.9: 136805452-136805452 on Build GRCh38
rs566612863 Chr.9: 136805813-136805813 on Build GRCh38
rs896135357 Chr.9: 136805617-136805617 on Build GRCh38
rs149692353 Chr.9: 136805400-136805400 on Build GRCh38
rs778866812 Chr.9: 136805501-136805501 on Build GRCh38
rs906128367 Chr.9: 136805752-136805752 on Build GRCh38

View Full Product Details