Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 124794938-124795444 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OLFML2A
Entrez Gene ID
169611
Gene Name
olfactomedin like 2A
Gene Aliases
PRO34319
UniGene
Hs.357004
Gene Chromosome Location
Chr.9: 124777158-124814891 on Build GRCh38
Transcripts
NM_182487.3 XM_005251760.4 XM_006716989.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7238891 c.355-3C>T p.? Chr.9: 124795021-124795021 on Build GRCh38
1459959 c.454C>T p.L152L Chr.9: 124795123-124795123 on Build GRCh38
4822343 c.375C>T p.L125L Chr.9: 124795044-124795044 on Build GRCh38
116296 c.366G>A p.M122I Chr.9: 124795035-124795035 on Build GRCh38
3699415 c.360G>A p.Q120Q Chr.9: 124795029-124795029 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760338206 Chr.9: 124795030-124795030 on Build GRCh38
rs779136629 Chr.9: 124795105-124795105 on Build GRCh38
rs760124792 Chr.9: 124795078-124795078 on Build GRCh38
rs776261691 Chr.9: 124795050-124795050 on Build GRCh38
rs753135545 Chr.9: 124795175-124795175 on Build GRCh38
rs965416574 Chr.9: 124795389-124795389 on Build GRCh38
rs1002865617 Chr.9: 124795299-124795299 on Build GRCh38
rs905477272 Chr.9: 124795259-124795259 on Build GRCh38
rs944557808 Chr.9: 124795267-124795267 on Build GRCh38
rs983839337 Chr.9: 124795139-124795139 on Build GRCh38
rs772069218 Chr.9: 124795123-124795123 on Build GRCh38
rs532748588 Chr.9: 124795188-124795188 on Build GRCh38
rs1032748092 Chr.9: 124794965-124794965 on Build GRCh38
rs934570838 Chr.9: 124795336-124795336 on Build GRCh38
rs529442416 Chr.9: 124794975-124794975 on Build GRCh38
rs1046586769 Chr.9: 124795217-124795217 on Build GRCh38
rs769400228 Chr.9: 124795142-124795142 on Build GRCh38
rs10986427 Chr.9: 124795089-124795089 on Build GRCh38
rs373492057 Chr.9: 124795087-124795087 on Build GRCh38
rs779825905 Chr.9: 124795129-124795129 on Build GRCh38
rs761373082 Chr.9: 124795059-124795059 on Build GRCh38
rs767866464 Chr.9: 124795080-124795080 on Build GRCh38
rs910614176 Chr.9: 124795260-124795261 on Build GRCh38
rs746350003 Chr.9: 124795125-124795125 on Build GRCh38
rs969547195 Chr.9: 124795005-124795005 on Build GRCh38
rs954087117 Chr.9: 124795101-124795101 on Build GRCh38
rs749415313 Chr.9: 124795011-124795011 on Build GRCh38
rs998004895 Chr.9: 124795419-124795419 on Build GRCh38
rs377537752 Chr.9: 124794995-124794995 on Build GRCh38
rs745684440 Chr.9: 124794990-124794990 on Build GRCh38
rs893806288 Chr.9: 124795357-124795357 on Build GRCh38
rs976309802 Chr.9: 124795269-124795269 on Build GRCh38
rs557323248 Chr.9: 124795367-124795367 on Build GRCh38
rs34219629 Chr.9: 124795006-124795007 on Build GRCh38
rs764743729 Chr.9: 124795060-124795060 on Build GRCh38
rs58204450 Chr.9: 124795383-124795383 on Build GRCh38
rs745811827 Chr.9: 124795018-124795018 on Build GRCh38
rs758884733 Chr.9: 124795148-124795148 on Build GRCh38
rs776354197 Chr.9: 124795131-124795131 on Build GRCh38
rs767048080 Chr.9: 124795206-124795206 on Build GRCh38
rs772735719 Chr.9: 124795149-124795149 on Build GRCh38
rs200167193 Chr.9: 124795040-124795040 on Build GRCh38
rs775386285 Chr.9: 124795026-124795026 on Build GRCh38
rs771127718 Chr.9: 124795017-124795017 on Build GRCh38
rs113431973 Chr.9: 124795194-124795194 on Build GRCh38
rs768106910 Chr.9: 124795159-124795159 on Build GRCh38
rs202194837 Chr.9: 124795086-124795086 on Build GRCh38
rs761369222 Chr.9: 124795143-124795145 on Build GRCh38
rs916977448 Chr.9: 124795195-124795195 on Build GRCh38
rs1021014404 Chr.9: 124795371-124795371 on Build GRCh38
rs762351331 Chr.9: 124795156-124795156 on Build GRCh38
rs142940447 Chr.9: 124795021-124795021 on Build GRCh38
rs531828500 Chr.9: 124795083-124795083 on Build GRCh38
rs373445727 Chr.9: 124794977-124794977 on Build GRCh38
rs781624840 Chr.9: 124794982-124794982 on Build GRCh38
rs537002088 Chr.9: 124795319-124795319 on Build GRCh38
rs1054163117 Chr.9: 124795345-124795345 on Build GRCh38
rs745903529 Chr.9: 124795122-124795122 on Build GRCh38
rs749871190 Chr.9: 124795066-124795066 on Build GRCh38
rs1044186702 Chr.9: 124795413-124795413 on Build GRCh38
rs376785996 Chr.9: 124795140-124795140 on Build GRCh38
rs10986428 Chr.9: 124795312-124795312 on Build GRCh38
rs779404897 Chr.9: 124795093-124795093 on Build GRCh38
rs756385611 Chr.9: 124795001-124795001 on Build GRCh38
rs768324525 Chr.9: 124795130-124795130 on Build GRCh38
rs754097331 Chr.9: 124795092-124795092 on Build GRCh38
rs760021064 Chr.9: 124795116-124795116 on Build GRCh38

View Full Product Details