Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 129095003-129095505 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRAT
Entrez Gene ID
1384
Gene Name
carnitine O-acetyltransferase
Gene Aliases
CAT1
UniGene
Hs.12068
Gene Chromosome Location
Chr.9: 129094794-129110791 on Build GRCh38
Transcripts
NM_000755.3 NM_001257363.1 XM_005251708.3 XM_017014274.1 XM_017014275.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3847814 c.1822G>A p.E608K Chr.9: 129095456-129095456 on Build GRCh38
6873552 c.1872C>A p.A624A Chr.9: 129095406-129095406 on Build GRCh38
6244342 c.1818C>A p.Y606* Chr.9: 129095460-129095460 on Build GRCh38
3654799 c.1864C>T p.P622S Chr.9: 129095414-129095414 on Build GRCh38
7000758 c.1846G>T p.A616S Chr.9: 129095432-129095432 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776673270 Chr.9: 129095414-129095414 on Build GRCh38
rs747989201 Chr.9: 129095477-129095477 on Build GRCh38
rs947302653 Chr.9: 129095117-129095117 on Build GRCh38
rs529448801 Chr.9: 129095450-129095450 on Build GRCh38
rs914214561 Chr.9: 129095108-129095108 on Build GRCh38
rs1039403835 Chr.9: 129095299-129095299 on Build GRCh38
rs781091098 Chr.9: 129095189-129095189 on Build GRCh38
rs768702880 Chr.9: 129095381-129095381 on Build GRCh38
rs925634795 Chr.9: 129095466-129095466 on Build GRCh38
rs770646309 Chr.9: 129095365-129095366 on Build GRCh38
rs886512645 Chr.9: 129095146-129095146 on Build GRCh38
rs979836754 Chr.9: 129095465-129095465 on Build GRCh38
rs751985419 Chr.9: 129095458-129095458 on Build GRCh38
rs17455684 Chr.9: 129095338-129095338 on Build GRCh38
rs544209490 Chr.9: 129095175-129095175 on Build GRCh38
rs552808792 Chr.9: 129095056-129095056 on Build GRCh38
rs1012914896 Chr.9: 129095218-129095218 on Build GRCh38
rs935880183 Chr.9: 129095030-129095039 on Build GRCh38
rs867806081 Chr.9: 129095484-129095484 on Build GRCh38
rs1012240026 Chr.9: 129095448-129095448 on Build GRCh38
rs982262940 Chr.9: 129095259-129095259 on Build GRCh38
rs147892426 Chr.9: 129095089-129095089 on Build GRCh38
rs934342107 Chr.9: 129095083-129095083 on Build GRCh38
rs997358613 Chr.9: 129095136-129095136 on Build GRCh38
rs1051832735 Chr.9: 129095144-129095144 on Build GRCh38
rs143822726 Chr.9: 129095435-129095435 on Build GRCh38
rs748123046 Chr.9: 129095403-129095403 on Build GRCh38
rs755812329 Chr.9: 129095373-129095373 on Build GRCh38
rs1052751317 Chr.9: 129095091-129095091 on Build GRCh38
rs17486059 Chr.9: 129095133-129095133 on Build GRCh38
rs561708819 Chr.9: 129095167-129095167 on Build GRCh38
rs759666546 Chr.9: 129095431-129095431 on Build GRCh38
rs947035496 Chr.9: 129095286-129095286 on Build GRCh38
rs779060953 Chr.9: 129095385-129095385 on Build GRCh38
rs921865724 Chr.9: 129095340-129095340 on Build GRCh38
rs551012337 Chr.9: 129095411-129095411 on Build GRCh38
rs112134266 Chr.9: 129095306-129095306 on Build GRCh38
rs760770816 Chr.9: 129095410-129095410 on Build GRCh38
rs532999065 Chr.9: 129095235-129095235 on Build GRCh38
rs779910980 Chr.9: 129095467-129095467 on Build GRCh38
rs776627595 Chr.9: 129095461-129095462 on Build GRCh38
rs558434911 Chr.9: 129095134-129095134 on Build GRCh38
rs1009421940 Chr.9: 129095059-129095059 on Build GRCh38
rs1044555 Chr.9: 129095276-129095276 on Build GRCh38
rs777404087 Chr.9: 129095480-129095480 on Build GRCh38
rs1024336096 Chr.9: 129095454-129095454 on Build GRCh38
rs764316315 Chr.9: 129095438-129095438 on Build GRCh38
rs996192245 Chr.9: 129095324-129095324 on Build GRCh38
rs1004864086 Chr.9: 129095156-129095156 on Build GRCh38
rs2296770 Chr.9: 129095481-129095481 on Build GRCh38
rs879245871 Chr.9: 129095217-129095217 on Build GRCh38
rs11559172 Chr.9: 129095241-129095241 on Build GRCh38
rs369183439 Chr.9: 129095265-129095265 on Build GRCh38
rs564381366 Chr.9: 129095188-129095188 on Build GRCh38
rs988528282 Chr.9: 129095269-129095269 on Build GRCh38
rs1023886884 Chr.9: 129095227-129095227 on Build GRCh38
rs559420906 Chr.9: 129095246-129095246 on Build GRCh38
rs575299367 Chr.9: 129095171-129095171 on Build GRCh38
rs535290438 Chr.9: 129095123-129095123 on Build GRCh38
rs762626138 Chr.9: 129095464-129095464 on Build GRCh38
rs913098477 Chr.9: 129095238-129095238 on Build GRCh38
rs758754267 Chr.9: 129095346-129095346 on Build GRCh38
rs181730785 Chr.9: 129095486-129095486 on Build GRCh38
rs981637728 Chr.9: 129095044-129095044 on Build GRCh38
rs367953219 Chr.9: 129095377-129095377 on Build GRCh38
rs772079837 Chr.9: 129095478-129095478 on Build GRCh38
rs1052954857 Chr.9: 129095052-129095052 on Build GRCh38
rs749984082 Chr.9: 129095354-129095354 on Build GRCh38
rs1049062532 Chr.9: 129095342-129095342 on Build GRCh38
rs753394063 Chr.9: 129095473-129095473 on Build GRCh38
rs914247126 Chr.9: 129095279-129095279 on Build GRCh38
rs1019028508 Chr.9: 129095063-129095063 on Build GRCh38
rs928390108 Chr.9: 129095260-129095260 on Build GRCh38
rs773085650 Chr.9: 129095409-129095409 on Build GRCh38
rs2229419 Chr.9: 129095351-129095351 on Build GRCh38
rs778580722 Chr.9: 129095476-129095476 on Build GRCh38
rs779671477 Chr.9: 129095375-129095375 on Build GRCh38
rs149016840 Chr.9: 129095434-129095434 on Build GRCh38
rs17459086 Chr.9: 129095408-129095408 on Build GRCh38
rs186434192 Chr.9: 129095287-129095287 on Build GRCh38
rs375902962 Chr.9: 129095245-129095245 on Build GRCh38

View Full Product Details