Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 49590997-49591493 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAGE2A
Entrez Gene ID
729447
Gene Name
G antigen 2A
Gene Aliases
CT4.2, GAGE-2, GAGE-2A, GAGE2
UniGene
Hs.460641
Gene Chromosome Location
Chr.X: 49589515-49596827 on Build GRCh38
Transcripts
NM_001127212.1

SNP Detail

SNP ID Chromosome Location
rs782430757 Chr.X: 49591208-49591208 on Build GRCh38
rs781864628 Chr.X: 49591350-49591350 on Build GRCh38
rs781795933 Chr.X: 49591271-49591271 on Build GRCh38
rs5906821 Chr.X: 49591290-49591290 on Build GRCh38
rs782617924 Chr.X: 49591356-49591356 on Build GRCh38
rs782136866 Chr.X: 49591306-49591306 on Build GRCh38
rs781889853 Chr.X: 49591339-49591339 on Build GRCh38
rs782095130 Chr.X: 49591260-49591260 on Build GRCh38
rs202091876 Chr.X: 49591168-49591168 on Build GRCh38
rs782121533 Chr.X: 49591170-49591170 on Build GRCh38
rs200636160 Chr.X: 49591237-49591237 on Build GRCh38
rs782110551 Chr.X: 49591248-49591248 on Build GRCh38
rs782567731 Chr.X: 49591239-49591239 on Build GRCh38
rs782721321 Chr.X: 49591188-49591188 on Build GRCh38
rs112425647 Chr.X: 49591401-49591401 on Build GRCh38
rs782749845 Chr.X: 49591347-49591347 on Build GRCh38
rs782190214 Chr.X: 49591302-49591302 on Build GRCh38
rs782398935 Chr.X: 49591304-49591304 on Build GRCh38
rs781826029 Chr.X: 49591155-49591155 on Build GRCh38
rs782358678 Chr.X: 49591296-49591296 on Build GRCh38
rs781973809 Chr.X: 49591312-49591312 on Build GRCh38
rs782680763 Chr.X: 49591279-49591279 on Build GRCh38
rs781804610 Chr.X: 49591261-49591261 on Build GRCh38
rs782385785 Chr.X: 49591249-49591249 on Build GRCh38
rs369376588 Chr.X: 49591387-49591388 on Build GRCh38
rs782805716 Chr.X: 49591164-49591164 on Build GRCh38
rs781838138 Chr.X: 49591203-49591203 on Build GRCh38
rs782058124 Chr.X: 49591325-49591325 on Build GRCh38
rs782623251 Chr.X: 49591270-49591270 on Build GRCh38
rs782462551 Chr.X: 49591352-49591352 on Build GRCh38
rs782729976 Chr.X: 49591309-49591309 on Build GRCh38
rs113157222 Chr.X: 49591438-49591438 on Build GRCh38
rs782146001 Chr.X: 49591162-49591162 on Build GRCh38
rs782766457 Chr.X: 49591262-49591262 on Build GRCh38
rs782031240 Chr.X: 49591245-49591245 on Build GRCh38
rs782736319 Chr.X: 49591333-49591333 on Build GRCh38
rs782456159 Chr.X: 49591285-49591286 on Build GRCh38
rs5953311 Chr.X: 49591026-49591026 on Build GRCh38
rs782508519 Chr.X: 49591233-49591233 on Build GRCh38
rs782208002 Chr.X: 49591291-49591291 on Build GRCh38
rs782651808 Chr.X: 49591234-49591234 on Build GRCh38
rs782452890 Chr.X: 49591278-49591278 on Build GRCh38
rs781909525 Chr.X: 49591230-49591230 on Build GRCh38
rs59427157 Chr.X: 49591406-49591407 on Build GRCh38
rs781953355 Chr.X: 49591254-49591254 on Build GRCh38
rs782474596 Chr.X: 49591266-49591266 on Build GRCh38
rs781938096 Chr.X: 49591156-49591156 on Build GRCh38
rs781918490 Chr.X: 49591301-49591301 on Build GRCh38
rs782209643 Chr.X: 49591235-49591235 on Build GRCh38
rs782401289 Chr.X: 49591244-49591244 on Build GRCh38
rs782499790 Chr.X: 49591287-49591287 on Build GRCh38
rs781871920 Chr.X: 49591263-49591263 on Build GRCh38
rs782284018 Chr.X: 49591280-49591280 on Build GRCh38
rs781988443 Chr.X: 49591305-49591305 on Build GRCh38
rs57514394 Chr.X: 49591469-49591469 on Build GRCh38
rs782583005 Chr.X: 49591228-49591228 on Build GRCh38
rs60862992 Chr.X: 49591405-49591406 on Build GRCh38

View Full Product Details