Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 54920775-54921267 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRO
Entrez Gene ID
7216
Gene Name
trophinin
Gene Aliases
MAGE-d3, MAGED3
UniGene
Hs.633653
Gene Chromosome Location
Chr.X: 54920563-54931436 on Build GRCh38
Transcripts
NM_001039705.2 NM_001271183.1 NM_001271184.1 NM_016157.3 NM_177556.2 NM_177557.2 NR_073148.1 NR_073149.1 XM_006724600.2 XM_011530808.1 XM_011530809.1 XM_011530811.2 XM_011530812.1 XM_011530813.1 XM_011530814.1 XM_017029767.1 XM_017029768.1 XM_017029769.1 XM_017029770.1 XM_017029771.1 XM_017029772.1 XM_017029773.1

SNP Detail

SNP ID Chromosome Location
rs935903005 Chr.X: 54921179-54921179 on Build GRCh38
rs146989702 Chr.X: 54920827-54920827 on Build GRCh38
rs1020811483 Chr.X: 54920855-54920855 on Build GRCh38
rs908744362 Chr.X: 54921003-54921003 on Build GRCh38
rs574039835 Chr.X: 54921198-54921198 on Build GRCh38
rs917923941 Chr.X: 54920803-54920803 on Build GRCh38
rs891584890 Chr.X: 54920850-54920850 on Build GRCh38
rs747999681 Chr.X: 54921227-54921227 on Build GRCh38
rs756467807 Chr.X: 54920810-54920811 on Build GRCh38
rs915126539 Chr.X: 54921163-54921163 on Build GRCh38
rs1050072410 Chr.X: 54920831-54920831 on Build GRCh38
rs370937491 Chr.X: 54921146-54921146 on Build GRCh38
rs948064506 Chr.X: 54920810-54920810 on Build GRCh38
rs778193090 Chr.X: 54920868-54920868 on Build GRCh38
rs182867576 Chr.X: 54921056-54921056 on Build GRCh38
rs902393605 Chr.X: 54921177-54921177 on Build GRCh38
rs1042608876 Chr.X: 54921176-54921176 on Build GRCh38
rs984974233 Chr.X: 54920930-54920930 on Build GRCh38
rs995823921 Chr.X: 54920911-54920911 on Build GRCh38
rs965755569 Chr.X: 54920910-54920910 on Build GRCh38
rs1054702878 Chr.X: 54920832-54920832 on Build GRCh38
rs187209993 Chr.X: 54920892-54920892 on Build GRCh38
rs959993239 Chr.X: 54920884-54920884 on Build GRCh38
rs747875480 Chr.X: 54921016-54921016 on Build GRCh38
rs1054352994 Chr.X: 54921230-54921230 on Build GRCh38
rs952333197 Chr.X: 54920919-54920919 on Build GRCh38
rs946686893 Chr.X: 54921175-54921175 on Build GRCh38
rs998930132 Chr.X: 54920859-54920859 on Build GRCh38
rs1049425739 Chr.X: 54921074-54921074 on Build GRCh38
rs1011134747 Chr.X: 54920844-54920844 on Build GRCh38
rs779138771 Chr.X: 54921030-54921030 on Build GRCh38
rs1010422573 Chr.X: 54920856-54920856 on Build GRCh38
rs1000103711 Chr.X: 54920829-54920829 on Build GRCh38
rs992723566 Chr.X: 54921001-54921001 on Build GRCh38
rs1045067585 Chr.X: 54920812-54920812 on Build GRCh38
rs192456184 Chr.X: 54920913-54920913 on Build GRCh38
rs1018669884 Chr.X: 54920857-54920857 on Build GRCh38
rs1056513313 Chr.X: 54920801-54920801 on Build GRCh38
rs1040723662 Chr.X: 54920804-54920804 on Build GRCh38
rs908039247 Chr.X: 54920937-54920937 on Build GRCh38
rs962207366 Chr.X: 54920997-54920997 on Build GRCh38
rs185973384 Chr.X: 54921061-54921061 on Build GRCh38
rs925238955 Chr.X: 54921094-54921094 on Build GRCh38
rs984681901 Chr.X: 54920940-54920940 on Build GRCh38
rs1035652217 Chr.X: 54920878-54920878 on Build GRCh38
rs775853501 Chr.X: 54921072-54921072 on Build GRCh38

View Full Product Details