Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 57119579-57120087 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPIN2B
Entrez Gene ID
474343
Gene Name
spindlin family member 2B
Gene Aliases
SPIN-2, SPIN-2B, SPIN2_duplicate, TDRD26, dJ323P24.2
UniGene
Hs.460717
Gene Chromosome Location
Chr.X: 57112142-57121905 on Build GRCh38
Transcripts
NM_001006681.1 NM_001006682.1 NM_001006683.1 NM_001282461.1 NM_001282462.1 XM_005262008.4 XM_005262009.4 XM_005262010.1 XM_011530788.2 XM_011530789.2 XM_017029552.1

SNP Detail

SNP ID Chromosome Location
rs747994297 Chr.X: 57119802-57119802 on Build GRCh38
rs1018169222 Chr.X: 57119684-57119684 on Build GRCh38
rs779679512 Chr.X: 57119957-57119957 on Build GRCh38
rs758804929 Chr.X: 57119916-57119916 on Build GRCh38
rs752955593 Chr.X: 57119907-57119907 on Build GRCh38
rs757886936 Chr.X: 57119955-57119955 on Build GRCh38
rs753262603 Chr.X: 57119777-57119777 on Build GRCh38
rs753421766 Chr.X: 57120006-57120006 on Build GRCh38
rs771899871 Chr.X: 57120026-57120026 on Build GRCh38
rs772145184 Chr.X: 57119638-57119638 on Build GRCh38
rs957011368 Chr.X: 57120022-57120022 on Build GRCh38
rs1736863 Chr.X: 57119690-57119690 on Build GRCh38
rs752093364 Chr.X: 57119954-57119954 on Build GRCh38
rs199979060 Chr.X: 57119830-57119830 on Build GRCh38
rs759725620 Chr.X: 57119890-57119890 on Build GRCh38
rs992149889 Chr.X: 57120036-57120036 on Build GRCh38
rs364007 Chr.X: 57119772-57119772 on Build GRCh38
rs766437234 Chr.X: 57119845-57119845 on Build GRCh38
rs756369740 Chr.X: 57119784-57119784 on Build GRCh38
rs760283208 Chr.X: 57119806-57119806 on Build GRCh38
rs1736862 Chr.X: 57119696-57119696 on Build GRCh38
rs776742693 Chr.X: 57119846-57119846 on Build GRCh38
rs764588115 Chr.X: 57119991-57119991 on Build GRCh38
rs955262122 Chr.X: 57119673-57119673 on Build GRCh38
rs1736864 Chr.X: 57119689-57119689 on Build GRCh38
rs750265983 Chr.X: 57119714-57119714 on Build GRCh38
rs760600260 Chr.X: 57119842-57119842 on Build GRCh38
rs924017959 Chr.X: 57119981-57119981 on Build GRCh38
rs707345 Chr.X: 57119602-57119602 on Build GRCh38
rs775882172 Chr.X: 57119644-57119644 on Build GRCh38
rs771843968 Chr.X: 57119814-57119814 on Build GRCh38
rs778637262 Chr.X: 57119994-57119994 on Build GRCh38
rs761013157 Chr.X: 57119658-57119658 on Build GRCh38
rs878894553 Chr.X: 57119930-57119930 on Build GRCh38
rs967850229 Chr.X: 57119714-57119714 on Build GRCh38
rs759720127 Chr.X: 57119637-57119637 on Build GRCh38
rs1006747171 Chr.X: 57119678-57119678 on Build GRCh38
rs758660889 Chr.X: 57119829-57119830 on Build GRCh38
rs748835775 Chr.X: 57119975-57119975 on Build GRCh38
rs765475615 Chr.X: 57119895-57119895 on Build GRCh38
rs764432914 Chr.X: 57119937-57119937 on Build GRCh38
rs993809748 Chr.X: 57119625-57119625 on Build GRCh38
rs767862610 Chr.X: 57119664-57119664 on Build GRCh38

View Full Product Details