Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 65502495-65502991 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZC3H12B
Entrez Gene ID
340554
Gene Name
zinc finger CCCH-type containing 12B
Gene Aliases
CXorf32, MCPIP2
UniGene
Hs.21249
Gene Chromosome Location
Chr.X: 65034815-65507887 on Build GRCh38
Transcripts
NM_001010888.3 XM_011530939.2 XM_017029478.1 XM_017029479.1 XM_017029480.1 XM_017029481.1 XM_017029482.1 XM_017029483.1 XM_017029484.1

SNP Detail

SNP ID Chromosome Location
rs368356777 Chr.X: 65502870-65502870 on Build GRCh38
rs373987687 Chr.X: 65502687-65502687 on Build GRCh38
rs865857585 Chr.X: 65502763-65502763 on Build GRCh38
rs750374120 Chr.X: 65502840-65502840 on Build GRCh38
rs749055478 Chr.X: 65502618-65502618 on Build GRCh38
rs925024748 Chr.X: 65502828-65502828 on Build GRCh38
rs370582971 Chr.X: 65502887-65502887 on Build GRCh38
rs1010742248 Chr.X: 65502672-65502672 on Build GRCh38
rs747943213 Chr.X: 65502894-65502894 on Build GRCh38
rs768266916 Chr.X: 65502779-65502779 on Build GRCh38
rs769459646 Chr.X: 65502609-65502609 on Build GRCh38
rs573884415 Chr.X: 65502918-65502918 on Build GRCh38
rs779281412 Chr.X: 65502560-65502560 on Build GRCh38
rs752707090 Chr.X: 65502733-65502733 on Build GRCh38
rs769595611 Chr.X: 65502709-65502709 on Build GRCh38
rs760832460 Chr.X: 65502817-65502817 on Build GRCh38
rs773443238 Chr.X: 65502661-65502661 on Build GRCh38
rs768565241 Chr.X: 65502620-65502620 on Build GRCh38
rs1002101579 Chr.X: 65502815-65502815 on Build GRCh38
rs766676462 Chr.X: 65502710-65502710 on Build GRCh38
rs1023452277 Chr.X: 65502778-65502778 on Build GRCh38
rs777837819 Chr.X: 65502929-65502929 on Build GRCh38
rs765480548 Chr.X: 65502845-65502845 on Build GRCh38
rs905137980 Chr.X: 65502808-65502808 on Build GRCh38
rs753303498 Chr.X: 65502568-65502568 on Build GRCh38
rs186386233 Chr.X: 65502561-65502561 on Build GRCh38
rs369883605 Chr.X: 65502532-65502532 on Build GRCh38
rs761811198 Chr.X: 65502647-65502647 on Build GRCh38
rs767327735 Chr.X: 65502653-65502656 on Build GRCh38
rs752130811 Chr.X: 65502861-65502861 on Build GRCh38
rs759804399 Chr.X: 65502713-65502713 on Build GRCh38
rs758539246 Chr.X: 65502772-65502772 on Build GRCh38
rs753129536 Chr.X: 65502846-65502846 on Build GRCh38
rs868058917 Chr.X: 65502722-65502722 on Build GRCh38
rs755664779 Chr.X: 65502767-65502767 on Build GRCh38
rs961770025 Chr.X: 65502927-65502927 on Build GRCh38
rs774144741 Chr.X: 65502625-65502625 on Build GRCh38
rs753390684 Chr.X: 65502868-65502868 on Build GRCh38
rs985114779 Chr.X: 65502914-65502914 on Build GRCh38
rs758334616 Chr.X: 65502924-65502924 on Build GRCh38
rs755409380 Chr.X: 65502559-65502559 on Build GRCh38
rs374042825 Chr.X: 65502776-65502776 on Build GRCh38
rs778796872 Chr.X: 65502787-65502787 on Build GRCh38
rs749916260 Chr.X: 65502764-65502764 on Build GRCh38
rs747983053 Chr.X: 65502796-65502796 on Build GRCh38
rs377369268 Chr.X: 65502879-65502879 on Build GRCh38
rs746955344 Chr.X: 65502931-65502931 on Build GRCh38
rs966582167 Chr.X: 65502786-65502786 on Build GRCh38
rs771191242 Chr.X: 65502825-65502825 on Build GRCh38
rs772001538 Chr.X: 65502651-65502651 on Build GRCh38
rs780760315 Chr.X: 65502587-65502587 on Build GRCh38
rs867891151 Chr.X: 65502526-65502526 on Build GRCh38
rs753080155 Chr.X: 65502736-65502736 on Build GRCh38
rs763277063 Chr.X: 65502859-65502859 on Build GRCh38
rs764558199 Chr.X: 65502860-65502860 on Build GRCh38
rs781174103 Chr.X: 65502813-65502813 on Build GRCh38
rs780632228 Chr.X: 65502759-65502759 on Build GRCh38
rs772020269 Chr.X: 65502807-65502807 on Build GRCh38
rs756665932 Chr.X: 65502583-65502583 on Build GRCh38
rs759811849 Chr.X: 65502841-65502841 on Build GRCh38
rs765454587 Chr.X: 65502589-65502589 on Build GRCh38
rs758976126 Chr.X: 65502742-65502742 on Build GRCh38
rs772825957 Chr.X: 65502893-65502893 on Build GRCh38
rs765890816 Chr.X: 65502866-65502866 on Build GRCh38
rs779734586 Chr.X: 65502615-65502615 on Build GRCh38
rs960605127 Chr.X: 65502558-65502558 on Build GRCh38
rs754232430 Chr.X: 65502550-65502550 on Build GRCh38

View Full Product Details