Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 152869103-152869604 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NSDHL
Entrez Gene ID
50814
Gene Name
NAD(P) dependent steroid dehydrogenase-like
Gene Aliases
H105E3, SDR31E1, XAP104
UniGene
Hs.57698
Gene Chromosome Location
Chr.X: 152830967-152869363 on Build GRCh38
Transcripts
NM_001129765.1 NM_015922.2 XM_011531178.2 XM_017029564.1

SNP Detail

SNP ID Chromosome Location
rs182657412 Chr.X: 152869266-152869266 on Build GRCh38
rs782236914 Chr.X: 152869262-152869262 on Build GRCh38
rs12558405 Chr.X: 152869435-152869435 on Build GRCh38
rs1010558224 Chr.X: 152869506-152869506 on Build GRCh38
rs977633375 Chr.X: 152869261-152869261 on Build GRCh38
rs1021805530 Chr.X: 152869517-152869517 on Build GRCh38
rs1047574158 Chr.X: 152869412-152869412 on Build GRCh38
rs899110537 Chr.X: 152869392-152869392 on Build GRCh38
rs782010818 Chr.X: 152869288-152869288 on Build GRCh38
rs782195589 Chr.X: 152869349-152869349 on Build GRCh38
rs969016476 Chr.X: 152869533-152869533 on Build GRCh38
rs782234098 Chr.X: 152869218-152869218 on Build GRCh38
rs781924147 Chr.X: 152869146-152869146 on Build GRCh38
rs782188221 Chr.X: 152869219-152869219 on Build GRCh38
rs111690259 Chr.X: 152869430-152869430 on Build GRCh38
rs782770511 Chr.X: 152869137-152869137 on Build GRCh38
rs969279531 Chr.X: 152869256-152869256 on Build GRCh38
rs782416397 Chr.X: 152869328-152869328 on Build GRCh38
rs948739695 Chr.X: 152869355-152869355 on Build GRCh38
rs782331380 Chr.X: 152869224-152869224 on Build GRCh38
rs1017349172 Chr.X: 152869433-152869433 on Build GRCh38
rs145978994 Chr.X: 152869245-152869245 on Build GRCh38
rs929101486 Chr.X: 152869407-152869407 on Build GRCh38
rs782354458 Chr.X: 152869142-152869142 on Build GRCh38
rs782160469 Chr.X: 152869398-152869398 on Build GRCh38
rs782365620 Chr.X: 152869362-152869362 on Build GRCh38
rs781956436 Chr.X: 152869371-152869371 on Build GRCh38
rs938782662 Chr.X: 152869303-152869303 on Build GRCh38
rs141761159 Chr.X: 152869422-152869422 on Build GRCh38
rs782661738 Chr.X: 152869348-152869348 on Build GRCh38
rs890159685 Chr.X: 152869415-152869415 on Build GRCh38
rs373088998 Chr.X: 152869141-152869141 on Build GRCh38
rs1031846650 Chr.X: 152869564-152869564 on Build GRCh38
rs186807967 Chr.X: 152869542-152869542 on Build GRCh38
rs782201839 Chr.X: 152869163-152869163 on Build GRCh38

View Full Product Details