Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 153594460-153594959 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM58A
Entrez Gene ID
92002
Gene Name
family with sequence similarity 58 member A
Gene Aliases
STAR
UniGene
Hs.496943
Gene Chromosome Location
Chr.X: 153587925-153599177 on Build GRCh38
Transcripts
NM_001130997.2 NM_152274.4 XM_005277920.4 XM_005277921.4 XM_011531214.2 XM_011531215.2

SNP Detail

SNP ID Chromosome Location
rs1053807918 Chr.X: 153594856-153594856 on Build GRCh38
rs782501590 Chr.X: 153594740-153594740 on Build GRCh38
rs190360145 Chr.X: 153594529-153594529 on Build GRCh38
rs781799914 Chr.X: 153594579-153594579 on Build GRCh38
rs782558422 Chr.X: 153594631-153594631 on Build GRCh38
rs142632816 Chr.X: 153594628-153594628 on Build GRCh38
rs782571286 Chr.X: 153594936-153594936 on Build GRCh38
rs923246561 Chr.X: 153594781-153594781 on Build GRCh38
rs782788324 Chr.X: 153594540-153594540 on Build GRCh38
rs781965328 Chr.X: 153594824-153594824 on Build GRCh38
rs201043377 Chr.X: 153594639-153594639 on Build GRCh38
rs183758563 Chr.X: 153594590-153594590 on Build GRCh38
rs781876804 Chr.X: 153594711-153594711 on Build GRCh38
rs918989098 Chr.X: 153594624-153594624 on Build GRCh38
rs782277665 Chr.X: 153594692-153594692 on Build GRCh38
rs1042412551 Chr.X: 153594787-153594787 on Build GRCh38
rs150562029 Chr.X: 153594638-153594638 on Build GRCh38
rs781984911 Chr.X: 153594714-153594714 on Build GRCh38
rs371751760 Chr.X: 153594716-153594716 on Build GRCh38
rs1012411134 Chr.X: 153594924-153594924 on Build GRCh38
rs146658835 Chr.X: 153594935-153594935 on Build GRCh38
rs781843718 Chr.X: 153594593-153594593 on Build GRCh38
rs1025231159 Chr.X: 153594553-153594553 on Build GRCh38
rs143133700 Chr.X: 153594539-153594539 on Build GRCh38
rs782289301 Chr.X: 153594813-153594813 on Build GRCh38
rs782055608 Chr.X: 153594534-153594534 on Build GRCh38
rs782715378 Chr.X: 153594616-153594616 on Build GRCh38
rs374423745 Chr.X: 153594552-153594552 on Build GRCh38
rs782038786 Chr.X: 153594684-153594684 on Build GRCh38
rs782080063 Chr.X: 153594912-153594912 on Build GRCh38
rs782608728 Chr.X: 153594657-153594657 on Build GRCh38
rs782498226 Chr.X: 153594641-153594641 on Build GRCh38
rs782319474 Chr.X: 153594683-153594683 on Build GRCh38
rs782469736 Chr.X: 153594614-153594614 on Build GRCh38
rs782206940 Chr.X: 153594663-153594663 on Build GRCh38
rs1000877975 Chr.X: 153594834-153594834 on Build GRCh38
rs782688675 Chr.X: 153594785-153594785 on Build GRCh38
rs902528557 Chr.X: 153594816-153594816 on Build GRCh38
rs981165355 Chr.X: 153594611-153594611 on Build GRCh38
rs782789648 Chr.X: 153594522-153594522 on Build GRCh38
rs971471069 Chr.X: 153594565-153594565 on Build GRCh38

View Full Product Details