Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 231694133-231694643 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DISC1
Entrez Gene ID
27185
Gene Name
disrupted in schizophrenia 1
Gene Aliases
C1orf136, SCZD9
UniGene
Hs.13318
Gene Chromosome Location
Chr.1: 231626815-232041272 on Build GRCh38
Transcripts
NM_001012957.1 NM_001012958.1 NM_001012959.1 NM_001164537.1 NM_001164538.1 NM_001164539.1 NM_001164540.1 NM_001164541.1 NM_001164542.1 NM_001164544.1 NM_001164545.1 NM_001164546.1 NM_001164547.1 NM_001164548.1 NM_001164549.1 NM_001164550.1 NM_001164551.1 NM_001164552.1 NM_001164553.1 NM_001164554.1 NM_001164555.1 NM_001164556.1 NM_018662.2 NR_028393.1 NR_028394.1 NR_028395.1 NR_028396.1 NR_028397.1 NR_028398.1 NR_028399.1 NR_028400.1

Gene Symbol
TSNAX-DISC1
Entrez Gene ID
100303453
Gene Name
TSNAX-DISC1 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.13318
Gene Chromosome Location
Chr.1: 231528653-232041272 on Build GRCh38
Transcripts
NM_001012957.1 NM_001012958.1 NM_001012959.1 NM_001164537.1 NM_001164538.1 NM_001164539.1 NM_001164540.1 NM_001164541.1 NM_001164542.1 NM_001164544.1 NM_001164545.1 NM_001164546.1 NM_001164547.1 NM_001164548.1 NM_001164549.1 NM_001164550.1 NM_001164551.1 NM_001164552.1 NM_001164553.1 NM_001164554.1 NM_001164555.1 NM_001164556.1 NM_018662.2 NR_028393.1 NR_028394.1 NR_028395.1 NR_028396.1 NR_028397.1 NR_028398.1 NR_028399.1 NR_028400.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7257118 c.558C>T p.S186S Chr.1: 231694316-231694316 on Build GRCh38
6541965 c.536C>T p.S179L Chr.1: 231694294-231694294 on Build GRCh38
1998581 c.717C>T p.C239C Chr.1: 231694475-231694475 on Build GRCh38
7216869 c.791G>A p.R264Q Chr.1: 231694549-231694549 on Build GRCh38
4958215 c.423T>A p.A141A Chr.1: 231694181-231694181 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765577746 Chr.1: 231694579-231694579 on Build GRCh38
rs559000734 Chr.1: 231694343-231694343 on Build GRCh38
rs751883731 Chr.1: 231694308-231694308 on Build GRCh38
rs762099070 Chr.1: 231694487-231694487 on Build GRCh38
rs199530992 Chr.1: 231694468-231694468 on Build GRCh38
rs747906699 Chr.1: 231694358-231694358 on Build GRCh38
rs778749511 Chr.1: 231694613-231694613 on Build GRCh38
rs913903792 Chr.1: 231694384-231694384 on Build GRCh38
rs769513744 Chr.1: 231694422-231694422 on Build GRCh38
rs146439119 Chr.1: 231694426-231694426 on Build GRCh38
rs932186534 Chr.1: 231694177-231694177 on Build GRCh38
rs138228295 Chr.1: 231694598-231694598 on Build GRCh38
rs766759086 Chr.1: 231694588-231694588 on Build GRCh38
rs780719614 Chr.1: 231694518-231694518 on Build GRCh38
rs377484189 Chr.1: 231694535-231694535 on Build GRCh38
rs764811265 Chr.1: 231694527-231694529 on Build GRCh38
rs776775277 Chr.1: 231694572-231694572 on Build GRCh38
rs762545623 Chr.1: 231694485-231694485 on Build GRCh38
rs201249128 Chr.1: 231694456-231694456 on Build GRCh38
rs973491556 Chr.1: 231694159-231694159 on Build GRCh38
rs139667828 Chr.1: 231694313-231694313 on Build GRCh38
rs768870892 Chr.1: 231694571-231694571 on Build GRCh38
rs776311202 Chr.1: 231694272-231694272 on Build GRCh38
rs373288445 Chr.1: 231694534-231694534 on Build GRCh38
rs766374745 Chr.1: 231694367-231694367 on Build GRCh38
rs771557495 Chr.1: 231694258-231694258 on Build GRCh38
rs750925617 Chr.1: 231694512-231694512 on Build GRCh38
rs767741003 Chr.1: 231694316-231694316 on Build GRCh38
rs150489951 Chr.1: 231694224-231694224 on Build GRCh38
rs775660207 Chr.1: 231694560-231694560 on Build GRCh38
rs750026954 Chr.1: 231694540-231694557 on Build GRCh38
rs148973353 Chr.1: 231694548-231694548 on Build GRCh38
rs774425560 Chr.1: 231694256-231694256 on Build GRCh38
rs867093901 Chr.1: 231694492-231694492 on Build GRCh38
rs763492083 Chr.1: 231694582-231694582 on Build GRCh38
rs759594286 Chr.1: 231694447-231694447 on Build GRCh38
rs367543084 Chr.1: 231694230-231694230 on Build GRCh38
rs754450096 Chr.1: 231694418-231694418 on Build GRCh38
rs754230811 Chr.1: 231694481-231694481 on Build GRCh38
rs181767113 Chr.1: 231694249-231694249 on Build GRCh38
rs747828145 Chr.1: 231694419-231694419 on Build GRCh38
rs367543086 Chr.1: 231694599-231694599 on Build GRCh38
rs375607284 Chr.1: 231694440-231694440 on Build GRCh38
rs750801544 Chr.1: 231694204-231694204 on Build GRCh38
rs199893176 Chr.1: 231694524-231694524 on Build GRCh38
rs767632505 Chr.1: 231694448-231694448 on Build GRCh38
rs763792460 Chr.1: 231694210-231694210 on Build GRCh38
rs760521090 Chr.1: 231694184-231694184 on Build GRCh38
rs765087160 Chr.1: 231694276-231694276 on Build GRCh38
rs780432594 Chr.1: 231694229-231694229 on Build GRCh38
rs201600649 Chr.1: 231694215-231694215 on Build GRCh38
rs763179161 Chr.1: 231694286-231694286 on Build GRCh38
rs760289681 Chr.1: 231694267-231694267 on Build GRCh38
rs755437485 Chr.1: 231694231-231694231 on Build GRCh38
rs774376929 Chr.1: 231694442-231694442 on Build GRCh38
rs752952040 Chr.1: 231694389-231694389 on Build GRCh38
rs757944052 Chr.1: 231694344-231694344 on Build GRCh38
rs761627224 Chr.1: 231694273-231694273 on Build GRCh38
rs755631643 Chr.1: 231694593-231694593 on Build GRCh38
rs141270877 Chr.1: 231694347-231694347 on Build GRCh38
rs879001090 Chr.1: 231694597-231694597 on Build GRCh38
rs186593988 Chr.1: 231694266-231694266 on Build GRCh38
rs775068297 Chr.1: 231694176-231694176 on Build GRCh38
rs3738401 Chr.1: 231694549-231694549 on Build GRCh38
rs761174007 Chr.1: 231694343-231694343 on Build GRCh38
rs367543085 Chr.1: 231694526-231694526 on Build GRCh38
rs745397878 Chr.1: 231694254-231694254 on Build GRCh38
rs764251510 Chr.1: 231694480-231694480 on Build GRCh38
rs201901960 Chr.1: 231694352-231694352 on Build GRCh38
rs945470362 Chr.1: 231694165-231694165 on Build GRCh38
rs986232343 Chr.1: 231694461-231694461 on Build GRCh38
rs754390526 Chr.1: 231694401-231694401 on Build GRCh38
rs143796295 Chr.1: 231694237-231694237 on Build GRCh38
rs756502841 Chr.1: 231694398-231694398 on Build GRCh38
rs769613668 Chr.1: 231694361-231694361 on Build GRCh38
rs760628896 Chr.1: 231694565-231694565 on Build GRCh38
rs199617790 Chr.1: 231694399-231694399 on Build GRCh38
rs753852441 Chr.1: 231694213-231694213 on Build GRCh38
rs773758385 Chr.1: 231694162-231694162 on Build GRCh38
rs113312552 Chr.1: 231694517-231694517 on Build GRCh38
rs746525075 Chr.1: 231694417-231694417 on Build GRCh38
rs1055229412 Chr.1: 231694410-231694410 on Build GRCh38
rs779350258 Chr.1: 231694406-231694406 on Build GRCh38
rs750325934 Chr.1: 231694279-231694279 on Build GRCh38
rs750932225 Chr.1: 231694581-231694581 on Build GRCh38
rs761815665 Chr.1: 231694317-231694317 on Build GRCh38
rs888103515 Chr.1: 231694244-231694244 on Build GRCh38
rs765745403 Chr.1: 231694500-231694500 on Build GRCh38
rs139091980 Chr.1: 231694240-231694240 on Build GRCh38
rs778277620 Chr.1: 231694253-231694253 on Build GRCh38
rs757157671 Chr.1: 231694607-231694607 on Build GRCh38
rs750498211 Chr.1: 231694221-231694221 on Build GRCh38
rs201258158 Chr.1: 231694574-231694574 on Build GRCh38
rs758482654 Chr.1: 231694223-231694223 on Build GRCh38
rs997930434 Chr.1: 231694449-231694449 on Build GRCh38
rs759010166 Chr.1: 231694163-231694163 on Build GRCh38
rs201556843 Chr.1: 231694465-231694465 on Build GRCh38
rs752258922 Chr.1: 231694590-231694590 on Build GRCh38
rs773166710 Chr.1: 231694425-231694425 on Build GRCh38
rs777504676 Chr.1: 231694527-231694527 on Build GRCh38
rs772075803 Chr.1: 231694619-231694619 on Build GRCh38
rs779060951 Chr.1: 231694542-231694542 on Build GRCh38
rs373365825 Chr.1: 231694469-231694469 on Build GRCh38
rs765171325 Chr.1: 231694217-231694217 on Build GRCh38
rs112577310 Chr.1: 231694335-231694335 on Build GRCh38
rs139420445 Chr.1: 231694405-231694405 on Build GRCh38
rs756445061 Chr.1: 231694338-231694338 on Build GRCh38
rs369315072 Chr.1: 231694353-231694353 on Build GRCh38
rs200704946 Chr.1: 231694385-231694385 on Build GRCh38
rs768544712 Chr.1: 231694269-231694269 on Build GRCh38
rs770237650 Chr.1: 231694242-231694242 on Build GRCh38
rs542010946 Chr.1: 231694292-231694292 on Build GRCh38
rs774606244 Chr.1: 231694368-231694368 on Build GRCh38
rs753537068 Chr.1: 231694606-231694606 on Build GRCh38
rs141459782 Chr.1: 231694573-231694573 on Build GRCh38
rs151311391 Chr.1: 231694191-231694191 on Build GRCh38
rs146244276 Chr.1: 231694618-231694618 on Build GRCh38
rs773346712 Chr.1: 231694362-231694362 on Build GRCh38
rs149702548 Chr.1: 231694537-231694537 on Build GRCh38
rs755865070 Chr.1: 231694525-231694525 on Build GRCh38
rs766559698 Chr.1: 231694306-231694306 on Build GRCh38
rs770528086 Chr.1: 231694156-231694156 on Build GRCh38

View Full Product Details