Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 231723805-231724317 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DISC1
Entrez Gene ID
27185
Gene Name
disrupted in schizophrenia 1
Gene Aliases
C1orf136, SCZD9
UniGene
Hs.13318
Gene Chromosome Location
Chr.1: 231626815-232041272 on Build GRCh38
Transcripts
NM_001012957.1 NM_001012958.1 NM_001012959.1 NM_001164537.1 NM_001164538.1 NM_001164539.1 NM_001164540.1 NM_001164541.1 NM_001164542.1 NM_001164544.1 NM_001164545.1 NM_001164546.1 NM_001164547.1 NM_001164548.1 NM_001164549.1 NM_001164550.1 NM_001164551.1 NM_001164552.1 NM_001164553.1 NM_001164555.1 NM_001164556.1 NM_018662.2 NR_028393.1 NR_028394.1 NR_028395.1 NR_028396.1 NR_028397.1 NR_028398.1 NR_028400.1

Gene Symbol
TSNAX-DISC1
Entrez Gene ID
100303453
Gene Name
TSNAX-DISC1 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.13318
Gene Chromosome Location
Chr.1: 231528653-232041272 on Build GRCh38
Transcripts
NM_001012957.1 NM_001012958.1 NM_001012959.1 NM_001164537.1 NM_001164538.1 NM_001164539.1 NM_001164540.1 NM_001164541.1 NM_001164542.1 NM_001164544.1 NM_001164545.1 NM_001164546.1 NM_001164547.1 NM_001164548.1 NM_001164549.1 NM_001164550.1 NM_001164551.1 NM_001164552.1 NM_001164553.1 NM_001164555.1 NM_001164556.1 NM_018662.2 NR_028393.1 NR_028394.1 NR_028395.1 NR_028396.1 NR_028397.1 NR_028398.1 NR_028400.1

SNP Detail

SNP ID Chromosome Location
rs1015661513 Chr.1: 231724130-231724130 on Build GRCh38
rs932338645 Chr.1: 231723941-231723941 on Build GRCh38
rs960934566 Chr.1: 231724183-231724183 on Build GRCh38
rs111811855 Chr.1: 231724186-231724186 on Build GRCh38
rs925646758 Chr.1: 231724213-231724213 on Build GRCh38
rs12126864 Chr.1: 231724200-231724200 on Build GRCh38
rs138623735 Chr.1: 231723857-231723857 on Build GRCh38
rs1049902456 Chr.1: 231723946-231723946 on Build GRCh38
rs114788152 Chr.1: 231724092-231724092 on Build GRCh38
rs771306896 Chr.1: 231724033-231724033 on Build GRCh38
rs908736756 Chr.1: 231723905-231723905 on Build GRCh38
rs140640340 Chr.1: 231724268-231724269 on Build GRCh38
rs932449970 Chr.1: 231723913-231723913 on Build GRCh38
rs905686642 Chr.1: 231724290-231724290 on Build GRCh38
rs554945891 Chr.1: 231724268-231724268 on Build GRCh38
rs1011005512 Chr.1: 231724100-231724100 on Build GRCh38
rs984196206 Chr.1: 231723901-231723901 on Build GRCh38
rs537144453 Chr.1: 231723911-231723911 on Build GRCh38
rs949915729 Chr.1: 231724269-231724269 on Build GRCh38
rs568002263 Chr.1: 231723869-231723869 on Build GRCh38
rs961517996 Chr.1: 231723922-231723922 on Build GRCh38
rs905734776 Chr.1: 231724001-231724001 on Build GRCh38
rs1002580325 Chr.1: 231723841-231723841 on Build GRCh38
rs935645974 Chr.1: 231724215-231724215 on Build GRCh38
rs941098184 Chr.1: 231723862-231723862 on Build GRCh38
rs925634524 Chr.1: 231723963-231723963 on Build GRCh38
rs201760408 Chr.1: 231724242-231724242 on Build GRCh38
rs535789901 Chr.1: 231724260-231724260 on Build GRCh38
rs1014013985 Chr.1: 231723992-231723992 on Build GRCh38
rs559591962 Chr.1: 231723836-231723836 on Build GRCh38
rs573152727 Chr.1: 231724161-231724161 on Build GRCh38
rs929465218 Chr.1: 231724223-231724223 on Build GRCh38
rs748242232 Chr.1: 231723855-231723855 on Build GRCh38
rs547770430 Chr.1: 231723864-231723864 on Build GRCh38
rs747510781 Chr.1: 231723993-231723993 on Build GRCh38
rs570535516 Chr.1: 231724091-231724091 on Build GRCh38
rs922813649 Chr.1: 231723927-231723927 on Build GRCh38
rs1035305076 Chr.1: 231724165-231724165 on Build GRCh38
rs952675735 Chr.1: 231724078-231724078 on Build GRCh38
rs935781995 Chr.1: 231723972-231723972 on Build GRCh38
rs535200231 Chr.1: 231724236-231724236 on Build GRCh38
rs201897757 Chr.1: 231724176-231724176 on Build GRCh38
rs550355250 Chr.1: 231723977-231723977 on Build GRCh38
rs552192973 Chr.1: 231723902-231723902 on Build GRCh38
rs1001455453 Chr.1: 231724022-231724022 on Build GRCh38
rs200534864 Chr.1: 231723967-231723967 on Build GRCh38
rs149274936 Chr.1: 231724152-231724152 on Build GRCh38
rs922793702 Chr.1: 231724163-231724163 on Build GRCh38
rs977047306 Chr.1: 231723923-231723923 on Build GRCh38
rs778284866 Chr.1: 231723976-231723976 on Build GRCh38
rs891649795 Chr.1: 231723981-231723981 on Build GRCh38
rs187653977 Chr.1: 231724286-231724286 on Build GRCh38

View Full Product Details