Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 112438112-112438633 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZDHHC6
Entrez Gene ID
64429
Gene Name
zinc finger DHHC-type containing 6
Gene Aliases
DHHC-6, ZNF376
UniGene
Hs.196990
Gene Chromosome Location
Chr.10: 112424411-112448335 on Build GRCh38
Transcripts
NM_001303134.1 NM_022494.2 XM_005270051.4 XM_006717948.3 XM_006717949.3 XM_011540091.2 XM_011540092.2 XM_011540093.1 XM_011540094.2 XM_011540095.2 XM_017016561.1 XM_017016562.1 XM_017016563.1 XM_017016564.1 XM_017016565.1 XM_017016566.1 XM_017016567.1 XM_017016568.1 XM_017016569.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5817142 c.703A>G p.K235E Chr.10: 112438368-112438368 on Build GRCh38
5954197 c.732G>T p.E244D Chr.10: 112438339-112438339 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs185671421 Chr.10: 112438299-112438299 on Build GRCh38
rs552559862 Chr.10: 112438154-112438154 on Build GRCh38
rs762716412 Chr.10: 112438492-112438492 on Build GRCh38
rs553587968 Chr.10: 112438243-112438243 on Build GRCh38
rs539845271 Chr.10: 112438537-112438537 on Build GRCh38
rs916625519 Chr.10: 112438180-112438180 on Build GRCh38
rs181923674 Chr.10: 112438158-112438158 on Build GRCh38
rs763784762 Chr.10: 112438435-112438435 on Build GRCh38
rs1021166758 Chr.10: 112438375-112438375 on Build GRCh38
rs34823410 Chr.10: 112438345-112438345 on Build GRCh38
rs1009490146 Chr.10: 112438350-112438350 on Build GRCh38
rs982404878 Chr.10: 112438583-112438583 on Build GRCh38
rs772405523 Chr.10: 112438418-112438418 on Build GRCh38
rs1047024600 Chr.10: 112438241-112438241 on Build GRCh38
rs568707865 Chr.10: 112438255-112438255 on Build GRCh38
rs887971067 Chr.10: 112438503-112438503 on Build GRCh38
rs969453522 Chr.10: 112438212-112438213 on Build GRCh38
rs779528926 Chr.10: 112438310-112438310 on Build GRCh38
rs1015566462 Chr.10: 112438545-112438545 on Build GRCh38
rs964887078 Chr.10: 112438514-112438514 on Build GRCh38
rs748915545 Chr.10: 112438335-112438335 on Build GRCh38
rs754199359 Chr.10: 112438595-112438595 on Build GRCh38
rs942480170 Chr.10: 112438191-112438191 on Build GRCh38
rs975174721 Chr.10: 112438208-112438208 on Build GRCh38
rs116492857 Chr.10: 112438594-112438594 on Build GRCh38
rs3736059 Chr.10: 112438224-112438224 on Build GRCh38
rs1049210661 Chr.10: 112438429-112438429 on Build GRCh38
rs762275943 Chr.10: 112438410-112438410 on Build GRCh38
rs527934061 Chr.10: 112438451-112438451 on Build GRCh38
rs905017651 Chr.10: 112438300-112438300 on Build GRCh38
rs908423939 Chr.10: 112438249-112438249 on Build GRCh38
rs955030552 Chr.10: 112438578-112438578 on Build GRCh38
rs1001067276 Chr.10: 112438423-112438423 on Build GRCh38
rs979533167 Chr.10: 112438139-112438139 on Build GRCh38
rs910720402 Chr.10: 112438174-112438174 on Build GRCh38
rs922346257 Chr.10: 112438543-112438543 on Build GRCh38
rs935247110 Chr.10: 112438242-112438242 on Build GRCh38
rs774855252 Chr.10: 112438359-112438359 on Build GRCh38
rs1056529540 Chr.10: 112438327-112438327 on Build GRCh38
rs550178756 Chr.10: 112438229-112438229 on Build GRCh38

View Full Product Details