Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 11340737-11341236 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALNT18
Entrez Gene ID
374378
Gene Name
polypeptide N-acetylgalactosaminyltransferase 18
Gene Aliases
GALNACT18, GALNT15, GALNTL4, GalNAc-T15, GalNAc-T18
UniGene
Hs.655152
Gene Chromosome Location
Chr.11: 11270874-11622045 on Build GRCh38
Transcripts
NM_198516.2 XM_006718224.2 XM_011520068.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3710005 c.1276G>A p.E426K Chr.11: 11340821-11340821 on Build GRCh38
4775748 c.1234G>A p.E412K Chr.11: 11340863-11340863 on Build GRCh38
6567773 c.1209C>T p.L403L Chr.11: 11340888-11340888 on Build GRCh38
923228 c.1098G>A p.W366* Chr.11: 11340999-11340999 on Build GRCh38
4521278 c.1103G>T p.C368F Chr.11: 11340994-11340994 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs922814658 Chr.11: 11341125-11341125 on Build GRCh38
rs555837466 Chr.11: 11340810-11340810 on Build GRCh38
rs140071891 Chr.11: 11341143-11341143 on Build GRCh38
rs184878171 Chr.11: 11340788-11340788 on Build GRCh38
rs764348941 Chr.11: 11340864-11340864 on Build GRCh38
rs79582300 Chr.11: 11341086-11341086 on Build GRCh38
rs751976870 Chr.11: 11340962-11340962 on Build GRCh38
rs759476756 Chr.11: 11340843-11340843 on Build GRCh38
rs921059194 Chr.11: 11340888-11340888 on Build GRCh38
rs200976406 Chr.11: 11340856-11340856 on Build GRCh38
rs958446619 Chr.11: 11340769-11340769 on Build GRCh38
rs117391050 Chr.11: 11340806-11340806 on Build GRCh38
rs761875040 Chr.11: 11340949-11340949 on Build GRCh38
rs140776632 Chr.11: 11340837-11340837 on Build GRCh38
rs944779612 Chr.11: 11341182-11341182 on Build GRCh38
rs766308075 Chr.11: 11340776-11340776 on Build GRCh38
rs553486283 Chr.11: 11340944-11340944 on Build GRCh38
rs756342470 Chr.11: 11340789-11340789 on Build GRCh38
rs149181829 Chr.11: 11340959-11340959 on Build GRCh38
rs758936741 Chr.11: 11340958-11340958 on Build GRCh38
rs757746786 Chr.11: 11340800-11340800 on Build GRCh38
rs568922217 Chr.11: 11340821-11340821 on Build GRCh38
rs566902816 Chr.11: 11340803-11340803 on Build GRCh38
rs533662280 Chr.11: 11341130-11341130 on Build GRCh38
rs377098746 Chr.11: 11340937-11340937 on Build GRCh38
rs749344784 Chr.11: 11340795-11340795 on Build GRCh38
rs766284531 Chr.11: 11340826-11340826 on Build GRCh38
rs779358894 Chr.11: 11340890-11340890 on Build GRCh38
rs767429534 Chr.11: 11340848-11340848 on Build GRCh38
rs544802323 Chr.11: 11341090-11341090 on Build GRCh38
rs772636305 Chr.11: 11340823-11340823 on Build GRCh38
rs765338769 Chr.11: 11341034-11341034 on Build GRCh38
rs911280340 Chr.11: 11341150-11341150 on Build GRCh38
rs767916650 Chr.11: 11340785-11340785 on Build GRCh38
rs1001323384 Chr.11: 11341012-11341012 on Build GRCh38
rs770479060 Chr.11: 11340908-11340908 on Build GRCh38
rs750272887 Chr.11: 11340989-11340989 on Build GRCh38
rs772092466 Chr.11: 11340916-11340916 on Build GRCh38
rs867574644 Chr.11: 11341170-11341170 on Build GRCh38
rs750456085 Chr.11: 11340954-11340954 on Build GRCh38
rs773335179 Chr.11: 11341118-11341118 on Build GRCh38
rs944728907 Chr.11: 11340915-11340915 on Build GRCh38
rs936077495 Chr.11: 11341160-11341160 on Build GRCh38
rs751358045 Chr.11: 11340781-11340781 on Build GRCh38
rs745336160 Chr.11: 11340986-11340986 on Build GRCh38
rs756741253 Chr.11: 11340893-11340893 on Build GRCh38
rs755366124 Chr.11: 11340970-11340970 on Build GRCh38
rs757349269 Chr.11: 11340887-11340887 on Build GRCh38
rs754806772 Chr.11: 11340783-11340783 on Build GRCh38
rs989994623 Chr.11: 11341087-11341087 on Build GRCh38
rs189247074 Chr.11: 11341184-11341184 on Build GRCh38
rs74703597 Chr.11: 11340930-11340930 on Build GRCh38
rs752489000 Chr.11: 11340849-11340849 on Build GRCh38
rs79347342 Chr.11: 11341071-11341071 on Build GRCh38
rs76913025 Chr.11: 11341091-11341091 on Build GRCh38
rs934156017 Chr.11: 11341128-11341128 on Build GRCh38
rs764285689 Chr.11: 11340943-11340943 on Build GRCh38
rs951774360 Chr.11: 11341065-11341065 on Build GRCh38
rs1054236983 Chr.11: 11340885-11340885 on Build GRCh38
rs748816684 Chr.11: 11340906-11340906 on Build GRCh38
rs762117391 Chr.11: 11341009-11341009 on Build GRCh38
rs762821461 Chr.11: 11340772-11340772 on Build GRCh38
rs562624638 Chr.11: 11341080-11341080 on Build GRCh38
rs768505451 Chr.11: 11341007-11341007 on Build GRCh38
rs781343671 Chr.11: 11340976-11340976 on Build GRCh38
rs922742422 Chr.11: 11340830-11340830 on Build GRCh38
rs115125023 Chr.11: 11340911-11340911 on Build GRCh38
rs769789482 Chr.11: 11340950-11340950 on Build GRCh38
rs755903986 Chr.11: 11340990-11340990 on Build GRCh38
rs577502852 Chr.11: 11340912-11340912 on Build GRCh38
rs760618046 Chr.11: 11340996-11340996 on Build GRCh38
rs777467014 Chr.11: 11340902-11340902 on Build GRCh38
rs780515661 Chr.11: 11340961-11340961 on Build GRCh38
rs566710091 Chr.11: 11341178-11341178 on Build GRCh38
rs775765910 Chr.11: 11340807-11340807 on Build GRCh38
rs150122075 Chr.11: 11340901-11340901 on Build GRCh38
rs780730440 Chr.11: 11340894-11340894 on Build GRCh38
rs747173548 Chr.11: 11340776-11340777 on Build GRCh38
rs776231748 Chr.11: 11341008-11341008 on Build GRCh38
rs748979478 Chr.11: 11340979-11340979 on Build GRCh38
rs770437636 Chr.11: 11340982-11340982 on Build GRCh38
rs372315275 Chr.11: 11340805-11340805 on Build GRCh38
rs146437361 Chr.11: 11340825-11340825 on Build GRCh38
rs753916277 Chr.11: 11340866-11340866 on Build GRCh38
rs778594872 Chr.11: 11340984-11340984 on Build GRCh38
rs59816464 Chr.11: 11341037-11341037 on Build GRCh38

View Full Product Details