Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 18245753-18246292 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SAA2
Entrez Gene ID
6289
Gene Name
serum amyloid A2
Gene Aliases
-
UniGene
Hs.731376
Gene Chromosome Location
Chr.11: 18238233-18248674 on Build GRCh38
Transcripts
NM_001127380.2 NM_030754.4 NM_001199744.1

Gene Symbol
SAA2-SAA4
Entrez Gene ID
100528017
Gene Name
SAA2-SAA4 readthrough
Gene Aliases
-
UniGene
Hs.731376
Gene Chromosome Location
Chr.11: 18231355-18248674 on Build GRCh38
Transcripts
NM_001127380.2 NM_030754.4 NM_001199744.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4563981 c.99G>A p.R33R Chr.11: 18246041-18246041 on Build GRCh38
4411985 c.92-5T>G p.? Chr.11: 18246053-18246053 on Build GRCh38
6131890 c.208G>T p.A70S Chr.11: 18245932-18245932 on Build GRCh38
1561474 c.137A>C p.N46T Chr.11: 18246003-18246003 on Build GRCh38
4145612 c.224T>C p.V75A Chr.11: 18245916-18245916 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767805711 Chr.11: 18245898-18245898 on Build GRCh38
rs752256509 Chr.11: 18245991-18245991 on Build GRCh38
rs779503471 Chr.11: 18246012-18246012 on Build GRCh38
rs200930008 Chr.11: 18246053-18246053 on Build GRCh38
rs879005028 Chr.11: 18245908-18245908 on Build GRCh38
rs767218643 Chr.11: 18246045-18246046 on Build GRCh38
rs202167679 Chr.11: 18246050-18246051 on Build GRCh38
rs768875822 Chr.11: 18246042-18246042 on Build GRCh38
rs775562403 Chr.11: 18245952-18245952 on Build GRCh38
rs753201171 Chr.11: 18246064-18246065 on Build GRCh38
rs367983868 Chr.11: 18246038-18246038 on Build GRCh38
rs766267295 Chr.11: 18245971-18245971 on Build GRCh38
rs753669559 Chr.11: 18245899-18245899 on Build GRCh38
rs571096139 Chr.11: 18245934-18245934 on Build GRCh38
rs71469388 Chr.11: 18245916-18245916 on Build GRCh38
rs201840682 Chr.11: 18246067-18246068 on Build GRCh38
rs1044068853 Chr.11: 18246016-18246016 on Build GRCh38
rs576273360 Chr.11: 18246185-18246185 on Build GRCh38
rs752707688 Chr.11: 18245797-18245797 on Build GRCh38
rs755751426 Chr.11: 18246086-18246086 on Build GRCh38
rs373402346 Chr.11: 18246015-18246015 on Build GRCh38
rs200272839 Chr.11: 18246122-18246122 on Build GRCh38
rs949717238 Chr.11: 18246008-18246008 on Build GRCh38
rs78891342 Chr.11: 18245887-18245887 on Build GRCh38
rs758832111 Chr.11: 18246068-18246068 on Build GRCh38
rs767262411 Chr.11: 18245989-18245989 on Build GRCh38
rs759857504 Chr.11: 18245897-18245897 on Build GRCh38
rs761456152 Chr.11: 18245901-18245901 on Build GRCh38
rs144459542 Chr.11: 18246052-18246052 on Build GRCh38
rs756547312 Chr.11: 18245993-18245993 on Build GRCh38
rs757842152 Chr.11: 18246010-18246010 on Build GRCh38
rs375627268 Chr.11: 18246146-18246146 on Build GRCh38
rs754431086 Chr.11: 18245987-18245987 on Build GRCh38
rs113441609 Chr.11: 18245909-18245909 on Build GRCh38
rs1671927 Chr.11: 18245826-18245826 on Build GRCh38
rs527844633 Chr.11: 18245923-18245923 on Build GRCh38
rs375310102 Chr.11: 18246064-18246064 on Build GRCh38
rs771099289 Chr.11: 18245890-18245890 on Build GRCh38
rs751076942 Chr.11: 18245985-18245985 on Build GRCh38
rs374508530 Chr.11: 18245995-18245995 on Build GRCh38
rs1136766 Chr.11: 18245924-18245924 on Build GRCh38
rs766911330 Chr.11: 18246059-18246059 on Build GRCh38
rs771513906 Chr.11: 18245930-18245930 on Build GRCh38
rs138605229 Chr.11: 18245918-18245918 on Build GRCh38
rs779519299 Chr.11: 18245911-18245911 on Build GRCh38
rs4757631 Chr.11: 18246155-18246155 on Build GRCh38
rs769059305 Chr.11: 18245882-18245882 on Build GRCh38
rs2445172 Chr.11: 18245806-18245806 on Build GRCh38
rs2445171 Chr.11: 18245809-18245809 on Build GRCh38
rs1009438129 Chr.11: 18246217-18246217 on Build GRCh38
rs746001826 Chr.11: 18246014-18246014 on Build GRCh38
rs201251084 Chr.11: 18245889-18245889 on Build GRCh38
rs386751154 Chr.11: 18246123-18246124 on Build GRCh38
rs376452063 Chr.11: 18245874-18245874 on Build GRCh38
rs762672586 Chr.11: 18246043-18246043 on Build GRCh38
rs765275938 Chr.11: 18245906-18245906 on Build GRCh38
rs576283026 Chr.11: 18245838-18245838 on Build GRCh38
rs576320118 Chr.11: 18245852-18245852 on Build GRCh38
rs868572530 Chr.11: 18245940-18245940 on Build GRCh38
rs964733958 Chr.11: 18246233-18246233 on Build GRCh38
rs750424628 Chr.11: 18245968-18245968 on Build GRCh38
rs751024797 Chr.11: 18245912-18245912 on Build GRCh38
rs905575783 Chr.11: 18246019-18246019 on Build GRCh38
rs1671923 Chr.11: 18246104-18246104 on Build GRCh38
rs978655376 Chr.11: 18246267-18246267 on Build GRCh38
rs878994849 Chr.11: 18245910-18245910 on Build GRCh38
rs1017611373 Chr.11: 18246231-18246231 on Build GRCh38
rs772436713 Chr.11: 18245944-18245944 on Build GRCh38
rs764493342 Chr.11: 18246070-18246070 on Build GRCh38
rs890549956 Chr.11: 18246194-18246194 on Build GRCh38
rs749623042 Chr.11: 18245996-18245996 on Build GRCh38
rs146160224 Chr.11: 18245926-18245926 on Build GRCh38
rs764910787 Chr.11: 18245961-18245961 on Build GRCh38
rs550132053 Chr.11: 18246017-18246017 on Build GRCh38
rs774759478 Chr.11: 18245893-18245893 on Build GRCh38
rs746635254 Chr.11: 18245876-18245876 on Build GRCh38
rs748564206 Chr.11: 18245921-18245921 on Build GRCh38
rs75350417 Chr.11: 18245888-18245888 on Build GRCh38
rs76546965 Chr.11: 18245884-18245884 on Build GRCh38
rs765642518 Chr.11: 18246051-18246055 on Build GRCh38
rs796319938 Chr.11: 18245887-18245888 on Build GRCh38
rs762902088 Chr.11: 18245970-18245970 on Build GRCh38
rs761735429 Chr.11: 18245959-18245959 on Build GRCh38
rs769464603 Chr.11: 18246059-18246068 on Build GRCh38
rs978707492 Chr.11: 18245789-18245789 on Build GRCh38
rs759083650 Chr.11: 18246058-18246058 on Build GRCh38
rs752199546 Chr.11: 18246060-18246060 on Build GRCh38
rs79620496 Chr.11: 18245931-18245931 on Build GRCh38
rs993695207 Chr.11: 18246131-18246131 on Build GRCh38
rs539592580 Chr.11: 18246124-18246124 on Build GRCh38

View Full Product Details