Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 27055314-27055826 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BBOX1
Entrez Gene ID
8424
Gene Name
gamma-butyrobetaine hydroxylase 1
Gene Aliases
BBH, BBOX, G-BBH, gamma-BBH
UniGene
Hs.591996
Gene Chromosome Location
Chr.11: 27040705-27127807 on Build GRCh38
Transcripts
NM_003986.2 XM_005253159.2 XM_005253160.2 XM_005253161.2 XM_011520402.1 XM_017018403.1 NR_125768.1

Gene Symbol
BBOX1-AS1
Entrez Gene ID
103695435
Gene Name
BBOX1 antisense RNA 1
Gene Aliases
-
UniGene
Hs.196133
Gene Chromosome Location
Chr.11: 27047186-27220113 on Build GRCh38
Transcripts
NM_003986.2 XM_005253159.2 XM_005253160.2 XM_005253161.2 XM_011520402.1 XM_017018403.1 NR_125768.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4775644 c.76G>C p.E26Q Chr.11: 27055506-27055506 on Build GRCh38
687634 c.175G>C p.D59H Chr.11: 27055605-27055605 on Build GRCh38
120836 c.177T>A p.D59E Chr.11: 27055607-27055607 on Build GRCh38
3670748 c.101T>A p.L34* Chr.11: 27055531-27055531 on Build GRCh38
1297941 c.180G>A p.V60V Chr.11: 27055610-27055610 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs184088604 Chr.11: 27055769-27055769 on Build GRCh38
rs575442166 Chr.11: 27055594-27055594 on Build GRCh38
rs765894356 Chr.11: 27055507-27055507 on Build GRCh38
rs138619370 Chr.11: 27055565-27055565 on Build GRCh38
rs148033507 Chr.11: 27055438-27055438 on Build GRCh38
rs10767598 Chr.11: 27055571-27055571 on Build GRCh38
rs368465428 Chr.11: 27055437-27055437 on Build GRCh38
rs556658970 Chr.11: 27055530-27055530 on Build GRCh38
rs779558464 Chr.11: 27055412-27055412 on Build GRCh38
rs187949127 Chr.11: 27055582-27055582 on Build GRCh38
rs551267414 Chr.11: 27055429-27055429 on Build GRCh38
rs574993717 Chr.11: 27055773-27055773 on Build GRCh38
rs536072159 Chr.11: 27055415-27055415 on Build GRCh38
rs749354502 Chr.11: 27055672-27055672 on Build GRCh38
rs759343729 Chr.11: 27055440-27055440 on Build GRCh38
rs941773281 Chr.11: 27055614-27055614 on Build GRCh38
rs200195953 Chr.11: 27055467-27055467 on Build GRCh38
rs975719573 Chr.11: 27055496-27055496 on Build GRCh38
rs746920195 Chr.11: 27055520-27055520 on Build GRCh38
rs764775193 Chr.11: 27055606-27055606 on Build GRCh38
rs761196390 Chr.11: 27055483-27055483 on Build GRCh38
rs768633123 Chr.11: 27055432-27055432 on Build GRCh38
rs147395380 Chr.11: 27055742-27055742 on Build GRCh38
rs761374971 Chr.11: 27055501-27055502 on Build GRCh38
rs1013909822 Chr.11: 27055423-27055423 on Build GRCh38
rs907459017 Chr.11: 27055743-27055743 on Build GRCh38
rs933190374 Chr.11: 27055519-27055519 on Build GRCh38
rs773228917 Chr.11: 27055738-27055738 on Build GRCh38
rs757839184 Chr.11: 27055406-27055406 on Build GRCh38
rs752818562 Chr.11: 27055662-27055662 on Build GRCh38
rs941447811 Chr.11: 27055573-27055573 on Build GRCh38
rs528123757 Chr.11: 27055754-27055754 on Build GRCh38
rs141755764 Chr.11: 27055473-27055473 on Build GRCh38
rs758517397 Chr.11: 27055425-27055425 on Build GRCh38
rs781503812 Chr.11: 27055658-27055658 on Build GRCh38
rs776350781 Chr.11: 27055692-27055692 on Build GRCh38
rs12798621 Chr.11: 27055746-27055746 on Build GRCh38
rs776124862 Chr.11: 27055586-27055586 on Build GRCh38
rs1041373075 Chr.11: 27055706-27055706 on Build GRCh38
rs938216552 Chr.11: 27055649-27055649 on Build GRCh38
rs905410094 Chr.11: 27055343-27055343 on Build GRCh38
rs767202576 Chr.11: 27055503-27055505 on Build GRCh38
rs1015614325 Chr.11: 27055789-27055789 on Build GRCh38
rs12049843 Chr.11: 27055785-27055785 on Build GRCh38
rs766992020 Chr.11: 27055648-27055648 on Build GRCh38
rs746869939 Chr.11: 27055684-27055684 on Build GRCh38
rs777770420 Chr.11: 27055671-27055671 on Build GRCh38
rs778849057 Chr.11: 27055546-27055546 on Build GRCh38
rs144522986 Chr.11: 27055547-27055547 on Build GRCh38
rs896782001 Chr.11: 27055668-27055668 on Build GRCh38
rs763638880 Chr.11: 27055476-27055476 on Build GRCh38
rs769667333 Chr.11: 27055587-27055587 on Build GRCh38
rs888761036 Chr.11: 27055612-27055612 on Build GRCh38
rs756186779 Chr.11: 27055663-27055663 on Build GRCh38
rs751858108 Chr.11: 27055528-27055528 on Build GRCh38
rs899652957 Chr.11: 27055765-27055765 on Build GRCh38
rs372006148 Chr.11: 27055390-27055390 on Build GRCh38
rs202054890 Chr.11: 27055536-27055536 on Build GRCh38
rs974238451 Chr.11: 27055603-27055603 on Build GRCh38
rs755482560 Chr.11: 27055658-27055673 on Build GRCh38
rs765427088 Chr.11: 27055352-27055352 on Build GRCh38
rs188738260 Chr.11: 27055788-27055788 on Build GRCh38
rs779823808 Chr.11: 27055673-27055673 on Build GRCh38
rs2305095 Chr.11: 27055430-27055430 on Build GRCh38
rs768287336 Chr.11: 27055690-27055690 on Build GRCh38
rs751997909 Chr.11: 27055650-27055650 on Build GRCh38
rs763615718 Chr.11: 27055624-27055624 on Build GRCh38
rs772925558 Chr.11: 27055701-27055701 on Build GRCh38
rs750982836 Chr.11: 27055607-27055607 on Build GRCh38
rs758839124 Chr.11: 27055522-27055522 on Build GRCh38
rs780510064 Chr.11: 27055526-27055526 on Build GRCh38
rs555454326 Chr.11: 27055538-27055538 on Build GRCh38
rs377355425 Chr.11: 27055384-27055384 on Build GRCh38
rs754381876 Chr.11: 27055582-27055582 on Build GRCh38
rs753452828 Chr.11: 27055482-27055482 on Build GRCh38
rs747745913 Chr.11: 27055696-27055696 on Build GRCh38
rs540166569 Chr.11: 27055471-27055471 on Build GRCh38
rs376941503 Chr.11: 27055404-27055404 on Build GRCh38
rs769557990 Chr.11: 27055699-27055699 on Build GRCh38
rs56349897 Chr.11: 27055342-27055342 on Build GRCh38
rs746274716 Chr.11: 27055424-27055424 on Build GRCh38

View Full Product Details