Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 65013803-65014294 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL2
Entrez Gene ID
402
Gene Name
ADP ribosylation factor like GTPase 2
Gene Aliases
ARFL2
UniGene
Hs.502836
Gene Chromosome Location
Chr.11: 65014113-65022185 on Build GRCh38
Transcripts
NM_001199745.1 NM_001667.3 NR_037650.1

Gene Symbol
ARL2-SNX15
Entrez Gene ID
100528018
Gene Name
ARL2-SNX15 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.80132
Gene Chromosome Location
Chr.11: 65014113-65040572 on Build GRCh38
Transcripts
NM_001199745.1 NM_001667.3 NR_037650.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2164883 c.58C>G p.L20V Chr.11: 65014265-65014265 on Build GRCh38
6352007 c.64C>G p.L22V Chr.11: 65014271-65014271 on Build GRCh38
5373245 c.48G>A p.E16E Chr.11: 65014255-65014255 on Build GRCh38
7114837 c.19C>T p.L7L Chr.11: 65014226-65014226 on Build GRCh38
7114836 c.9C>G p.L3L Chr.11: 65014216-65014216 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1041760917 Chr.11: 65014005-65014005 on Build GRCh38
rs561992738 Chr.11: 65014146-65014146 on Build GRCh38
rs1012721797 Chr.11: 65014045-65014045 on Build GRCh38
rs376174797 Chr.11: 65013988-65013988 on Build GRCh38
rs1027795969 Chr.11: 65014023-65014023 on Build GRCh38
rs567248938 Chr.11: 65013952-65013952 on Build GRCh38
rs78852382 Chr.11: 65013840-65013840 on Build GRCh38
rs1054557003 Chr.11: 65014125-65014125 on Build GRCh38
rs993457667 Chr.11: 65014058-65014058 on Build GRCh38
rs893637831 Chr.11: 65013994-65013994 on Build GRCh38
rs1021307631 Chr.11: 65013999-65013999 on Build GRCh38
rs765723064 Chr.11: 65014212-65014212 on Build GRCh38
rs777969505 Chr.11: 65014164-65014164 on Build GRCh38
rs751817327 Chr.11: 65014165-65014165 on Build GRCh38
rs781160125 Chr.11: 65014051-65014051 on Build GRCh38
rs545530857 Chr.11: 65014011-65014011 on Build GRCh38
rs749190023 Chr.11: 65014192-65014193 on Build GRCh38
rs746698417 Chr.11: 65014200-65014200 on Build GRCh38
rs552940678 Chr.11: 65014038-65014038 on Build GRCh38
rs972466656 Chr.11: 65013934-65013934 on Build GRCh38
rs756343837 Chr.11: 65014267-65014267 on Build GRCh38
rs1010794478 Chr.11: 65013997-65013997 on Build GRCh38
rs779917314 Chr.11: 65014176-65014176 on Build GRCh38
rs112166431 Chr.11: 65014198-65014198 on Build GRCh38
rs958500244 Chr.11: 65013851-65013851 on Build GRCh38
rs777462885 Chr.11: 65014208-65014208 on Build GRCh38
rs1056861268 Chr.11: 65014020-65014020 on Build GRCh38
rs1026661823 Chr.11: 65014062-65014062 on Build GRCh38
rs752054244 Chr.11: 65013992-65013992 on Build GRCh38
rs910653375 Chr.11: 65013966-65013966 on Build GRCh38
rs755841395 Chr.11: 65014172-65014172 on Build GRCh38
rs932020100 Chr.11: 65014121-65014121 on Build GRCh38
rs778508915 Chr.11: 65014188-65014188 on Build GRCh38
rs576916846 Chr.11: 65014000-65014000 on Build GRCh38
rs938322910 Chr.11: 65014236-65014236 on Build GRCh38
rs374015070 Chr.11: 65014262-65014262 on Build GRCh38
rs893032263 Chr.11: 65013824-65013831 on Build GRCh38
rs145706074 Chr.11: 65013962-65013962 on Build GRCh38
rs753112407 Chr.11: 65013854-65013854 on Build GRCh38
rs753679803 Chr.11: 65014264-65014264 on Build GRCh38
rs987879229 Chr.11: 65014097-65014097 on Build GRCh38
rs894233149 Chr.11: 65014032-65014032 on Build GRCh38
rs762992082 Chr.11: 65014233-65014233 on Build GRCh38
rs759906878 Chr.11: 65014209-65014209 on Build GRCh38
rs1016139738 Chr.11: 65014049-65014049 on Build GRCh38
rs990539151 Chr.11: 65014022-65014022 on Build GRCh38
rs915431633 Chr.11: 65014152-65014152 on Build GRCh38
rs921929122 Chr.11: 65014120-65014120 on Build GRCh38
rs368381614 Chr.11: 65013901-65013901 on Build GRCh38
rs990947557 Chr.11: 65013853-65013853 on Build GRCh38
rs113298333 Chr.11: 65014273-65014273 on Build GRCh38
rs572718601 Chr.11: 65014054-65014054 on Build GRCh38
rs989602613 Chr.11: 65014105-65014105 on Build GRCh38
rs910600929 Chr.11: 65014102-65014102 on Build GRCh38
rs954709777 Chr.11: 65014066-65014066 on Build GRCh38
rs749136746 Chr.11: 65014187-65014187 on Build GRCh38
rs542131784 Chr.11: 65014060-65014060 on Build GRCh38
rs750611552 Chr.11: 65014104-65014104 on Build GRCh38
rs373234095 Chr.11: 65013977-65013977 on Build GRCh38
rs757327583 Chr.11: 65014004-65014004 on Build GRCh38
rs866184217 Chr.11: 65014013-65014013 on Build GRCh38
rs796855950 Chr.11: 65013832-65013832 on Build GRCh38
rs766018470 Chr.11: 65014259-65014259 on Build GRCh38
rs764204642 Chr.11: 65014235-65014235 on Build GRCh38
rs371780654 Chr.11: 65013993-65013993 on Build GRCh38
rs184873330 Chr.11: 65014251-65014251 on Build GRCh38
rs772978493 Chr.11: 65014199-65014199 on Build GRCh38
rs1040428498 Chr.11: 65013983-65013983 on Build GRCh38
rs771161160 Chr.11: 65014201-65014201 on Build GRCh38
rs763473572 Chr.11: 65013920-65013920 on Build GRCh38
rs193245768 Chr.11: 65013910-65013910 on Build GRCh38
rs550533269 Chr.11: 65013985-65013985 on Build GRCh38
rs369937985 Chr.11: 65014157-65014157 on Build GRCh38
rs959078356 Chr.11: 65014017-65014017 on Build GRCh38
rs919288699 Chr.11: 65013948-65013948 on Build GRCh38
rs867567492 Chr.11: 65014230-65014230 on Build GRCh38
rs959141997 Chr.11: 65013842-65013842 on Build GRCh38
rs776046018 Chr.11: 65014231-65014231 on Build GRCh38

View Full Product Details