Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 65351605-65352104 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPF2
Entrez Gene ID
5977
Gene Name
double PHD fingers 2
Gene Aliases
REQ, UBID4, ubi-d4
UniGene
Hs.13495
Gene Chromosome Location
Chr.11: 65333754-65352980 on Build GRCh38
Transcripts
NM_006268.4 XM_005274149.2 XM_017018101.1 XM_017018102.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1704239 c.1172C>T p.S391F Chr.11: 65351755-65351755 on Build GRCh38
3451772 c.1149C>T p.S383S Chr.11: 65351732-65351732 on Build GRCh38
6365104 c.1167C>A p.N389K Chr.11: 65351750-65351750 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs781511855 Chr.11: 65351784-65351784 on Build GRCh38
rs772541919 Chr.11: 65351672-65351672 on Build GRCh38
rs201395223 Chr.11: 65351705-65351705 on Build GRCh38
rs1042558862 Chr.11: 65351947-65351947 on Build GRCh38
rs555296733 Chr.11: 65351982-65351982 on Build GRCh38
rs778618525 Chr.11: 65351800-65351803 on Build GRCh38
rs895366896 Chr.11: 65351985-65351985 on Build GRCh38
rs760241195 Chr.11: 65351734-65351734 on Build GRCh38
rs959363028 Chr.11: 65351736-65351736 on Build GRCh38
rs932340156 Chr.11: 65351885-65351885 on Build GRCh38
rs1035013085 Chr.11: 65351647-65351647 on Build GRCh38
rs961199169 Chr.11: 65352072-65352072 on Build GRCh38
rs922304147 Chr.11: 65351880-65351880 on Build GRCh38
rs746234689 Chr.11: 65351919-65351919 on Build GRCh38
rs1028044310 Chr.11: 65352025-65352025 on Build GRCh38
rs920697017 Chr.11: 65351761-65351762 on Build GRCh38
rs752174476 Chr.11: 65351864-65351864 on Build GRCh38
rs983927094 Chr.11: 65352054-65352054 on Build GRCh38
rs372284211 Chr.11: 65352021-65352021 on Build GRCh38
rs765911720 Chr.11: 65351739-65351739 on Build GRCh38
rs575446528 Chr.11: 65351992-65351992 on Build GRCh38
rs761879455 Chr.11: 65351659-65351659 on Build GRCh38
rs774405814 Chr.11: 65351652-65351652 on Build GRCh38
rs778915788 Chr.11: 65351796-65351796 on Build GRCh38
rs547060405 Chr.11: 65351977-65351977 on Build GRCh38
rs35387404 Chr.11: 65351945-65351945 on Build GRCh38
rs757388869 Chr.11: 65351781-65351781 on Build GRCh38
rs758009351 Chr.11: 65352029-65352029 on Build GRCh38
rs199638754 Chr.11: 65351836-65351838 on Build GRCh38
rs566661095 Chr.11: 65351950-65351950 on Build GRCh38
rs768537493 Chr.11: 65351791-65351791 on Build GRCh38
rs751527501 Chr.11: 65351770-65351770 on Build GRCh38
rs759872695 Chr.11: 65351750-65351750 on Build GRCh38
rs976656328 Chr.11: 65351869-65351869 on Build GRCh38
rs1015468471 Chr.11: 65352068-65352068 on Build GRCh38
rs902677223 Chr.11: 65351970-65351970 on Build GRCh38
rs768590373 Chr.11: 65351632-65351632 on Build GRCh38
rs1049828296 Chr.11: 65351888-65351888 on Build GRCh38
rs369343587 Chr.11: 65351971-65351971 on Build GRCh38
rs753511183 Chr.11: 65351744-65351744 on Build GRCh38
rs546668385 Chr.11: 65351928-65351928 on Build GRCh38
rs539496082 Chr.11: 65351829-65351829 on Build GRCh38
rs893571116 Chr.11: 65351908-65351908 on Build GRCh38
rs908355221 Chr.11: 65351785-65351785 on Build GRCh38
rs111991224 Chr.11: 65351716-65351716 on Build GRCh38
rs753199658 Chr.11: 65351758-65351758 on Build GRCh38
rs759394018 Chr.11: 65351650-65351650 on Build GRCh38
rs570504731 Chr.11: 65351804-65351804 on Build GRCh38
rs764748473 Chr.11: 65351788-65351788 on Build GRCh38
rs754909793 Chr.11: 65351768-65351768 on Build GRCh38
rs765632713 Chr.11: 65351752-65351752 on Build GRCh38
rs767043735 Chr.11: 65351662-65351662 on Build GRCh38
rs758885245 Chr.11: 65351768-65351768 on Build GRCh38

View Full Product Details