Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 65539604-65540092 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LTBP3
Entrez Gene ID
4054
Gene Name
latent transforming growth factor beta binding protein 3
Gene Aliases
DASS, LTBP-3, LTBP2, STHAG6, pp6425
UniGene
Hs.289019
Gene Chromosome Location
Chr.11: 65538559-65558388 on Build GRCh38
Transcripts
NM_001130144.2 NM_001164266.1 NM_021070.4 XM_011545032.2 XM_011545033.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4035414 c.3407-1G>T p.? Chr.11: 65539629-65539629 on Build GRCh38
6425569 c.3389G>A p.R1130H Chr.11: 65539878-65539878 on Build GRCh38
6425570 c.3248G>A p.R1083H Chr.11: 65539878-65539878 on Build GRCh38
5630252 c.3402C>T p.G1134G Chr.11: 65539724-65539724 on Build GRCh38
5447954 c.3399C>T p.R1133R Chr.11: 65539727-65539727 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781035535 Chr.11: 65539638-65539638 on Build GRCh38
rs558144911 Chr.11: 65539807-65539807 on Build GRCh38
rs915915394 Chr.11: 65539970-65539970 on Build GRCh38
rs763932569 Chr.11: 65539794-65539794 on Build GRCh38
rs754312507 Chr.11: 65539902-65539902 on Build GRCh38
rs371428846 Chr.11: 65539627-65539627 on Build GRCh38
rs751488209 Chr.11: 65539707-65539707 on Build GRCh38
rs891777322 Chr.11: 65540026-65540026 on Build GRCh38
rs780579883 Chr.11: 65539744-65539744 on Build GRCh38
rs773233610 Chr.11: 65539768-65539768 on Build GRCh38
rs901290783 Chr.11: 65539684-65539684 on Build GRCh38
rs369207441 Chr.11: 65539906-65539906 on Build GRCh38
rs776771520 Chr.11: 65539909-65539909 on Build GRCh38
rs752365016 Chr.11: 65539689-65539689 on Build GRCh38
rs911333789 Chr.11: 65539995-65539995 on Build GRCh38
rs1054379851 Chr.11: 65539905-65539905 on Build GRCh38
rs939910538 Chr.11: 65539866-65539866 on Build GRCh38
rs1024449167 Chr.11: 65539918-65539918 on Build GRCh38
rs779328140 Chr.11: 65539760-65539760 on Build GRCh38
rs922472115 Chr.11: 65539978-65539978 on Build GRCh38
rs907434061 Chr.11: 65540047-65540047 on Build GRCh38
rs988974106 Chr.11: 65539817-65539817 on Build GRCh38
rs143200916 Chr.11: 65540045-65540045 on Build GRCh38
rs770989324 Chr.11: 65539907-65539907 on Build GRCh38
rs762957783 Chr.11: 65539700-65539700 on Build GRCh38
rs907180740 Chr.11: 65539816-65539816 on Build GRCh38
rs569342451 Chr.11: 65539721-65539721 on Build GRCh38
rs934107346 Chr.11: 65539884-65539884 on Build GRCh38
rs534239532 Chr.11: 65539832-65539832 on Build GRCh38
rs763745752 Chr.11: 65539942-65539942 on Build GRCh38
rs750972026 Chr.11: 65539623-65539623 on Build GRCh38
rs887593623 Chr.11: 65539987-65539987 on Build GRCh38
rs889758877 Chr.11: 65539740-65539740 on Build GRCh38
rs748632960 Chr.11: 65539869-65539869 on Build GRCh38
rs761857145 Chr.11: 65540018-65540018 on Build GRCh38
rs373698568 Chr.11: 65539663-65539663 on Build GRCh38
rs768369471 Chr.11: 65539933-65539933 on Build GRCh38
rs760835865 Chr.11: 65539676-65539676 on Build GRCh38
rs755347057 Chr.11: 65539847-65539847 on Build GRCh38
rs1045991961 Chr.11: 65540040-65540040 on Build GRCh38
rs953592405 Chr.11: 65539930-65539930 on Build GRCh38
rs914746124 Chr.11: 65539803-65539803 on Build GRCh38
rs947916882 Chr.11: 65539976-65539976 on Build GRCh38
rs1055841750 Chr.11: 65539966-65539966 on Build GRCh38
rs375395244 Chr.11: 65539654-65539654 on Build GRCh38
rs748664824 Chr.11: 65539763-65539763 on Build GRCh38
rs767143287 Chr.11: 65539808-65539808 on Build GRCh38
rs187689518 Chr.11: 65539668-65539668 on Build GRCh38
rs578222622 Chr.11: 65539809-65539809 on Build GRCh38
rs149115544 Chr.11: 65539637-65539637 on Build GRCh38
rs1012083121 Chr.11: 65539834-65539834 on Build GRCh38
rs922390571 Chr.11: 65539873-65539873 on Build GRCh38
rs779288121 Chr.11: 65539859-65539859 on Build GRCh38
rs763991235 Chr.11: 65539705-65539705 on Build GRCh38
rs938686900 Chr.11: 65539951-65539951 on Build GRCh38
rs777159634 Chr.11: 65539668-65539669 on Build GRCh38
rs753499466 Chr.11: 65539835-65539835 on Build GRCh38
rs747233808 Chr.11: 65539769-65539769 on Build GRCh38
rs970508468 Chr.11: 65539827-65539827 on Build GRCh38
rs772160343 Chr.11: 65539766-65539766 on Build GRCh38
rs983770970 Chr.11: 65539844-65539844 on Build GRCh38
rs972621001 Chr.11: 65539977-65539977 on Build GRCh38
rs1028583414 Chr.11: 65540060-65540060 on Build GRCh38
rs758834505 Chr.11: 65539885-65539885 on Build GRCh38
rs967970194 Chr.11: 65539938-65539938 on Build GRCh38
rs549912712 Chr.11: 65539693-65539693 on Build GRCh38
rs766787396 Chr.11: 65539829-65539829 on Build GRCh38
rs1002150082 Chr.11: 65540056-65540056 on Build GRCh38
rs770307972 Chr.11: 65539666-65539666 on Build GRCh38
rs771349317 Chr.11: 65539657-65539658 on Build GRCh38
rs926322628 Chr.11: 65539830-65539830 on Build GRCh38
rs889863271 Chr.11: 65539886-65539886 on Build GRCh38
rs536307181 Chr.11: 65539673-65539673 on Build GRCh38
rs542670094 Chr.11: 65539964-65539964 on Build GRCh38
rs1033870849 Chr.11: 65539936-65539936 on Build GRCh38
rs1000626384 Chr.11: 65539897-65539897 on Build GRCh38
rs765045821 Chr.11: 65539679-65539679 on Build GRCh38
rs1024395398 Chr.11: 65539820-65539820 on Build GRCh38
rs951867225 Chr.11: 65539702-65539702 on Build GRCh38
rs771081037 Chr.11: 65539775-65539775 on Build GRCh38
rs373015000 Chr.11: 65539674-65539674 on Build GRCh38
rs1048759666 Chr.11: 65539659-65539659 on Build GRCh38
rs1054389826 Chr.11: 65540014-65540014 on Build GRCh38
rs933844755 Chr.11: 65539980-65539980 on Build GRCh38
rs1015106416 Chr.11: 65539903-65539904 on Build GRCh38
rs538479771 Chr.11: 65539748-65539748 on Build GRCh38
rs756880007 Chr.11: 65539708-65539708 on Build GRCh38
rs774131721 Chr.11: 65539989-65539989 on Build GRCh38
rs878853036 Chr.11: 65540034-65540034 on Build GRCh38
rs983821625 Chr.11: 65539947-65539947 on Build GRCh38
rs965722320 Chr.11: 65539904-65539904 on Build GRCh38
rs769061467 Chr.11: 65539753-65539753 on Build GRCh38
rs1036873338 Chr.11: 65539871-65539871 on Build GRCh38
rs936055103 Chr.11: 65540004-65540004 on Build GRCh38
rs781038627 Chr.11: 65539670-65539670 on Build GRCh38
rs1049561409 Chr.11: 65539988-65539988 on Build GRCh38
rs755762566 Chr.11: 65539726-65539726 on Build GRCh38
rs773471184 Chr.11: 65539675-65539675 on Build GRCh38
rs956271105 Chr.11: 65539790-65539790 on Build GRCh38
rs776895791 Chr.11: 65539787-65539787 on Build GRCh38
rs762718826 Chr.11: 65539789-65539789 on Build GRCh38
rs909513638 Chr.11: 65539956-65539956 on Build GRCh38
rs893256135 Chr.11: 65539624-65539624 on Build GRCh38
rs1028327090 Chr.11: 65539690-65539690 on Build GRCh38
rs11227217 Chr.11: 65539931-65539931 on Build GRCh38
rs377361170 Chr.11: 65539711-65539711 on Build GRCh38
rs922216292 Chr.11: 65539756-65539756 on Build GRCh38
rs926245835 Chr.11: 65539644-65539644 on Build GRCh38

View Full Product Details