Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 42513170-42513679 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNAP23
Entrez Gene ID
8773
Gene Name
synaptosome associated protein 23
Gene Aliases
HsT17016, SNAP-23, SNAP23A, SNAP23B
UniGene
Hs.511149
Gene Chromosome Location
Chr.15: 42491244-42533061 on Build GRCh38
Transcripts
NM_003825.3 NM_130798.2 XM_006720725.2 XM_017022692.1 XM_017022693.1 XM_017022694.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4818282 c.103C>G p.Q35E Chr.15: 42513402-42513402 on Build GRCh38
5781137 c.122C>T p.T41I Chr.15: 42513421-42513421 on Build GRCh38
2187410 c.131T>C p.M44T Chr.15: 42513430-42513430 on Build GRCh38
6496098 c.103C>T p.Q35* Chr.15: 42513402-42513402 on Build GRCh38
700560 c.142C>A p.Q48K Chr.15: 42513441-42513441 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs931286056 Chr.15: 42513644-42513644 on Build GRCh38
rs749743300 Chr.15: 42513472-42513472 on Build GRCh38
rs543964969 Chr.15: 42513568-42513568 on Build GRCh38
rs369292370 Chr.15: 42513466-42513466 on Build GRCh38
rs889617864 Chr.15: 42513216-42513216 on Build GRCh38
rs766684707 Chr.15: 42513196-42513196 on Build GRCh38
rs750313327 Chr.15: 42513424-42513424 on Build GRCh38
rs965847478 Chr.15: 42513564-42513564 on Build GRCh38
rs1030589212 Chr.15: 42513420-42513420 on Build GRCh38
rs746298506 Chr.15: 42513558-42513558 on Build GRCh38
rs773574805 Chr.15: 42513317-42513317 on Build GRCh38
rs781353945 Chr.15: 42513323-42513323 on Build GRCh38
rs535028743 Chr.15: 42513344-42513344 on Build GRCh38
rs969947014 Chr.15: 42513552-42513552 on Build GRCh38
rs908459627 Chr.15: 42513655-42513655 on Build GRCh38
rs764799240 Chr.15: 42513291-42513291 on Build GRCh38
rs778841289 Chr.15: 42513483-42513483 on Build GRCh38
rs763726827 Chr.15: 42513241-42513241 on Build GRCh38
rs777872404 Chr.15: 42513208-42513208 on Build GRCh38
rs771092908 Chr.15: 42513345-42513345 on Build GRCh38
rs748748266 Chr.15: 42513336-42513336 on Build GRCh38
rs999166736 Chr.15: 42513408-42513408 on Build GRCh38
rs558225693 Chr.15: 42513384-42513384 on Build GRCh38
rs754573435 Chr.15: 42513439-42513439 on Build GRCh38
rs775897162 Chr.15: 42513656-42513656 on Build GRCh38
rs956742648 Chr.15: 42513542-42513542 on Build GRCh38
rs372575887 Chr.15: 42513485-42513485 on Build GRCh38
rs746603389 Chr.15: 42513305-42513305 on Build GRCh38
rs752123301 Chr.15: 42513351-42513351 on Build GRCh38
rs749907761 Chr.15: 42513294-42513294 on Build GRCh38
rs770393723 Chr.15: 42513341-42513341 on Build GRCh38
rs773473732 Chr.15: 42513342-42513342 on Build GRCh38
rs755376385 Chr.15: 42513308-42513308 on Build GRCh38
rs1019539574 Chr.15: 42513561-42513561 on Build GRCh38
rs772158614 Chr.15: 42513493-42513493 on Build GRCh38
rs761782201 Chr.15: 42513392-42513392 on Build GRCh38
rs751238982 Chr.15: 42513436-42513436 on Build GRCh38
rs748764454 Chr.15: 42513285-42513285 on Build GRCh38
rs150249939 Chr.15: 42513199-42513199 on Build GRCh38
rs921110502 Chr.15: 42513569-42513569 on Build GRCh38
rs757169156 Chr.15: 42513255-42513255 on Build GRCh38
rs766924800 Chr.15: 42513345-42513350 on Build GRCh38
rs1019770249 Chr.15: 42513373-42513373 on Build GRCh38
rs553251620 Chr.15: 42513215-42513215 on Build GRCh38
rs781650435 Chr.15: 42513443-42513443 on Build GRCh38
rs939650595 Chr.15: 42513229-42513229 on Build GRCh38
rs760547832 Chr.15: 42513240-42513240 on Build GRCh38
rs75170357 Chr.15: 42513430-42513430 on Build GRCh38
rs908238352 Chr.15: 42513220-42513220 on Build GRCh38
rs757937500 Chr.15: 42513427-42513427 on Build GRCh38
rs753823812 Chr.15: 42513250-42513250 on Build GRCh38
rs920586827 Chr.15: 42513619-42513619 on Build GRCh38
rs928051404 Chr.15: 42513595-42513595 on Build GRCh38
rs868414732 Chr.15: 42513624-42513624 on Build GRCh38
rs965070448 Chr.15: 42513405-42513405 on Build GRCh38
rs765139961 Chr.15: 42513396-42513396 on Build GRCh38
rs779705852 Chr.15: 42513304-42513304 on Build GRCh38
rs542924637 Chr.15: 42513608-42513608 on Build GRCh38
rs774385732 Chr.15: 42513234-42513234 on Build GRCh38
rs778453778 Chr.15: 42513469-42513469 on Build GRCh38
rs770475209 Chr.15: 42513365-42513365 on Build GRCh38
rs565886511 Chr.15: 42513261-42513261 on Build GRCh38
rs759616719 Chr.15: 42513382-42513382 on Build GRCh38
rs756495909 Chr.15: 42513468-42513468 on Build GRCh38
rs895549741 Chr.15: 42513338-42513338 on Build GRCh38
rs984063310 Chr.15: 42513638-42513638 on Build GRCh38
rs916776307 Chr.15: 42513499-42513499 on Build GRCh38
rs574192476 Chr.15: 42513600-42513600 on Build GRCh38
rs557573166 Chr.15: 42513586-42513586 on Build GRCh38
rs771145819 Chr.15: 42513476-42513476 on Build GRCh38
rs758099038 Chr.15: 42513301-42513301 on Build GRCh38
rs763254874 Chr.15: 42513345-42513347 on Build GRCh38

View Full Product Details