Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 42532045-42532552 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRC57
Entrez Gene ID
255252
Gene Name
leucine rich repeat containing 57
Gene Aliases
-
UniGene
Hs.234681
Gene Chromosome Location
Chr.15: 42528332-42548804 on Build GRCh38
Transcripts
XM_011521423.2 NM_003825.3 NM_130798.2 XM_006720725.2

Gene Symbol
SNAP23
Entrez Gene ID
8773
Gene Name
synaptosome associated protein 23
Gene Aliases
HsT17016, SNAP-23, SNAP23A, SNAP23B
UniGene
Hs.511149
Gene Chromosome Location
Chr.15: 42491244-42533061 on Build GRCh38
Transcripts
XM_011521423.2 NM_003825.3 NM_130798.2 XM_006720725.2

SNP Detail

SNP ID Chromosome Location
rs944717852 Chr.15: 42532310-42532310 on Build GRCh38
rs572699561 Chr.15: 42532135-42532135 on Build GRCh38
rs1027430616 Chr.15: 42532167-42532167 on Build GRCh38
rs767221469 Chr.15: 42532332-42532332 on Build GRCh38
rs564171566 Chr.15: 42532147-42532147 on Build GRCh38
rs1032033928 Chr.15: 42532202-42532202 on Build GRCh38
rs189831391 Chr.15: 42532247-42532247 on Build GRCh38
rs200587308 Chr.15: 42532384-42532385 on Build GRCh38
rs11022 Chr.15: 42532380-42532380 on Build GRCh38
rs901186860 Chr.15: 42532187-42532187 on Build GRCh38
rs901245569 Chr.15: 42532420-42532420 on Build GRCh38
rs116060757 Chr.15: 42532399-42532399 on Build GRCh38
rs935354356 Chr.15: 42532429-42532429 on Build GRCh38
rs766836227 Chr.15: 42532095-42532095 on Build GRCh38
rs367817702 Chr.15: 42532115-42532116 on Build GRCh38
rs117057596 Chr.15: 42532453-42532453 on Build GRCh38
rs879945358 Chr.15: 42532422-42532422 on Build GRCh38
rs75635089 Chr.15: 42532108-42532108 on Build GRCh38
rs547726640 Chr.15: 42532387-42532387 on Build GRCh38
rs1048593127 Chr.15: 42532123-42532123 on Build GRCh38
rs926109336 Chr.15: 42532272-42532272 on Build GRCh38
rs892014349 Chr.15: 42532292-42532292 on Build GRCh38
rs386783476 Chr.15: 42532384-42532386 on Build GRCh38
rs528376253 Chr.15: 42532236-42532236 on Build GRCh38
rs73404763 Chr.15: 42532278-42532278 on Build GRCh38
rs114902006 Chr.15: 42532386-42532386 on Build GRCh38
rs563109682 Chr.15: 42532331-42532331 on Build GRCh38
rs142265126 Chr.15: 42532318-42532318 on Build GRCh38
rs1060265 Chr.15: 42532384-42532384 on Build GRCh38
rs541013228 Chr.15: 42532190-42532190 on Build GRCh38
rs1040333459 Chr.15: 42532317-42532317 on Build GRCh38
rs960657715 Chr.15: 42532244-42532244 on Build GRCh38
rs918834547 Chr.15: 42532111-42532111 on Build GRCh38
rs964737123 Chr.15: 42532152-42532152 on Build GRCh38
rs762393881 Chr.15: 42532252-42532252 on Build GRCh38
rs947768884 Chr.15: 42532265-42532265 on Build GRCh38
rs983770194 Chr.15: 42532235-42532235 on Build GRCh38
rs778364151 Chr.15: 42532495-42532495 on Build GRCh38
rs999502886 Chr.15: 42532199-42532201 on Build GRCh38
rs1053782508 Chr.15: 42532291-42532291 on Build GRCh38
rs372744266 Chr.15: 42532486-42532486 on Build GRCh38
rs776675829 Chr.15: 42532251-42532251 on Build GRCh38
rs765866602 Chr.15: 42532271-42532271 on Build GRCh38
rs529038195 Chr.15: 42532373-42532373 on Build GRCh38
rs190885244 Chr.15: 42532417-42532417 on Build GRCh38
rs996208860 Chr.15: 42532346-42532346 on Build GRCh38
rs1041109178 Chr.15: 42532174-42532174 on Build GRCh38
rs952030559 Chr.15: 42532177-42532177 on Build GRCh38
rs62019290 Chr.15: 42532110-42532110 on Build GRCh38
rs562349763 Chr.15: 42532069-42532069 on Build GRCh38
rs1053161302 Chr.15: 42532075-42532075 on Build GRCh38
rs891789230 Chr.15: 42532122-42532122 on Build GRCh38
rs960631688 Chr.15: 42532477-42532477 on Build GRCh38
rs1008860315 Chr.15: 42532142-42532142 on Build GRCh38
rs995988923 Chr.15: 42532159-42532159 on Build GRCh38

View Full Product Details