Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 43360649-43361154 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZSCAN29
Entrez Gene ID
146050
Gene Name
zinc finger and SCAN domain containing 29
Gene Aliases
ZNF690, Zfp690
UniGene
Hs.418287
Gene Chromosome Location
Chr.15: 43358172-43371032 on Build GRCh38
Transcripts
NM_152455.3 XM_006720401.3 XM_011521266.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6998523 c.2532G>T p.K844N Chr.15: 43361100-43361100 on Build GRCh38
3754325 c.2508C>T p.H836H Chr.15: 43361124-43361124 on Build GRCh38
5916474 c.2497C>T p.L833F Chr.15: 43361135-43361135 on Build GRCh38
4453070 c.2487A>T p.K829N Chr.15: 43361145-43361145 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs971244123 Chr.15: 43360875-43360875 on Build GRCh38
rs202173229 Chr.15: 43361108-43361108 on Build GRCh38
rs10518812 Chr.15: 43360739-43360739 on Build GRCh38
rs58954604 Chr.15: 43360934-43360934 on Build GRCh38
rs748854245 Chr.15: 43361002-43361002 on Build GRCh38
rs137944091 Chr.15: 43361107-43361107 on Build GRCh38
rs183095976 Chr.15: 43360848-43360848 on Build GRCh38
rs79786746 Chr.15: 43360831-43360831 on Build GRCh38
rs555395977 Chr.15: 43361054-43361054 on Build GRCh38
rs1024079780 Chr.15: 43360871-43360871 on Build GRCh38
rs750676470 Chr.15: 43361029-43361029 on Build GRCh38
rs896314065 Chr.15: 43360858-43360858 on Build GRCh38
rs769218009 Chr.15: 43361081-43361081 on Build GRCh38
rs149461798 Chr.15: 43361123-43361123 on Build GRCh38
rs549809797 Chr.15: 43361012-43361012 on Build GRCh38
rs763208930 Chr.15: 43361112-43361112 on Build GRCh38
rs1012461840 Chr.15: 43360878-43360878 on Build GRCh38
rs772994507 Chr.15: 43360888-43360892 on Build GRCh38
rs973688615 Chr.15: 43360671-43360671 on Build GRCh38
rs756035163 Chr.15: 43361045-43361045 on Build GRCh38
rs907548202 Chr.15: 43361091-43361091 on Build GRCh38
rs1055360301 Chr.15: 43361077-43361077 on Build GRCh38
rs766644002 Chr.15: 43361113-43361113 on Build GRCh38
rs576043169 Chr.15: 43360915-43360915 on Build GRCh38
rs940480583 Chr.15: 43361100-43361100 on Build GRCh38
rs750583772 Chr.15: 43360693-43360693 on Build GRCh38
rs746583185 Chr.15: 43361075-43361075 on Build GRCh38
rs35278805 Chr.15: 43361124-43361124 on Build GRCh38
rs1041817744 Chr.15: 43360777-43360777 on Build GRCh38
rs191249783 Chr.15: 43360827-43360827 on Build GRCh38
rs1000882208 Chr.15: 43360972-43360972 on Build GRCh38
rs569656886 Chr.15: 43361034-43361034 on Build GRCh38
rs762034528 Chr.15: 43361098-43361098 on Build GRCh38
rs60924629 Chr.15: 43360933-43360933 on Build GRCh38
rs751637493 Chr.15: 43361114-43361114 on Build GRCh38
rs547503271 Chr.15: 43360953-43360953 on Build GRCh38
rs922138781 Chr.15: 43360685-43360687 on Build GRCh38
rs775119312 Chr.15: 43361074-43361074 on Build GRCh38
rs1048679278 Chr.15: 43360767-43360767 on Build GRCh38
rs200107245 Chr.15: 43361042-43361042 on Build GRCh38
rs1014056658 Chr.15: 43360870-43360870 on Build GRCh38
rs956814219 Chr.15: 43360980-43360980 on Build GRCh38
rs781072204 Chr.15: 43360773-43360773 on Build GRCh38
rs187742338 Chr.15: 43361086-43361086 on Build GRCh38
rs1000237121 Chr.15: 43360819-43360819 on Build GRCh38
rs1051903368 Chr.15: 43360738-43360738 on Build GRCh38
rs770315103 Chr.15: 43361041-43361041 on Build GRCh38
rs945795086 Chr.15: 43360771-43360771 on Build GRCh38
rs1020080604 Chr.15: 43361035-43361035 on Build GRCh38
rs369665095 Chr.15: 43361053-43361053 on Build GRCh38
rs777837567 Chr.15: 43360974-43360974 on Build GRCh38
rs374000879 Chr.15: 43361058-43361058 on Build GRCh38
rs987659786 Chr.15: 43361010-43361010 on Build GRCh38
rs904183250 Chr.15: 43360809-43360809 on Build GRCh38
rs371736070 Chr.15: 43361031-43361031 on Build GRCh38
rs752682924 Chr.15: 43360843-43360843 on Build GRCh38
rs543616507 Chr.15: 43360947-43360947 on Build GRCh38
rs778381285 Chr.15: 43361057-43361057 on Build GRCh38
rs377557566 Chr.15: 43361061-43361061 on Build GRCh38
rs370490171 Chr.15: 43361092-43361092 on Build GRCh38
rs755058248 Chr.15: 43361120-43361120 on Build GRCh38
rs777758246 Chr.15: 43361047-43361047 on Build GRCh38
rs903746526 Chr.15: 43360940-43360940 on Build GRCh38
rs920131605 Chr.15: 43360691-43360691 on Build GRCh38

View Full Product Details