Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 26447476-26447979 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHDDS
Entrez Gene ID
79947
Gene Name
dehydrodolichyl diphosphate synthase subunit
Gene Aliases
CIT, CPT, DS, HDS, RP59
UniGene
Hs.369385
Gene Chromosome Location
Chr.1: 26432282-26471306 on Build GRCh38
Transcripts
NM_001243564.1 NM_001243565.1 NM_001319959.1 NM_024887.3 NM_205861.2 XM_006710912.2 XM_006710913.2 XM_006710914.2 XM_006710915.2 XM_006710916.2 XM_006710917.2 XM_006710918.2 XM_011542183.2 XM_011542184.2 XM_011542185.2 XM_011542186.2 XM_017002379.1 XM_017002380.1 XM_017002381.1 XM_017002382.1 XM_017002383.1 XM_017002384.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6584024 c.486C>G p.I162M Chr.1: 26447604-26447604 on Build GRCh38
907671 c.466T>C p.Y156H Chr.1: 26447584-26447584 on Build GRCh38
6373827 c.542G>C p.S181T Chr.1: 26447660-26447660 on Build GRCh38
3804803 c.492T>A p.N164K Chr.1: 26447610-26447610 on Build GRCh38
907672 c.494C>T p.A165V Chr.1: 26447612-26447612 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs886688061 Chr.1: 26447917-26447917 on Build GRCh38
rs772924790 Chr.1: 26447696-26447696 on Build GRCh38
rs201898489 Chr.1: 26447542-26447542 on Build GRCh38
rs763275058 Chr.1: 26447549-26447549 on Build GRCh38
rs769323514 Chr.1: 26447660-26447660 on Build GRCh38
rs932486700 Chr.1: 26447802-26447802 on Build GRCh38
rs900788331 Chr.1: 26447846-26447846 on Build GRCh38
rs936734474 Chr.1: 26447881-26447881 on Build GRCh38
rs367557625 Chr.1: 26447630-26447630 on Build GRCh38
rs764831063 Chr.1: 26447535-26447535 on Build GRCh38
rs776132110 Chr.1: 26447528-26447528 on Build GRCh38
rs752178339 Chr.1: 26447565-26447565 on Build GRCh38
rs779041144 Chr.1: 26447716-26447716 on Build GRCh38
rs770708595 Chr.1: 26447759-26447759 on Build GRCh38
rs756509826 Chr.1: 26447595-26447595 on Build GRCh38
rs370849250 Chr.1: 26447521-26447521 on Build GRCh38
rs1025197385 Chr.1: 26447719-26447719 on Build GRCh38
rs1015222321 Chr.1: 26447944-26447944 on Build GRCh38
rs774102743 Chr.1: 26447768-26447768 on Build GRCh38
rs750044465 Chr.1: 26447539-26447539 on Build GRCh38
rs200852161 Chr.1: 26447688-26447688 on Build GRCh38
rs753960562 Chr.1: 26447597-26447597 on Build GRCh38
rs1056907267 Chr.1: 26447706-26447706 on Build GRCh38
rs896478931 Chr.1: 26447737-26447737 on Build GRCh38
rs769252429 Chr.1: 26447746-26447746 on Build GRCh38
rs375903302 Chr.1: 26447638-26447638 on Build GRCh38
rs982452900 Chr.1: 26447753-26447753 on Build GRCh38
rs372941732 Chr.1: 26447637-26447637 on Build GRCh38
rs950989592 Chr.1: 26447720-26447720 on Build GRCh38
rs569375282 Chr.1: 26447952-26447952 on Build GRCh38
rs376267529 Chr.1: 26447531-26447531 on Build GRCh38
rs895293358 Chr.1: 26447914-26447914 on Build GRCh38
rs746837932 Chr.1: 26447727-26447727 on Build GRCh38
rs781042066 Chr.1: 26447731-26447731 on Build GRCh38
rs1013687215 Chr.1: 26447915-26447915 on Build GRCh38
rs768075911 Chr.1: 26447586-26447586 on Build GRCh38
rs754070431 Chr.1: 26447704-26447704 on Build GRCh38
rs552360072 Chr.1: 26447841-26447841 on Build GRCh38
rs149948788 Chr.1: 26447668-26447668 on Build GRCh38
rs201038798 Chr.1: 26447559-26447559 on Build GRCh38
rs1005588537 Chr.1: 26447929-26447929 on Build GRCh38
rs1006409155 Chr.1: 26447832-26447833 on Build GRCh38
rs746749832 Chr.1: 26447627-26447627 on Build GRCh38
rs748749709 Chr.1: 26447755-26447755 on Build GRCh38
rs757487649 Chr.1: 26447713-26447713 on Build GRCh38
rs550186277 Chr.1: 26447515-26447515 on Build GRCh38
rs774865740 Chr.1: 26447676-26447676 on Build GRCh38
rs772765751 Chr.1: 26447750-26447750 on Build GRCh38
rs754699307 Chr.1: 26447730-26447730 on Build GRCh38
rs922956992 Chr.1: 26447557-26447557 on Build GRCh38
rs202148886 Chr.1: 26447583-26447583 on Build GRCh38
rs529638382 Chr.1: 26447682-26447682 on Build GRCh38
rs1042805853 Chr.1: 26447579-26447579 on Build GRCh38
rs758260694 Chr.1: 26447598-26447598 on Build GRCh38
rs756192012 Chr.1: 26447690-26447690 on Build GRCh38
rs912465308 Chr.1: 26447835-26447835 on Build GRCh38
rs779814399 Chr.1: 26447611-26447611 on Build GRCh38
rs186191592 Chr.1: 26447779-26447779 on Build GRCh38
rs748086202 Chr.1: 26447744-26447744 on Build GRCh38
rs538864907 Chr.1: 26447780-26447780 on Build GRCh38
rs1033812946 Chr.1: 26447665-26447665 on Build GRCh38
rs776512735 Chr.1: 26447634-26447634 on Build GRCh38
rs1055147828 Chr.1: 26447884-26447884 on Build GRCh38
rs369429285 Chr.1: 26447594-26447594 on Build GRCh38
rs775930566 Chr.1: 26447786-26447786 on Build GRCh38
rs1025125296 Chr.1: 26447916-26447916 on Build GRCh38
rs1042388583 Chr.1: 26447854-26447854 on Build GRCh38
rs1050979651 Chr.1: 26447822-26447822 on Build GRCh38
rs764232401 Chr.1: 26447701-26447701 on Build GRCh38
rs772779004 Chr.1: 26447667-26447667 on Build GRCh38
rs866202896 Chr.1: 26447507-26447507 on Build GRCh38
rs765283728 Chr.1: 26447858-26447858 on Build GRCh38
rs752561243 Chr.1: 26447922-26447922 on Build GRCh38
rs752722280 Chr.1: 26447593-26447593 on Build GRCh38

View Full Product Details