Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 27012327-27012823 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM46B
Entrez Gene ID
115572
Gene Name
family with sequence similarity 46 member B
Gene Aliases
-
UniGene
Hs.632378
Gene Chromosome Location
Chr.1: 27005020-27012842 on Build GRCh38
Transcripts
NM_052943.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3487634 c.134C>T p.P45L Chr.1: 27012537-27012537 on Build GRCh38
6126222 c.194G>A p.S65N Chr.1: 27012477-27012477 on Build GRCh38
5468726 c.8C>A p.P3Q Chr.1: 27012663-27012663 on Build GRCh38
1341464 c.218G>A p.R73H Chr.1: 27012453-27012453 on Build GRCh38
6497016 c.206C>A p.P69H Chr.1: 27012465-27012465 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377504560 Chr.1: 27012494-27012494 on Build GRCh38
rs766887556 Chr.1: 27012478-27012478 on Build GRCh38
rs779935361 Chr.1: 27012416-27012416 on Build GRCh38
rs199535191 Chr.1: 27012386-27012386 on Build GRCh38
rs971671168 Chr.1: 27012654-27012654 on Build GRCh38
rs767965947 Chr.1: 27012495-27012495 on Build GRCh38
rs1030612130 Chr.1: 27012634-27012634 on Build GRCh38
rs1033444743 Chr.1: 27012617-27012617 on Build GRCh38
rs756389783 Chr.1: 27012518-27012518 on Build GRCh38
rs754177179 Chr.1: 27012595-27012595 on Build GRCh38
rs776112407 Chr.1: 27012432-27012432 on Build GRCh38
rs989302843 Chr.1: 27012369-27012369 on Build GRCh38
rs746292601 Chr.1: 27012685-27012685 on Build GRCh38
rs578233653 Chr.1: 27012601-27012601 on Build GRCh38
rs559699502 Chr.1: 27012739-27012739 on Build GRCh38
rs1021062902 Chr.1: 27012607-27012607 on Build GRCh38
rs138542880 Chr.1: 27012479-27012479 on Build GRCh38
rs35810604 Chr.1: 27012614-27012614 on Build GRCh38
rs747718885 Chr.1: 27012557-27012557 on Build GRCh38
rs780786522 Chr.1: 27012603-27012603 on Build GRCh38
rs751401047 Chr.1: 27012599-27012599 on Build GRCh38
rs910715789 Chr.1: 27012768-27012768 on Build GRCh38
rs894931395 Chr.1: 27012777-27012777 on Build GRCh38
rs775927833 Chr.1: 27012579-27012579 on Build GRCh38
rs981008389 Chr.1: 27012693-27012693 on Build GRCh38
rs985027463 Chr.1: 27012764-27012764 on Build GRCh38
rs781059179 Chr.1: 27012552-27012552 on Build GRCh38
rs551498371 Chr.1: 27012773-27012773 on Build GRCh38
rs777399569 Chr.1: 27012618-27012618 on Build GRCh38
rs528573100 Chr.1: 27012765-27012765 on Build GRCh38
rs776800126 Chr.1: 27012397-27012397 on Build GRCh38
rs768560725 Chr.1: 27012431-27012431 on Build GRCh38
rs116726456 Chr.1: 27012596-27012596 on Build GRCh38
rs148050298 Chr.1: 27012469-27012469 on Build GRCh38
rs746764507 Chr.1: 27012424-27012424 on Build GRCh38
rs1050965559 Chr.1: 27012591-27012591 on Build GRCh38
rs780132172 Chr.1: 27012546-27012546 on Build GRCh38
rs774689393 Chr.1: 27012571-27012571 on Build GRCh38
rs542614118 Chr.1: 27012735-27012735 on Build GRCh38
rs1010749455 Chr.1: 27012663-27012663 on Build GRCh38
rs1013079491 Chr.1: 27012779-27012779 on Build GRCh38
rs754703912 Chr.1: 27012548-27012548 on Build GRCh38
rs745760498 Chr.1: 27012412-27012412 on Build GRCh38
rs760920374 Chr.1: 27012582-27012582 on Build GRCh38
rs778020592 Chr.1: 27012407-27012407 on Build GRCh38
rs556070580 Chr.1: 27012508-27012508 on Build GRCh38
rs764397161 Chr.1: 27012587-27012587 on Build GRCh38
rs969828939 Chr.1: 27012743-27012769 on Build GRCh38
rs1024555894 Chr.1: 27012644-27012644 on Build GRCh38
rs772645200 Chr.1: 27012703-27012703 on Build GRCh38
rs539335606 Chr.1: 27012437-27012437 on Build GRCh38
rs767733030 Chr.1: 27012578-27012578 on Build GRCh38
rs763385717 Chr.1: 27012476-27012476 on Build GRCh38
rs369265939 Chr.1: 27012360-27012360 on Build GRCh38
rs201781588 Chr.1: 27012554-27012554 on Build GRCh38
rs769445450 Chr.1: 27012558-27012558 on Build GRCh38
rs377530137 Chr.1: 27012378-27012378 on Build GRCh38
rs750904634 Chr.1: 27012357-27012357 on Build GRCh38
rs570359008 Chr.1: 27012413-27012413 on Build GRCh38
rs76246176 Chr.1: 27012780-27012780 on Build GRCh38
rs558376071 Chr.1: 27012600-27012600 on Build GRCh38
rs958823113 Chr.1: 27012619-27012619 on Build GRCh38
rs777933549 Chr.1: 27012520-27012520 on Build GRCh38
rs769249037 Chr.1: 27012451-27012451 on Build GRCh38
rs529605196 Chr.1: 27012712-27012712 on Build GRCh38
rs373891927 Chr.1: 27012363-27012363 on Build GRCh38
rs903484611 Chr.1: 27012743-27012743 on Build GRCh38
rs761115270 Chr.1: 27012709-27012709 on Build GRCh38
rs921386591 Chr.1: 27012694-27012694 on Build GRCh38
rs1050913693 Chr.1: 27012726-27012726 on Build GRCh38
rs772654039 Chr.1: 27012563-27012563 on Build GRCh38
rs1004507533 Chr.1: 27012746-27012746 on Build GRCh38
rs541168500 Chr.1: 27012598-27012598 on Build GRCh38
rs758965437 Chr.1: 27012691-27012691 on Build GRCh38
rs4970471 Chr.1: 27012612-27012612 on Build GRCh38
rs974523184 Chr.1: 27012688-27012688 on Build GRCh38
rs775918408 Chr.1: 27012704-27012704 on Build GRCh38
rs982787607 Chr.1: 27012659-27012659 on Build GRCh38
rs773936443 Chr.1: 27012643-27012643 on Build GRCh38
rs760139038 Chr.1: 27012577-27012577 on Build GRCh38
rs569639678 Chr.1: 27012545-27012545 on Build GRCh38
rs1022555561 Chr.1: 27012684-27012684 on Build GRCh38
rs748979913 Chr.1: 27012620-27012620 on Build GRCh38
rs770524427 Chr.1: 27012551-27012564 on Build GRCh38
rs753978511 Chr.1: 27012525-27012525 on Build GRCh38
rs770406893 Chr.1: 27012642-27012642 on Build GRCh38
rs536948702 Chr.1: 27012585-27012596 on Build GRCh38
rs772744023 Chr.1: 27012463-27012463 on Build GRCh38
rs1035143836 Chr.1: 27012707-27012707 on Build GRCh38
rs566005435 Chr.1: 27012356-27012356 on Build GRCh38
rs570268118 Chr.1: 27012358-27012358 on Build GRCh38
rs972080231 Chr.1: 27012721-27012721 on Build GRCh38
rs370980393 Chr.1: 27012565-27012565 on Build GRCh38

View Full Product Details