Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 39764871-39765420 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IKZF3
Entrez Gene ID
22806
Gene Name
IKAROS family zinc finger 3
Gene Aliases
AIO, AIOLOS, ZNFN1A3
UniGene
Hs.371680
Gene Chromosome Location
Chr.17: 39757715-39864188 on Build GRCh38
Transcripts
NM_001257408.1 NM_001257409.1 NM_001257410.1 NM_001257411.1 NM_001257412.1 NM_001257413.1 NM_001257414.1 NM_001284514.1 NM_001284515.1 NM_001284516.1 NM_012481.4 NM_183228.2 NM_183229.2 NM_183230.2 NM_183231.2 NM_183232.2

SNP Detail

SNP ID Chromosome Location
rs778364591 Chr.17: 39765262-39765262 on Build GRCh38
rs927893836 Chr.17: 39765040-39765040 on Build GRCh38
rs1027782333 Chr.17: 39765226-39765226 on Build GRCh38
rs887910238 Chr.17: 39765252-39765252 on Build GRCh38
rs957574112 Chr.17: 39765313-39765313 on Build GRCh38
rs538729941 Chr.17: 39765102-39765102 on Build GRCh38
rs2941509 Chr.17: 39764941-39764941 on Build GRCh38
rs550789387 Chr.17: 39765389-39765389 on Build GRCh38
rs564023066 Chr.17: 39765127-39765127 on Build GRCh38
rs775968961 Chr.17: 39765361-39765361 on Build GRCh38
rs983902788 Chr.17: 39764967-39764967 on Build GRCh38
rs996331629 Chr.17: 39764903-39764903 on Build GRCh38
rs569381918 Chr.17: 39765396-39765396 on Build GRCh38
rs879228572 Chr.17: 39765172-39765172 on Build GRCh38
rs913512924 Chr.17: 39765370-39765370 on Build GRCh38
rs541567149 Chr.17: 39764998-39764998 on Build GRCh38
rs1033148774 Chr.17: 39765285-39765285 on Build GRCh38
rs528385729 Chr.17: 39765204-39765204 on Build GRCh38
rs959278024 Chr.17: 39765381-39765381 on Build GRCh38
rs942401273 Chr.17: 39765116-39765116 on Build GRCh38
rs957631516 Chr.17: 39765020-39765020 on Build GRCh38
rs770456297 Chr.17: 39765319-39765319 on Build GRCh38
rs561738012 Chr.17: 39765299-39765299 on Build GRCh38
rs1014934295 Chr.17: 39764977-39764977 on Build GRCh38
rs990690194 Chr.17: 39765087-39765087 on Build GRCh38
rs540395595 Chr.17: 39765212-39765212 on Build GRCh38
rs971492043 Chr.17: 39765322-39765322 on Build GRCh38
rs866742497 Chr.17: 39764944-39764944 on Build GRCh38
rs986500549 Chr.17: 39765394-39765394 on Build GRCh38
rs373643986 Chr.17: 39765005-39765005 on Build GRCh38
rs1034348049 Chr.17: 39765371-39765371 on Build GRCh38
rs529349825 Chr.17: 39765365-39765365 on Build GRCh38
rs937903384 Chr.17: 39765086-39765086 on Build GRCh38
rs140541594 Chr.17: 39765286-39765287 on Build GRCh38
rs944395498 Chr.17: 39765171-39765171 on Build GRCh38
rs1004982626 Chr.17: 39765266-39765266 on Build GRCh38
rs902192856 Chr.17: 39765152-39765152 on Build GRCh38
rs989376801 Chr.17: 39765343-39765343 on Build GRCh38
rs900799983 Chr.17: 39765203-39765203 on Build GRCh38
rs756952707 Chr.17: 39765198-39765198 on Build GRCh38
rs753382907 Chr.17: 39765182-39765182 on Build GRCh38
rs1014287958 Chr.17: 39765224-39765224 on Build GRCh38
rs981447080 Chr.17: 39765021-39765021 on Build GRCh38
rs946492090 Chr.17: 39765110-39765110 on Build GRCh38
rs981882623 Chr.17: 39765352-39765352 on Build GRCh38
rs913471733 Chr.17: 39765062-39765062 on Build GRCh38
rs574888974 Chr.17: 39765058-39765058 on Build GRCh38
rs185478724 Chr.17: 39765111-39765111 on Build GRCh38
rs1001546055 Chr.17: 39765274-39765274 on Build GRCh38
rs980263614 Chr.17: 39764943-39764943 on Build GRCh38

View Full Product Details