Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 40076793-40077301 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
THRA
Entrez Gene ID
7067
Gene Name
thyroid hormone receptor, alpha
Gene Aliases
AR7, CHNG6, EAR7, ERB-T-1, ERBA, ERBA1, NR1A1, THRA1, THRA2, c-ERBA-1
UniGene
Hs.724
Gene Chromosome Location
Chr.17: 40062193-40093867 on Build GRCh38
Transcripts
NM_001190918.1 NM_001190919.1 NM_003250.5 NM_199334.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7193820 c.102C>G p.S34S Chr.17: 40076919-40076919 on Build GRCh38
5893965 c.54-8C>T p.? Chr.17: 40076863-40076863 on Build GRCh38
1710204 c.101C>T p.S34F Chr.17: 40076918-40076918 on Build GRCh38
4733616 c.91G>A p.G31S Chr.17: 40076908-40076908 on Build GRCh38
1710202 c.61T>A p.S21T Chr.17: 40076878-40076878 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs944196472 Chr.17: 40077043-40077043 on Build GRCh38
rs762706791 Chr.17: 40076829-40076829 on Build GRCh38
rs906476659 Chr.17: 40077226-40077226 on Build GRCh38
rs750378693 Chr.17: 40076859-40076859 on Build GRCh38
rs552868450 Chr.17: 40076994-40076994 on Build GRCh38
rs112664727 Chr.17: 40077242-40077242 on Build GRCh38
rs957436989 Chr.17: 40077140-40077140 on Build GRCh38
rs942336070 Chr.17: 40076957-40076957 on Build GRCh38
rs1020383554 Chr.17: 40077167-40077167 on Build GRCh38
rs1045356440 Chr.17: 40077065-40077065 on Build GRCh38
rs530568508 Chr.17: 40076900-40076900 on Build GRCh38
rs780966269 Chr.17: 40076855-40076855 on Build GRCh38
rs369146770 Chr.17: 40076955-40076955 on Build GRCh38
rs747306415 Chr.17: 40076910-40076910 on Build GRCh38
rs535568640 Chr.17: 40077061-40077061 on Build GRCh38
rs946061110 Chr.17: 40077013-40077013 on Build GRCh38
rs905128786 Chr.17: 40077080-40077080 on Build GRCh38
rs1046763077 Chr.17: 40077265-40077265 on Build GRCh38
rs184687239 Chr.17: 40077177-40077177 on Build GRCh38
rs751271851 Chr.17: 40076986-40076986 on Build GRCh38
rs570720280 Chr.17: 40076951-40076951 on Build GRCh38
rs545748199 Chr.17: 40077173-40077173 on Build GRCh38
rs113215611 Chr.17: 40076874-40076874 on Build GRCh38
rs763955512 Chr.17: 40076969-40076969 on Build GRCh38
rs911444975 Chr.17: 40077210-40077210 on Build GRCh38
rs568279073 Chr.17: 40077028-40077028 on Build GRCh38
rs929645020 Chr.17: 40077262-40077262 on Build GRCh38
rs113694684 Chr.17: 40076862-40076862 on Build GRCh38
rs763863476 Chr.17: 40076830-40076830 on Build GRCh38
rs189473726 Chr.17: 40077241-40077241 on Build GRCh38
rs927354130 Chr.17: 40076992-40076992 on Build GRCh38
rs958803597 Chr.17: 40076997-40076997 on Build GRCh38
rs951047260 Chr.17: 40076899-40076899 on Build GRCh38
rs113098356 Chr.17: 40076815-40076815 on Build GRCh38
rs556432286 Chr.17: 40077081-40077081 on Build GRCh38
rs552205515 Chr.17: 40076907-40076907 on Build GRCh38
rs1025303649 Chr.17: 40077214-40077214 on Build GRCh38
rs113018832 Chr.17: 40076977-40076977 on Build GRCh38
rs199530759 Chr.17: 40076870-40076870 on Build GRCh38
rs373692031 Chr.17: 40076970-40076970 on Build GRCh38
rs377188886 Chr.17: 40076954-40076954 on Build GRCh38
rs990211063 Chr.17: 40076998-40076998 on Build GRCh38
rs778973434 Chr.17: 40076883-40076883 on Build GRCh38
rs896282336 Chr.17: 40077006-40077006 on Build GRCh38
rs912855062 Chr.17: 40077042-40077042 on Build GRCh38
rs990160607 Chr.17: 40077272-40077272 on Build GRCh38
rs974137980 Chr.17: 40077254-40077254 on Build GRCh38
rs151082578 Chr.17: 40076911-40076911 on Build GRCh38
rs955492152 Chr.17: 40077196-40077196 on Build GRCh38
rs1010364653 Chr.17: 40077159-40077159 on Build GRCh38
rs369077224 Chr.17: 40076825-40076825 on Build GRCh38
rs777230459 Chr.17: 40076947-40076947 on Build GRCh38
rs372494553 Chr.17: 40076844-40076844 on Build GRCh38
rs557078992 Chr.17: 40077129-40077129 on Build GRCh38
rs899565794 Chr.17: 40077103-40077103 on Build GRCh38
rs562128753 Chr.17: 40077255-40077255 on Build GRCh38
rs1126503 Chr.17: 40076927-40076927 on Build GRCh38
rs766208212 Chr.17: 40077084-40077084 on Build GRCh38
rs774981222 Chr.17: 40077198-40077198 on Build GRCh38
rs751304984 Chr.17: 40076842-40076842 on Build GRCh38
rs1038062551 Chr.17: 40076963-40076963 on Build GRCh38
rs931027678 Chr.17: 40077109-40077109 on Build GRCh38
rs75534088 Chr.17: 40077138-40077138 on Build GRCh38
rs1001924544 Chr.17: 40077136-40077136 on Build GRCh38
rs1002426565 Chr.17: 40077234-40077234 on Build GRCh38
rs761462156 Chr.17: 40076990-40076990 on Build GRCh38
rs552800414 Chr.17: 40076964-40076964 on Build GRCh38

View Full Product Details