Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 40798793-40799306 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRT28
Entrez Gene ID
162605
Gene Name
keratin 28
Gene Aliases
K25IRS4, KRT25D
UniGene
Hs.59736
Gene Chromosome Location
Chr.17: 40792196-40799959 on Build GRCh38
Transcripts
NM_181535.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4066237 c.518A>G p.D173G Chr.17: 40798932-40798932 on Build GRCh38
6080874 c.525C>A p.F175L Chr.17: 40798925-40798925 on Build GRCh38
4919457 c.488T>C p.L163P Chr.17: 40798962-40798962 on Build GRCh38
1302777 c.520G>T p.D174Y Chr.17: 40798930-40798930 on Build GRCh38
3755475 c.529C>T p.L177L Chr.17: 40798921-40798921 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746097691 Chr.17: 40798982-40798982 on Build GRCh38
rs746278911 Chr.17: 40798879-40798879 on Build GRCh38
rs769086519 Chr.17: 40799018-40799018 on Build GRCh38
rs370955389 Chr.17: 40799258-40799258 on Build GRCh38
rs566305920 Chr.17: 40798942-40798942 on Build GRCh38
rs960967146 Chr.17: 40799078-40799078 on Build GRCh38
rs200138772 Chr.17: 40799002-40799002 on Build GRCh38
rs777867606 Chr.17: 40798958-40798958 on Build GRCh38
rs202198261 Chr.17: 40799034-40799034 on Build GRCh38
rs183147626 Chr.17: 40798881-40798881 on Build GRCh38
rs771087577 Chr.17: 40798871-40798871 on Build GRCh38
rs1029920772 Chr.17: 40798869-40798869 on Build GRCh38
rs954336875 Chr.17: 40799138-40799138 on Build GRCh38
rs916461885 Chr.17: 40799228-40799228 on Build GRCh38
rs925396480 Chr.17: 40799153-40799153 on Build GRCh38
rs149445923 Chr.17: 40798986-40798986 on Build GRCh38
rs1050805338 Chr.17: 40799224-40799224 on Build GRCh38
rs143876681 Chr.17: 40799050-40799050 on Build GRCh38
rs918412233 Chr.17: 40798937-40798937 on Build GRCh38
rs531304717 Chr.17: 40798822-40798822 on Build GRCh38
rs561132276 Chr.17: 40799231-40799231 on Build GRCh38
rs991967784 Chr.17: 40799218-40799218 on Build GRCh38
rs372413870 Chr.17: 40799156-40799156 on Build GRCh38
rs531648383 Chr.17: 40799273-40799273 on Build GRCh38
rs963067149 Chr.17: 40799162-40799162 on Build GRCh38
rs7208254 Chr.17: 40799199-40799199 on Build GRCh38
rs773891647 Chr.17: 40799020-40799020 on Build GRCh38
rs972789763 Chr.17: 40799177-40799177 on Build GRCh38
rs200963334 Chr.17: 40798905-40798905 on Build GRCh38
rs767206491 Chr.17: 40798923-40798923 on Build GRCh38
rs941880896 Chr.17: 40799277-40799277 on Build GRCh38
rs771708702 Chr.17: 40799025-40799025 on Build GRCh38
rs72824010 Chr.17: 40799085-40799085 on Build GRCh38
rs867725064 Chr.17: 40799262-40799262 on Build GRCh38
rs979539627 Chr.17: 40799108-40799108 on Build GRCh38
rs547672470 Chr.17: 40798936-40798936 on Build GRCh38
rs933258213 Chr.17: 40799200-40799200 on Build GRCh38
rs1007575741 Chr.17: 40799145-40799145 on Build GRCh38
rs35583648 Chr.17: 40799184-40799184 on Build GRCh38
rs370593289 Chr.17: 40798946-40798946 on Build GRCh38
rs754881207 Chr.17: 40798944-40798945 on Build GRCh38
rs1044066442 Chr.17: 40799256-40799256 on Build GRCh38
rs768166301 Chr.17: 40798916-40798916 on Build GRCh38
rs754892167 Chr.17: 40798947-40798947 on Build GRCh38
rs7224150 Chr.17: 40799022-40799022 on Build GRCh38
rs776076378 Chr.17: 40798895-40798895 on Build GRCh38
rs1043418136 Chr.17: 40799001-40799001 on Build GRCh38
rs7222207 Chr.17: 40798921-40798921 on Build GRCh38
rs757474993 Chr.17: 40798968-40798968 on Build GRCh38
rs773766615 Chr.17: 40798919-40798919 on Build GRCh38
rs751628287 Chr.17: 40798959-40798959 on Build GRCh38
rs760530871 Chr.17: 40798932-40798932 on Build GRCh38
rs1052286861 Chr.17: 40798874-40798874 on Build GRCh38
rs777101669 Chr.17: 40798877-40798877 on Build GRCh38
rs749765011 Chr.17: 40799013-40799013 on Build GRCh38
rs549751709 Chr.17: 40799280-40799280 on Build GRCh38
rs531872413 Chr.17: 40799075-40799075 on Build GRCh38
rs753603928 Chr.17: 40799051-40799051 on Build GRCh38
rs988116411 Chr.17: 40799193-40799193 on Build GRCh38
rs1016921854 Chr.17: 40799153-40799153 on Build GRCh38
rs781619299 Chr.17: 40798969-40798969 on Build GRCh38
rs138546546 Chr.17: 40799092-40799092 on Build GRCh38
rs552617156 Chr.17: 40798930-40798930 on Build GRCh38
rs760332660 Chr.17: 40799040-40799040 on Build GRCh38

View Full Product Details