Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 41394758-41395256 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRT31
Entrez Gene ID
3881
Gene Name
keratin 31
Gene Aliases
HA1, Ha-1, KRTHA1, hHa1
UniGene
Hs.41696
Gene Chromosome Location
Chr.17: 41393724-41397592 on Build GRCh38
Transcripts
NM_002277.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5950040 c.960C>T p.N320N Chr.17: 41394985-41394985 on Build GRCh38
472771 c.901A>G p.T301A Chr.17: 41395044-41395044 on Build GRCh38
1383147 c.976G>A p.A326T Chr.17: 41394969-41394969 on Build GRCh38
2840540 c.1068C>T p.Y356Y Chr.17: 41394877-41394877 on Build GRCh38
7148463 c.1083G>C p.E361D Chr.17: 41394862-41394862 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs181766476 Chr.17: 41394944-41394944 on Build GRCh38
rs765550426 Chr.17: 41394858-41394858 on Build GRCh38
rs778589698 Chr.17: 41394946-41394946 on Build GRCh38
rs375562565 Chr.17: 41394976-41394976 on Build GRCh38
rs759685551 Chr.17: 41394904-41394904 on Build GRCh38
rs896386695 Chr.17: 41395103-41395103 on Build GRCh38
rs185078782 Chr.17: 41394815-41394815 on Build GRCh38
rs776080592 Chr.17: 41394801-41394801 on Build GRCh38
rs953757489 Chr.17: 41395199-41395199 on Build GRCh38
rs772204868 Chr.17: 41394954-41394954 on Build GRCh38
rs528481251 Chr.17: 41394910-41394910 on Build GRCh38
rs758083554 Chr.17: 41395081-41395081 on Build GRCh38
rs146671831 Chr.17: 41394973-41394973 on Build GRCh38
rs576563180 Chr.17: 41395048-41395048 on Build GRCh38
rs779801015 Chr.17: 41394936-41394936 on Build GRCh38
rs764396471 Chr.17: 41394913-41394913 on Build GRCh38
rs749365613 Chr.17: 41395099-41395099 on Build GRCh38
rs201871410 Chr.17: 41394943-41394943 on Build GRCh38
rs374685776 Chr.17: 41394967-41394967 on Build GRCh38
rs112069991 Chr.17: 41394830-41394830 on Build GRCh38
rs143943601 Chr.17: 41394876-41394876 on Build GRCh38
rs900350931 Chr.17: 41395033-41395033 on Build GRCh38
rs752037872 Chr.17: 41394871-41394871 on Build GRCh38
rs117612447 Chr.17: 41394847-41394847 on Build GRCh38
rs201100680 Chr.17: 41394997-41394997 on Build GRCh38
rs146247884 Chr.17: 41394924-41394924 on Build GRCh38
rs552595139 Chr.17: 41395104-41395104 on Build GRCh38
rs762708732 Chr.17: 41395222-41395222 on Build GRCh38
rs753358278 Chr.17: 41395071-41395071 on Build GRCh38
rs752421349 Chr.17: 41395078-41395078 on Build GRCh38
rs951729501 Chr.17: 41394872-41394872 on Build GRCh38
rs768291234 Chr.17: 41394945-41394945 on Build GRCh38
rs112895156 Chr.17: 41394884-41394884 on Build GRCh38
rs866668832 Chr.17: 41394891-41394891 on Build GRCh38
rs773503630 Chr.17: 41395063-41395063 on Build GRCh38
rs758652176 Chr.17: 41395081-41395082 on Build GRCh38
rs778625274 Chr.17: 41395020-41395020 on Build GRCh38
rs772228051 Chr.17: 41395116-41395116 on Build GRCh38
rs201426405 Chr.17: 41395074-41395074 on Build GRCh38
rs141715467 Chr.17: 41394903-41394903 on Build GRCh38
rs781183957 Chr.17: 41395095-41395095 on Build GRCh38
rs761099800 Chr.17: 41395212-41395212 on Build GRCh38
rs372548012 Chr.17: 41395038-41395038 on Build GRCh38
rs991417002 Chr.17: 41395167-41395167 on Build GRCh38
rs761666383 Chr.17: 41394902-41394902 on Build GRCh38
rs773628449 Chr.17: 41395197-41395197 on Build GRCh38
rs752188469 Chr.17: 41395221-41395221 on Build GRCh38
rs777714912 Chr.17: 41395085-41395085 on Build GRCh38
rs749160362 Chr.17: 41394889-41394889 on Build GRCh38
rs773148729 Chr.17: 41395040-41395040 on Build GRCh38
rs922224076 Chr.17: 41394829-41394829 on Build GRCh38
rs978136699 Chr.17: 41395189-41395189 on Build GRCh38
rs747997953 Chr.17: 41395027-41395027 on Build GRCh38
rs757773958 Chr.17: 41394923-41394923 on Build GRCh38
rs763824847 Chr.17: 41395224-41395224 on Build GRCh38
rs759839376 Chr.17: 41394848-41394848 on Build GRCh38
rs758246862 Chr.17: 41395028-41395028 on Build GRCh38
rs767887288 Chr.17: 41395049-41395049 on Build GRCh38
rs1035659841 Chr.17: 41395139-41395139 on Build GRCh38
rs746742544 Chr.17: 41395088-41395088 on Build GRCh38
rs558246350 Chr.17: 41395039-41395039 on Build GRCh38
rs754648711 Chr.17: 41395006-41395006 on Build GRCh38
rs753334304 Chr.17: 41395005-41395005 on Build GRCh38
rs774361936 Chr.17: 41395114-41395114 on Build GRCh38
rs139879817 Chr.17: 41394883-41394883 on Build GRCh38
rs772110327 Chr.17: 41394909-41394909 on Build GRCh38
rs372458218 Chr.17: 41394809-41394809 on Build GRCh38
rs893811375 Chr.17: 41394788-41394788 on Build GRCh38
rs376679048 Chr.17: 41394823-41394823 on Build GRCh38
rs774179895 Chr.17: 41394859-41394859 on Build GRCh38
rs377742443 Chr.17: 41395213-41395213 on Build GRCh38
rs746975526 Chr.17: 41395030-41395030 on Build GRCh38
rs745544228 Chr.17: 41394808-41394808 on Build GRCh38
rs142694941 Chr.17: 41394985-41394985 on Build GRCh38
rs185105039 Chr.17: 41395142-41395142 on Build GRCh38
rs754481069 Chr.17: 41395073-41395073 on Build GRCh38
rs777175784 Chr.17: 41395215-41395215 on Build GRCh38
rs537121079 Chr.17: 41395037-41395037 on Build GRCh38
rs764835624 Chr.17: 41395220-41395220 on Build GRCh38
rs751883392 Chr.17: 41394920-41394920 on Build GRCh38
rs935561751 Chr.17: 41394816-41394816 on Build GRCh38
rs139368068 Chr.17: 41394984-41394984 on Build GRCh38
rs768160718 Chr.17: 41394790-41394793 on Build GRCh38
rs374699090 Chr.17: 41394905-41394905 on Build GRCh38
rs747946461 Chr.17: 41394952-41394952 on Build GRCh38
rs202028025 Chr.17: 41394861-41394861 on Build GRCh38
rs370933699 Chr.17: 41395216-41395216 on Build GRCh38
rs763186214 Chr.17: 41394911-41394911 on Build GRCh38
rs372124501 Chr.17: 41395193-41395193 on Build GRCh38
rs76174069 Chr.17: 41394868-41394868 on Build GRCh38
rs774598805 Chr.17: 41395046-41395046 on Build GRCh38
rs1042398514 Chr.17: 41394810-41394810 on Build GRCh38
rs1004586037 Chr.17: 41395068-41395068 on Build GRCh38
rs968167368 Chr.17: 41395140-41395140 on Build GRCh38
rs982764075 Chr.17: 41394915-41394915 on Build GRCh38
rs768309821 Chr.17: 41394894-41394894 on Build GRCh38
rs372409338 Chr.17: 41394968-41394968 on Build GRCh38
rs563881573 Chr.17: 41395179-41395179 on Build GRCh38
rs994056132 Chr.17: 41395086-41395086 on Build GRCh38
rs770016719 Chr.17: 41394972-41394972 on Build GRCh38
rs575768087 Chr.17: 41395180-41395180 on Build GRCh38
rs976236445 Chr.17: 41395191-41395191 on Build GRCh38
rs866902276 Chr.17: 41395162-41395162 on Build GRCh38
rs777663715 Chr.17: 41395029-41395029 on Build GRCh38
rs1031193822 Chr.17: 41395147-41395147 on Build GRCh38
rs770834831 Chr.17: 41395036-41395036 on Build GRCh38
rs140159560 Chr.17: 41395174-41395174 on Build GRCh38
rs757199142 Chr.17: 41395093-41395093 on Build GRCh38
rs370138904 Chr.17: 41394821-41394821 on Build GRCh38
rs749999706 Chr.17: 41394925-41394925 on Build GRCh38
rs750810723 Chr.17: 41395013-41395019 on Build GRCh38
rs745319380 Chr.17: 41394875-41394875 on Build GRCh38
rs765816116 Chr.17: 41395002-41395002 on Build GRCh38
rs780695551 Chr.17: 41394873-41394873 on Build GRCh38
rs752841027 Chr.17: 41394907-41394907 on Build GRCh38
rs766584606 Chr.17: 41394837-41394837 on Build GRCh38
rs748397223 Chr.17: 41395115-41395115 on Build GRCh38
rs371841210 Chr.17: 41394928-41394928 on Build GRCh38
rs372890277 Chr.17: 41394819-41394819 on Build GRCh38
rs774142307 Chr.17: 41394900-41394900 on Build GRCh38

View Full Product Details