Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 7256918-7257447 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STS
Entrez Gene ID
412
Gene Name
steroid sulfatase (microsomal), isozyme S
Gene Aliases
ARSC, ARSC1, ASC, ES, SSDD, XLI
UniGene
Hs.522578
Gene Chromosome Location
Chr.X: 7147252-7354643 on Build GRCh38
Transcripts
NM_000351.5 NM_001320750.1 NM_001320751.1 NM_001320752.1 NM_001320753.1 NM_001320754.1

SNP Detail

SNP ID Chromosome Location
rs776067214 Chr.X: 7257403-7257403 on Build GRCh38
rs189905493 Chr.X: 7257383-7257383 on Build GRCh38
rs369635100 Chr.X: 7257293-7257293 on Build GRCh38
rs759057849 Chr.X: 7257395-7257395 on Build GRCh38
rs756268990 Chr.X: 7257372-7257372 on Build GRCh38
rs750716469 Chr.X: 7257382-7257382 on Build GRCh38
rs757048542 Chr.X: 7257197-7257197 on Build GRCh38
rs942514642 Chr.X: 7257151-7257151 on Build GRCh38
rs769521230 Chr.X: 7257308-7257308 on Build GRCh38
rs751973801 Chr.X: 7257254-7257254 on Build GRCh38
rs376655549 Chr.X: 7257234-7257234 on Build GRCh38
rs568827065 Chr.X: 7257136-7257136 on Build GRCh38
rs749108195 Chr.X: 7257373-7257373 on Build GRCh38
rs759997236 Chr.X: 7257245-7257245 on Build GRCh38
rs150111371 Chr.X: 7257242-7257242 on Build GRCh38
rs371667398 Chr.X: 7257129-7257129 on Build GRCh38
rs753698397 Chr.X: 7257191-7257191 on Build GRCh38
rs760368070 Chr.X: 7257258-7257258 on Build GRCh38
rs770607867 Chr.X: 7257374-7257374 on Build GRCh38
rs769969361 Chr.X: 7257232-7257232 on Build GRCh38
rs919221011 Chr.X: 7256990-7256990 on Build GRCh38
rs756110481 Chr.X: 7257414-7257414 on Build GRCh38
rs930653121 Chr.X: 7257067-7257067 on Build GRCh38
rs757758272 Chr.X: 7257259-7257259 on Build GRCh38
rs778764828 Chr.X: 7257199-7257199 on Build GRCh38
rs900918453 Chr.X: 7257174-7257174 on Build GRCh38
rs771440377 Chr.X: 7257238-7257238 on Build GRCh38
rs773065157 Chr.X: 7257368-7257370 on Build GRCh38
rs138730860 Chr.X: 7257271-7257271 on Build GRCh38
rs369366546 Chr.X: 7257404-7257404 on Build GRCh38
rs372806768 Chr.X: 7257425-7257425 on Build GRCh38
rs765707029 Chr.X: 7257073-7257073 on Build GRCh38
rs763668550 Chr.X: 7257268-7257268 on Build GRCh38
rs757189309 Chr.X: 7257211-7257211 on Build GRCh38
rs1039571895 Chr.X: 7257159-7257159 on Build GRCh38
rs780197948 Chr.X: 7257165-7257165 on Build GRCh38
rs750342991 Chr.X: 7257300-7257300 on Build GRCh38
rs1048765860 Chr.X: 7257125-7257125 on Build GRCh38
rs373509256 Chr.X: 7257336-7257336 on Build GRCh38
rs781171118 Chr.X: 7257224-7257224 on Build GRCh38
rs758637471 Chr.X: 7257156-7257156 on Build GRCh38
rs781320111 Chr.X: 7257332-7257332 on Build GRCh38
rs773710811 Chr.X: 7257233-7257233 on Build GRCh38
rs768177499 Chr.X: 7257412-7257412 on Build GRCh38
rs972037871 Chr.X: 7256974-7256974 on Build GRCh38
rs771387293 Chr.X: 7257379-7257379 on Build GRCh38
rs776225009 Chr.X: 7257422-7257422 on Build GRCh38
rs756997415 Chr.X: 7257279-7257279 on Build GRCh38
rs1043471103 Chr.X: 7257076-7257076 on Build GRCh38
rs755088803 Chr.X: 7257236-7257236 on Build GRCh38
rs770947575 Chr.X: 7257223-7257224 on Build GRCh38
rs755047283 Chr.X: 7257201-7257201 on Build GRCh38
rs757644040 Chr.X: 7256992-7256992 on Build GRCh38
rs187669216 Chr.X: 7257255-7257255 on Build GRCh38
rs747087410 Chr.X: 7257226-7257226 on Build GRCh38

View Full Product Details