Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 10445401-10445917 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MID1
Entrez Gene ID
4281
Gene Name
midline 1
Gene Aliases
BBBG1, FXY, GBBB1, MIDIN, OGS1, OS, OSX, RNF59, TRIM18, XPRF, ZNFXY
UniGene
Hs.27695
Gene Chromosome Location
Chr.X: 10445310-11111264 on Build GRCh38
Transcripts
NM_000381.3 NM_001098624.2 NM_001193277.1 NM_033289.1 NM_033290.3 XM_005274536.1 XM_005274537.1 XM_006724492.3 XM_011545525.2 XM_011545526.2 XM_011545527.2 XM_017029534.1 XM_017029535.1 XM_017029536.1 XM_017029537.1 XM_017029538.1 XM_017029539.1

SNP Detail

SNP ID Chromosome Location
rs993853602 Chr.X: 10445580-10445580 on Build GRCh38
rs747762216 Chr.X: 10445550-10445550 on Build GRCh38
rs945948248 Chr.X: 10445488-10445488 on Build GRCh38
rs979644904 Chr.X: 10445652-10445652 on Build GRCh38
rs997596084 Chr.X: 10445438-10445438 on Build GRCh38
rs981141742 Chr.X: 10445494-10445494 on Build GRCh38
rs986932353 Chr.X: 10445888-10445888 on Build GRCh38
rs898109814 Chr.X: 10445727-10445727 on Build GRCh38
rs776115133 Chr.X: 10445749-10445749 on Build GRCh38
rs898051570 Chr.X: 10445568-10445568 on Build GRCh38
rs991916372 Chr.X: 10445624-10445624 on Build GRCh38
rs923265697 Chr.X: 10445759-10445759 on Build GRCh38
rs998137990 Chr.X: 10445743-10445743 on Build GRCh38
rs990373151 Chr.X: 10445460-10445460 on Build GRCh38
rs1023930828 Chr.X: 10445626-10445626 on Build GRCh38
rs769801693 Chr.X: 10445892-10445892 on Build GRCh38
rs969253733 Chr.X: 10445643-10445643 on Build GRCh38
rs183965210 Chr.X: 10445718-10445718 on Build GRCh38
rs925219015 Chr.X: 10445693-10445693 on Build GRCh38
rs1025297615 Chr.X: 10445582-10445582 on Build GRCh38
rs192588923 Chr.X: 10445800-10445800 on Build GRCh38
rs190766836 Chr.X: 10445653-10445653 on Build GRCh38
rs139026561 Chr.X: 10445832-10445832 on Build GRCh38
rs772761314 Chr.X: 10445732-10445732 on Build GRCh38
rs778917321 Chr.X: 10445503-10445504 on Build GRCh38
rs960474842 Chr.X: 10445589-10445589 on Build GRCh38
rs1051065368 Chr.X: 10445477-10445477 on Build GRCh38
rs188862193 Chr.X: 10445742-10445742 on Build GRCh38
rs572722230 Chr.X: 10445845-10445845 on Build GRCh38
rs1055521831 Chr.X: 10445617-10445617 on Build GRCh38
rs889287284 Chr.X: 10445495-10445495 on Build GRCh38
rs889086851 Chr.X: 10445863-10445863 on Build GRCh38
rs777184185 Chr.X: 10445825-10445825 on Build GRCh38
rs779273941 Chr.X: 10445526-10445526 on Build GRCh38

View Full Product Details