Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 132878814-132879311 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPACA9
Entrez Gene ID
11092
Gene Name
sperm acrosome associated 9
Gene Aliases
C9orf9, Mast
UniGene
Hs.62595
Gene Chromosome Location
Chr.9: 132878320-132890314 on Build GRCh38
Transcripts
NM_001316897.1 NM_001316898.1 NM_001316900.1 NM_018956.4 NR_133631.1 XM_017014231.1

SNP Detail

SNP ID Chromosome Location
rs976988753 Chr.9: 132879087-132879087 on Build GRCh38
rs574290426 Chr.9: 132879195-132879195 on Build GRCh38
rs866093616 Chr.9: 132878997-132878997 on Build GRCh38
rs111490056 Chr.9: 132879100-132879100 on Build GRCh38
rs1001591535 Chr.9: 132878963-132878963 on Build GRCh38
rs1053269812 Chr.9: 132878877-132878877 on Build GRCh38
rs985037859 Chr.9: 132879096-132879096 on Build GRCh38
rs1027659910 Chr.9: 132878979-132878979 on Build GRCh38
rs541038157 Chr.9: 132879226-132879226 on Build GRCh38
rs905990442 Chr.9: 132879235-132879235 on Build GRCh38
rs887807330 Chr.9: 132879243-132879243 on Build GRCh38
rs940323312 Chr.9: 132879142-132879142 on Build GRCh38
rs985138649 Chr.9: 132879113-132879113 on Build GRCh38
rs1014817579 Chr.9: 132878913-132878913 on Build GRCh38
rs1052291502 Chr.9: 132879210-132879210 on Build GRCh38
rs574978514 Chr.9: 132879234-132879234 on Build GRCh38
rs903693838 Chr.9: 132878879-132878879 on Build GRCh38
rs767145455 Chr.9: 132879072-132879072 on Build GRCh38
rs557919125 Chr.9: 132878856-132878856 on Build GRCh38
rs953938466 Chr.9: 132879111-132879111 on Build GRCh38
rs1045851986 Chr.9: 132878946-132878946 on Build GRCh38
rs935654118 Chr.9: 132879139-132879139 on Build GRCh38
rs1015560075 Chr.9: 132879277-132879277 on Build GRCh38
rs368793783 Chr.9: 132879229-132879229 on Build GRCh38
rs879319191 Chr.9: 132879144-132879144 on Build GRCh38
rs753908634 Chr.9: 132879160-132879160 on Build GRCh38
rs1005486394 Chr.9: 132879062-132879062 on Build GRCh38
rs947762734 Chr.9: 132879163-132879163 on Build GRCh38
rs1004868543 Chr.9: 132879264-132879264 on Build GRCh38
rs961260323 Chr.9: 132879084-132879084 on Build GRCh38
rs922413049 Chr.9: 132879103-132879103 on Build GRCh38
rs913470157 Chr.9: 132879183-132879183 on Build GRCh38
rs1015507630 Chr.9: 132879080-132879080 on Build GRCh38
rs193211928 Chr.9: 132879130-132879130 on Build GRCh38
rs146146761 Chr.9: 132879165-132879165 on Build GRCh38
rs761489297 Chr.9: 132879003-132879003 on Build GRCh38
rs774178175 Chr.9: 132878897-132878897 on Build GRCh38
rs1049145310 Chr.9: 132879242-132879242 on Build GRCh38
rs1010222513 Chr.9: 132878995-132878995 on Build GRCh38
rs866796114 Chr.9: 132879278-132879278 on Build GRCh38
rs966503510 Chr.9: 132879000-132879000 on Build GRCh38
rs974867485 Chr.9: 132879011-132879011 on Build GRCh38
rs374736321 Chr.9: 132878954-132878954 on Build GRCh38
rs76110257 Chr.9: 132879239-132879239 on Build GRCh38
rs766831877 Chr.9: 132879198-132879198 on Build GRCh38

View Full Product Details