Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 60785573-60786072 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK1
Entrez Gene ID
983
Gene Name
cyclin dependent kinase 1
Gene Aliases
CDC2, CDC28A, P34CDC2
UniGene
Hs.732435
Gene Chromosome Location
Chr.10: 60778331-60794852 on Build GRCh38
Transcripts
NM_001170406.1 NM_001170407.1 NM_001320918.1 NM_001786.4 NM_033379.4 XM_005270303.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1348487 c.228A>G p.L76L Chr.10: 60785697-60785697 on Build GRCh38
366782 c.202G>A p.D68N Chr.10: 60785671-60785671 on Build GRCh38
5037133 c.259C>T p.L87L Chr.10: 60785728-60785728 on Build GRCh38
6849116 c.250T>C p.S84P Chr.10: 60785719-60785719 on Build GRCh38
7095487 c.254T>C p.M85T Chr.10: 60785723-60785723 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374332309 Chr.10: 60785767-60785767 on Build GRCh38
rs752848484 Chr.10: 60785618-60785618 on Build GRCh38
rs780820433 Chr.10: 60785842-60785842 on Build GRCh38
rs758387455 Chr.10: 60785828-60785828 on Build GRCh38
rs985951399 Chr.10: 60785674-60785674 on Build GRCh38
rs148632129 Chr.10: 60785697-60785697 on Build GRCh38
rs758106128 Chr.10: 60785647-60785647 on Build GRCh38
rs750432624 Chr.10: 60785645-60785645 on Build GRCh38
rs750048290 Chr.10: 60785956-60785956 on Build GRCh38
rs147689519 Chr.10: 60785740-60785740 on Build GRCh38
rs1045871727 Chr.10: 60785957-60785957 on Build GRCh38
rs768316764 Chr.10: 60785673-60785673 on Build GRCh38
rs778987482 Chr.10: 60785633-60785633 on Build GRCh38
rs565964874 Chr.10: 60785839-60785839 on Build GRCh38
rs760824235 Chr.10: 60785814-60785814 on Build GRCh38
rs11540347 Chr.10: 60785683-60785683 on Build GRCh38
rs893143489 Chr.10: 60786020-60786020 on Build GRCh38
rs757565151 Chr.10: 60785645-60785646 on Build GRCh38
rs2261327 Chr.10: 60786019-60786019 on Build GRCh38
rs779662361 Chr.10: 60785829-60785829 on Build GRCh38
rs779935732 Chr.10: 60785650-60785650 on Build GRCh38
rs907327895 Chr.10: 60785963-60785963 on Build GRCh38
rs1023381864 Chr.10: 60785890-60785890 on Build GRCh38
rs12415622 Chr.10: 60785997-60785997 on Build GRCh38
rs867317733 Chr.10: 60785819-60785819 on Build GRCh38
rs780782626 Chr.10: 60785650-60785650 on Build GRCh38
rs999518675 Chr.10: 60786001-60786001 on Build GRCh38
rs770857106 Chr.10: 60785748-60785748 on Build GRCh38
rs935481280 Chr.10: 60785679-60785679 on Build GRCh38
rs532901473 Chr.10: 60786045-60786045 on Build GRCh38
rs753032757 Chr.10: 60785811-60785811 on Build GRCh38
rs567245939 Chr.10: 60786035-60786035 on Build GRCh38
rs750292377 Chr.10: 60785825-60785825 on Build GRCh38
rs764281223 Chr.10: 60785631-60785631 on Build GRCh38
rs1048820261 Chr.10: 60785854-60785854 on Build GRCh38
rs951974005 Chr.10: 60786015-60786015 on Build GRCh38
rs757192370 Chr.10: 60785622-60785622 on Build GRCh38
rs370521219 Chr.10: 60785657-60785657 on Build GRCh38
rs948924059 Chr.10: 60785934-60785934 on Build GRCh38
rs142079607 Chr.10: 60785822-60785822 on Build GRCh38
rs144445383 Chr.10: 60785739-60785739 on Build GRCh38
rs570545366 Chr.10: 60785612-60785613 on Build GRCh38
rs373485880 Chr.10: 60785766-60785766 on Build GRCh38
rs1011679452 Chr.10: 60785615-60785615 on Build GRCh38
rs774156794 Chr.10: 60785869-60785869 on Build GRCh38
rs771169549 Chr.10: 60785763-60785763 on Build GRCh38
rs751716206 Chr.10: 60785639-60785640 on Build GRCh38
rs747663798 Chr.10: 60785688-60785688 on Build GRCh38
rs759629291 Chr.10: 60785770-60785770 on Build GRCh38
rs767855246 Chr.10: 60785801-60785801 on Build GRCh38
rs780626115 Chr.10: 60785677-60785677 on Build GRCh38
rs1027537783 Chr.10: 60785976-60785976 on Build GRCh38

View Full Product Details