Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.Y: 20575633-20576133 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF1AY
Entrez Gene ID
9086
Gene Name
eukaryotic translation initiation factor 1A, Y-linked
Gene Aliases
eIF-4C
UniGene
Hs.461178
Gene Chromosome Location
Chr.Y: 20575711-20593154 on Build GRCh38
Transcripts
NM_001278612.1 NM_004681.3

SNP Detail

SNP ID Chromosome Location
rs780651102 Chr.Y: 20575913-20575913 on Build GRCh38
rs756693957 Chr.Y: 20575903-20575903 on Build GRCh38
rs938508124 Chr.Y: 20575685-20575685 on Build GRCh38
rs987752557 Chr.Y: 20575849-20575849 on Build GRCh38
rs776035076 Chr.Y: 20575880-20575880 on Build GRCh38
rs763383187 Chr.Y: 20575771-20575771 on Build GRCh38
rs967799872 Chr.Y: 20575925-20575925 on Build GRCh38
rs9341310 Chr.Y: 20575915-20575915 on Build GRCh38
rs1019822058 Chr.Y: 20575768-20575768 on Build GRCh38
rs796263440 Chr.Y: 20576062-20576062 on Build GRCh38
rs979677748 Chr.Y: 20575990-20575990 on Build GRCh38
rs750495416 Chr.Y: 20575894-20575894 on Build GRCh38
rs376992333 Chr.Y: 20575870-20575870 on Build GRCh38

View Full Product Details