Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 44550853-44551405 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRTAP10-2
Entrez Gene ID
386679
Gene Name
keratin associated protein 10-2
Gene Aliases
KAP10.2, KAP18-2, KAP18.2, KRTAP10.2, KRTAP18-2, KRTAP18.2
UniGene
Hs.664660
Gene Chromosome Location
Chr.21: 44550357-44551505 on Build GRCh38
Transcripts
NM_198693.3 NR_130165.1 NM_001272037.1 NM_144991.2

Gene Symbol
TSPEAR
Entrez Gene ID
54084
Gene Name
thrombospondin type laminin G domain and EAR repeats
Gene Aliases
C21orf29, DFNB98, TSP-EAR
UniGene
Hs.660703
Gene Chromosome Location
Chr.21: 44497892-44711580 on Build GRCh38
Transcripts
NM_198693.3 NR_130165.1 NM_001272037.1 NM_144991.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3766665 c.319C>A p.P107T Chr.21: 44551140-44551140 on Build GRCh38
5729663 c.175C>T p.Q59* Chr.21: 44551284-44551284 on Build GRCh38
6840155 c.311G>A p.C104Y Chr.21: 44551148-44551148 on Build GRCh38
6094865 c.145C>G p.P49A Chr.21: 44551314-44551314 on Build GRCh38
725873 c.161C>T p.S54F Chr.21: 44551298-44551298 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782382021 Chr.21: 44551298-44551298 on Build GRCh38
rs782297019 Chr.21: 44551346-44551346 on Build GRCh38
rs78661539 Chr.21: 44551231-44551231 on Build GRCh38
rs782150986 Chr.21: 44551109-44551109 on Build GRCh38
rs374244983 Chr.21: 44550917-44550917 on Build GRCh38
rs782160184 Chr.21: 44550951-44550951 on Build GRCh38
rs782693221 Chr.21: 44551335-44551335 on Build GRCh38
rs382226 Chr.21: 44551027-44551027 on Build GRCh38
rs782755548 Chr.21: 44550877-44550877 on Build GRCh38
rs587707007 Chr.21: 44551179-44551179 on Build GRCh38
rs782099572 Chr.21: 44550957-44550957 on Build GRCh38
rs199695232 Chr.21: 44551196-44551196 on Build GRCh38
rs781965001 Chr.21: 44551207-44551207 on Build GRCh38
rs782333206 Chr.21: 44550955-44550955 on Build GRCh38
rs782122143 Chr.21: 44551006-44551006 on Build GRCh38
rs782030252 Chr.21: 44551386-44551386 on Build GRCh38
rs782517945 Chr.21: 44551138-44551138 on Build GRCh38
rs782653798 Chr.21: 44550896-44550896 on Build GRCh38
rs782662252 Chr.21: 44551199-44551199 on Build GRCh38
rs587746517 Chr.21: 44550918-44550918 on Build GRCh38
rs781847897 Chr.21: 44551186-44551186 on Build GRCh38
rs368800676 Chr.21: 44551365-44551365 on Build GRCh38
rs782742490 Chr.21: 44551121-44551121 on Build GRCh38
rs782002498 Chr.21: 44551328-44551328 on Build GRCh38
rs782638553 Chr.21: 44551005-44551005 on Build GRCh38
rs781919556 Chr.21: 44551115-44551115 on Build GRCh38
rs782190064 Chr.21: 44551286-44551286 on Build GRCh38
rs200608605 Chr.21: 44551281-44551281 on Build GRCh38
rs782430780 Chr.21: 44551204-44551204 on Build GRCh38
rs782378471 Chr.21: 44551375-44551375 on Build GRCh38
rs371219783 Chr.21: 44550881-44550881 on Build GRCh38
rs782349134 Chr.21: 44551244-44551244 on Build GRCh38
rs782690145 Chr.21: 44551030-44551030 on Build GRCh38
rs782417897 Chr.21: 44551152-44551152 on Build GRCh38
rs782263136 Chr.21: 44551294-44551294 on Build GRCh38
rs782255065 Chr.21: 44551373-44551373 on Build GRCh38
rs369556022 Chr.21: 44551153-44551153 on Build GRCh38
rs781891072 Chr.21: 44551264-44551264 on Build GRCh38
rs782364167 Chr.21: 44551178-44551178 on Build GRCh38
rs201722971 Chr.21: 44551187-44551187 on Build GRCh38
rs587654793 Chr.21: 44551293-44551293 on Build GRCh38
rs478967 Chr.21: 44551140-44551140 on Build GRCh38
rs587678058 Chr.21: 44551122-44551122 on Build GRCh38
rs374290804 Chr.21: 44551239-44551239 on Build GRCh38
rs782442914 Chr.21: 44551130-44551130 on Build GRCh38
rs781926576 Chr.21: 44550956-44550956 on Build GRCh38
rs782653408 Chr.21: 44551039-44551039 on Build GRCh38
rs782794896 Chr.21: 44551123-44551123 on Build GRCh38
rs782026527 Chr.21: 44551348-44551348 on Build GRCh38
rs782564487 Chr.21: 44551198-44551198 on Build GRCh38
rs782183041 Chr.21: 44551158-44551158 on Build GRCh38
rs782033116 Chr.21: 44551102-44551102 on Build GRCh38
rs781806951 Chr.21: 44551200-44551200 on Build GRCh38
rs78215665 Chr.21: 44551007-44551007 on Build GRCh38
rs372107705 Chr.21: 44551338-44551338 on Build GRCh38
rs587609542 Chr.21: 44551053-44551053 on Build GRCh38
rs782722193 Chr.21: 44551183-44551183 on Build GRCh38
rs782095780 Chr.21: 44551322-44551322 on Build GRCh38
rs782280768 Chr.21: 44551304-44551304 on Build GRCh38
rs186057899 Chr.21: 44551369-44551369 on Build GRCh38
rs374542929 Chr.21: 44551077-44551077 on Build GRCh38
rs587645783 Chr.21: 44551167-44551167 on Build GRCh38
rs782218527 Chr.21: 44551137-44551137 on Build GRCh38
rs199607801 Chr.21: 44551269-44551269 on Build GRCh38
rs587750879 Chr.21: 44551280-44551280 on Build GRCh38
rs782386244 Chr.21: 44550938-44550938 on Build GRCh38
rs782096863 Chr.21: 44551119-44551119 on Build GRCh38
rs782116286 Chr.21: 44551329-44551329 on Build GRCh38
rs200441874 Chr.21: 44550904-44550904 on Build GRCh38
rs201338715 Chr.21: 44551258-44551258 on Build GRCh38
rs782242774 Chr.21: 44551044-44551044 on Build GRCh38
rs233239 Chr.21: 44551110-44551110 on Build GRCh38
rs781922416 Chr.21: 44551318-44551318 on Build GRCh38
rs782138371 Chr.21: 44551181-44551181 on Build GRCh38
rs782658027 Chr.21: 44551282-44551282 on Build GRCh38
rs781956269 Chr.21: 44551083-44551083 on Build GRCh38
rs782072870 Chr.21: 44550882-44550882 on Build GRCh38
rs587665326 Chr.21: 44551097-44551097 on Build GRCh38
rs782215534 Chr.21: 44551212-44551212 on Build GRCh38
rs782451436 Chr.21: 44551334-44551334 on Build GRCh38
rs71199610 Chr.21: 44550889-44550903 on Build GRCh38
rs200496560 Chr.21: 44550928-44550928 on Build GRCh38
rs587667536 Chr.21: 44551342-44551342 on Build GRCh38
rs200799981 Chr.21: 44551263-44551263 on Build GRCh38
rs587774645 Chr.21: 44550949-44550949 on Build GRCh38
rs782103907 Chr.21: 44551232-44551232 on Build GRCh38
rs781855635 Chr.21: 44550898-44550898 on Build GRCh38
rs76536096 Chr.21: 44550891-44550891 on Build GRCh38
rs782717977 Chr.21: 44551324-44551324 on Build GRCh38
rs782586482 Chr.21: 44551356-44551356 on Build GRCh38
rs782667674 Chr.21: 44550947-44550948 on Build GRCh38
rs782555601 Chr.21: 44550903-44550903 on Build GRCh38
rs782750786 Chr.21: 44550967-44550967 on Build GRCh38
rs782364830 Chr.21: 44551080-44551080 on Build GRCh38
rs781834518 Chr.21: 44551274-44551274 on Build GRCh38
rs202199029 Chr.21: 44550952-44550952 on Build GRCh38
rs76021731 Chr.21: 44550889-44550889 on Build GRCh38
rs781884692 Chr.21: 44551125-44551125 on Build GRCh38
rs782129000 Chr.21: 44550876-44550876 on Build GRCh38
rs587749085 Chr.21: 44551101-44551101 on Build GRCh38
rs782331094 Chr.21: 44551305-44551305 on Build GRCh38
rs587703858 Chr.21: 44551300-44551300 on Build GRCh38
rs2329834 Chr.21: 44550929-44550929 on Build GRCh38
rs782660586 Chr.21: 44551069-44551069 on Build GRCh38
rs781880015 Chr.21: 44551332-44551332 on Build GRCh38
rs587596519 Chr.21: 44551021-44551021 on Build GRCh38
rs782283560 Chr.21: 44551225-44551225 on Build GRCh38
rs782192602 Chr.21: 44550906-44550906 on Build GRCh38
rs782545109 Chr.21: 44551057-44551057 on Build GRCh38
rs587724578 Chr.21: 44551358-44551358 on Build GRCh38
rs782534140 Chr.21: 44551339-44551339 on Build GRCh38
rs782475851 Chr.21: 44550895-44550895 on Build GRCh38
rs782391999 Chr.21: 44551223-44551223 on Build GRCh38
rs782477465 Chr.21: 44551277-44551277 on Build GRCh38
rs150498297 Chr.21: 44551037-44551037 on Build GRCh38
rs782613126 Chr.21: 44551205-44551205 on Build GRCh38
rs113146284 Chr.21: 44550890-44550890 on Build GRCh38
rs478947 Chr.21: 44551135-44551135 on Build GRCh38
rs373244856 Chr.21: 44551159-44551159 on Build GRCh38
rs201944330 Chr.21: 44551038-44551038 on Build GRCh38
rs587713619 Chr.21: 44551015-44551015 on Build GRCh38
rs587610872 Chr.21: 44551330-44551330 on Build GRCh38
rs200215960 Chr.21: 44550888-44550888 on Build GRCh38

View Full Product Details