Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 8530589-8531175 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANOS1
Entrez Gene ID
3730
Gene Name
anosmin 1
Gene Aliases
ADMLX, HH1, HHA, KAL, KAL1, KALIG-1, KMS, WFDC19
UniGene
Hs.521869
Gene Chromosome Location
Chr.X: 8528874-8732187 on Build GRCh38
Transcripts
NM_000216.3

SNP Detail

SNP ID Chromosome Location
rs762236639 Chr.X: 8530904-8530904 on Build GRCh38
rs6530182 Chr.X: 8531045-8531045 on Build GRCh38
rs6530183 Chr.X: 8531066-8531066 on Build GRCh38
rs180842620 Chr.X: 8530814-8530814 on Build GRCh38
rs749516324 Chr.X: 8530630-8530630 on Build GRCh38
rs6638834 Chr.X: 8530753-8530753 on Build GRCh38
rs887179041 Chr.X: 8530770-8530770 on Build GRCh38
rs959185537 Chr.X: 8530909-8530909 on Build GRCh38
rs1001788317 Chr.X: 8530738-8530738 on Build GRCh38
rs808127 Chr.X: 8530938-8530938 on Build GRCh38
rs184924343 Chr.X: 8530958-8530958 on Build GRCh38
rs781063587 Chr.X: 8531104-8531104 on Build GRCh38
rs188432292 Chr.X: 8530658-8530658 on Build GRCh38
rs5978893 Chr.X: 8530834-8530834 on Build GRCh38
rs199697084 Chr.X: 8530818-8530818 on Build GRCh38
rs1034617008 Chr.X: 8530751-8530751 on Build GRCh38
rs1016827302 Chr.X: 8531109-8531109 on Build GRCh38
rs5978894 Chr.X: 8531038-8531038 on Build GRCh38
rs757062981 Chr.X: 8531087-8531087 on Build GRCh38
rs906208917 Chr.X: 8530632-8530632 on Build GRCh38
rs6530184 Chr.X: 8531084-8531084 on Build GRCh38
rs369900751 Chr.X: 8530835-8530835 on Build GRCh38
rs6530181 Chr.X: 8530989-8530989 on Build GRCh38
rs1006409458 Chr.X: 8530939-8530939 on Build GRCh38
rs750327306 Chr.X: 8531125-8531125 on Build GRCh38
rs763659170 Chr.X: 8531072-8531072 on Build GRCh38

View Full Product Details