Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 4055085-4055677 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STIM1
Entrez Gene ID
6786
Gene Name
stromal interaction molecule 1
Gene Aliases
D11S4896E, GOK, IMD10, STRMK, TAM, TAM1
UniGene
Hs.501735
Gene Chromosome Location
Chr.11: 3855703-4093210 on Build GRCh38
Transcripts
NM_001277961.1 NM_001277962.1 NM_003156.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4447246 c.497+3C>T p.? Chr.11: 4055640-4055640 on Build GRCh38
4921703 c.462C>A p.F154L Chr.11: 4055602-4055602 on Build GRCh38
5372459 c.462C>T p.F154F Chr.11: 4055602-4055602 on Build GRCh38
5453081 c.387A>G p.V129V Chr.11: 4055527-4055527 on Build GRCh38
6331154 c.482G>T p.G161V Chr.11: 4055622-4055622 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1010907523 Chr.11: 4055271-4055271 on Build GRCh38
rs144602692 Chr.11: 4055598-4055598 on Build GRCh38
rs1015857908 Chr.11: 4055431-4055431 on Build GRCh38
rs149043599 Chr.11: 4055398-4055398 on Build GRCh38
rs771484089 Chr.11: 4055604-4055604 on Build GRCh38
rs770062289 Chr.11: 4055140-4055140 on Build GRCh38
rs1022693760 Chr.11: 4055356-4055356 on Build GRCh38
rs199513192 Chr.11: 4055539-4055539 on Build GRCh38
rs767039902 Chr.11: 4055507-4055507 on Build GRCh38
rs200734570 Chr.11: 4055615-4055615 on Build GRCh38
rs1038405546 Chr.11: 4055116-4055116 on Build GRCh38
rs766344213 Chr.11: 4055339-4055339 on Build GRCh38
rs1029733316 Chr.11: 4055403-4055403 on Build GRCh38
rs955559046 Chr.11: 4055404-4055404 on Build GRCh38
rs775688262 Chr.11: 4055221-4055221 on Build GRCh38
rs938944632 Chr.11: 4055183-4055183 on Build GRCh38
rs190436143 Chr.11: 4055410-4055410 on Build GRCh38
rs899854132 Chr.11: 4055613-4055613 on Build GRCh38
rs368922449 Chr.11: 4055515-4055515 on Build GRCh38
rs765707086 Chr.11: 4055521-4055521 on Build GRCh38
rs998640673 Chr.11: 4055229-4055229 on Build GRCh38
rs201671402 Chr.11: 4055583-4055583 on Build GRCh38
rs531842584 Chr.11: 4055655-4055655 on Build GRCh38
rs763289208 Chr.11: 4055637-4055637 on Build GRCh38
rs765361905 Chr.11: 4055508-4055508 on Build GRCh38
rs760506908 Chr.11: 4055255-4055255 on Build GRCh38
rs1052103314 Chr.11: 4055443-4055443 on Build GRCh38
rs776853897 Chr.11: 4055504-4055505 on Build GRCh38
rs768152391 Chr.11: 4055494-4055494 on Build GRCh38
rs116855870 Chr.11: 4055527-4055527 on Build GRCh38
rs200648767 Chr.11: 4055548-4055548 on Build GRCh38
rs758479123 Chr.11: 4055537-4055537 on Build GRCh38
rs764275626 Chr.11: 4055533-4055533 on Build GRCh38
rs759599608 Chr.11: 4055462-4055462 on Build GRCh38
rs200086415 Chr.11: 4055654-4055654 on Build GRCh38
rs540350859 Chr.11: 4055124-4055124 on Build GRCh38
rs73431657 Chr.11: 4055380-4055380 on Build GRCh38
rs200824556 Chr.11: 4055567-4055567 on Build GRCh38
rs747832260 Chr.11: 4055603-4055603 on Build GRCh38
rs776121110 Chr.11: 4055630-4055630 on Build GRCh38
rs200659679 Chr.11: 4055645-4055645 on Build GRCh38
rs201882860 Chr.11: 4055626-4055626 on Build GRCh38
rs962932906 Chr.11: 4055433-4055433 on Build GRCh38
rs376008411 Chr.11: 4055317-4055317 on Build GRCh38
rs201990461 Chr.11: 4055647-4055647 on Build GRCh38
rs200837007 Chr.11: 4055491-4055491 on Build GRCh38
rs769583191 Chr.11: 4055485-4055485 on Build GRCh38
rs779857263 Chr.11: 4055572-4055572 on Build GRCh38
rs768689027 Chr.11: 4055590-4055590 on Build GRCh38
rs972076211 Chr.11: 4055161-4055161 on Build GRCh38
rs143916878 Chr.11: 4055594-4055594 on Build GRCh38
rs775209514 Chr.11: 4055489-4055489 on Build GRCh38
rs199728303 Chr.11: 4055552-4055552 on Build GRCh38
rs963476096 Chr.11: 4055556-4055556 on Build GRCh38
rs201274425 Chr.11: 4055532-4055532 on Build GRCh38
rs1016279165 Chr.11: 4055137-4055137 on Build GRCh38
rs181620826 Chr.11: 4055501-4055501 on Build GRCh38
rs915683813 Chr.11: 4055119-4055119 on Build GRCh38
rs113922776 Chr.11: 4055357-4055357 on Build GRCh38
rs772826301 Chr.11: 4055182-4055182 on Build GRCh38
rs531281530 Chr.11: 4055194-4055195 on Build GRCh38
rs529099829 Chr.11: 4055414-4055414 on Build GRCh38
rs200351944 Chr.11: 4055540-4055540 on Build GRCh38
rs373956265 Chr.11: 4055482-4055482 on Build GRCh38
rs1030111673 Chr.11: 4055115-4055115 on Build GRCh38
rs1052997514 Chr.11: 4055248-4055248 on Build GRCh38
rs200227241 Chr.11: 4055490-4055490 on Build GRCh38
rs74966469 Chr.11: 4055564-4055564 on Build GRCh38
rs199893056 Chr.11: 4055628-4055628 on Build GRCh38
rs774370020 Chr.11: 4055653-4055653 on Build GRCh38

View Full Product Details