Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 35292417-35292919 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF76
Entrez Gene ID
7629
Gene Name
zinc finger protein 76
Gene Aliases
D6S229E, ZNF523, Zfp523
UniGene
Hs.388024
Gene Chromosome Location
Chr.6: 35259473-35295987 on Build GRCh38
Transcripts
NM_001292032.1 NM_003427.4 XM_011514846.2 XM_011514847.2 XM_011514849.2 XM_011514850.2 XM_011514854.2 XM_011514855.2 XM_011514856.2 XM_017011248.1 XM_017011249.1 XM_017011250.1 XM_017011251.1 XM_017011252.1 XM_017011253.1 XM_017011254.1 XM_017011255.1 XM_017011256.1 XM_017011257.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1078324 c.1064G>A p.R355Q Chr.6: 35292686-35292686 on Build GRCh38
5562163 c.960A>C p.P320P Chr.6: 35292582-35292582 on Build GRCh38
1078325 c.1108G>A p.E370K Chr.6: 35292730-35292730 on Build GRCh38
1643046 c.975C>T p.R325R Chr.6: 35292597-35292597 on Build GRCh38
4744901 c.1133A>T p.E378V Chr.6: 35292755-35292755 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772911192 Chr.6: 35292630-35292630 on Build GRCh38
rs4713848 Chr.6: 35292838-35292838 on Build GRCh38
rs367726779 Chr.6: 35292681-35292681 on Build GRCh38
rs976470798 Chr.6: 35292533-35292533 on Build GRCh38
rs761177970 Chr.6: 35292576-35292576 on Build GRCh38
rs775890871 Chr.6: 35292565-35292565 on Build GRCh38
rs754819723 Chr.6: 35292590-35292590 on Build GRCh38
rs1012874320 Chr.6: 35292448-35292448 on Build GRCh38
rs760935688 Chr.6: 35292846-35292846 on Build GRCh38
rs751710296 Chr.6: 35292655-35292655 on Build GRCh38
rs137993036 Chr.6: 35292631-35292631 on Build GRCh38
rs771067952 Chr.6: 35292550-35292550 on Build GRCh38
rs750665514 Chr.6: 35292752-35292752 on Build GRCh38
rs144970619 Chr.6: 35292729-35292729 on Build GRCh38
rs754610632 Chr.6: 35292637-35292638 on Build GRCh38
rs766353659 Chr.6: 35292852-35292852 on Build GRCh38
rs750352396 Chr.6: 35292584-35292584 on Build GRCh38
rs534275666 Chr.6: 35292756-35292756 on Build GRCh38
rs746363363 Chr.6: 35292602-35292602 on Build GRCh38
rs778495738 Chr.6: 35292703-35292704 on Build GRCh38
rs906673519 Chr.6: 35292561-35292561 on Build GRCh38
rs186092203 Chr.6: 35292504-35292504 on Build GRCh38
rs755930667 Chr.6: 35292507-35292507 on Build GRCh38
rs1046671232 Chr.6: 35292792-35292792 on Build GRCh38
rs117241970 Chr.6: 35292813-35292813 on Build GRCh38
rs767165662 Chr.6: 35292582-35292582 on Build GRCh38
rs749832082 Chr.6: 35292688-35292688 on Build GRCh38
rs377680153 Chr.6: 35292588-35292588 on Build GRCh38
rs770122684 Chr.6: 35292560-35292560 on Build GRCh38
rs761565684 Chr.6: 35292612-35292612 on Build GRCh38
rs777813909 Chr.6: 35292593-35292593 on Build GRCh38
rs776404157 Chr.6: 35292718-35292718 on Build GRCh38
rs746648936 Chr.6: 35292895-35292895 on Build GRCh38
rs767670054 Chr.6: 35292744-35292744 on Build GRCh38
rs749735485 Chr.6: 35292767-35292767 on Build GRCh38
rs879649009 Chr.6: 35292458-35292458 on Build GRCh38
rs760217393 Chr.6: 35292716-35292716 on Build GRCh38
rs780489199 Chr.6: 35292686-35292686 on Build GRCh38
rs759569533 Chr.6: 35292722-35292722 on Build GRCh38
rs765122477 Chr.6: 35292648-35292648 on Build GRCh38
rs767112486 Chr.6: 35292621-35292621 on Build GRCh38
rs771933940 Chr.6: 35292673-35292673 on Build GRCh38
rs748355316 Chr.6: 35292530-35292530 on Build GRCh38
rs772591591 Chr.6: 35292791-35292791 on Build GRCh38
rs766364824 Chr.6: 35292635-35292635 on Build GRCh38
rs776265064 Chr.6: 35292812-35292812 on Build GRCh38
rs111486721 Chr.6: 35292490-35292490 on Build GRCh38
rs770887538 Chr.6: 35292595-35292595 on Build GRCh38
rs968283892 Chr.6: 35292857-35292857 on Build GRCh38
rs753748668 Chr.6: 35292589-35292589 on Build GRCh38
rs763907756 Chr.6: 35292837-35292837 on Build GRCh38
rs142958367 Chr.6: 35292609-35292609 on Build GRCh38
rs1557567 Chr.6: 35292726-35292726 on Build GRCh38
rs748775812 Chr.6: 35292611-35292611 on Build GRCh38
rs777531247 Chr.6: 35292795-35292795 on Build GRCh38
rs893975298 Chr.6: 35292853-35292853 on Build GRCh38
rs771912734 Chr.6: 35292710-35292710 on Build GRCh38
rs781245862 Chr.6: 35292672-35292672 on Build GRCh38
rs778065004 Chr.6: 35292542-35292542 on Build GRCh38
rs143517301 Chr.6: 35292568-35292568 on Build GRCh38
rs760077489 Chr.6: 35292559-35292559 on Build GRCh38
rs2210414 Chr.6: 35292544-35292544 on Build GRCh38
rs147863647 Chr.6: 35292567-35292567 on Build GRCh38
rs746905273 Chr.6: 35292789-35292790 on Build GRCh38
rs753291979 Chr.6: 35292879-35292879 on Build GRCh38
rs770546987 Chr.6: 35292808-35292808 on Build GRCh38
rs770872818 Chr.6: 35292832-35292833 on Build GRCh38
rs770526444 Chr.6: 35292638-35292638 on Build GRCh38
rs756166612 Chr.6: 35292585-35292585 on Build GRCh38
rs755563605 Chr.6: 35292769-35292769 on Build GRCh38
rs747289649 Chr.6: 35292594-35292594 on Build GRCh38
rs373946973 Chr.6: 35292656-35292656 on Build GRCh38
rs778333298 Chr.6: 35292888-35292888 on Build GRCh38
rs774825701 Chr.6: 35292704-35292704 on Build GRCh38
rs960666960 Chr.6: 35292882-35292882 on Build GRCh38
rs368442038 Chr.6: 35292885-35292885 on Build GRCh38
rs777732653 Chr.6: 35292773-35292773 on Build GRCh38
rs747006074 Chr.6: 35292548-35292548 on Build GRCh38
rs201236290 Chr.6: 35292890-35292890 on Build GRCh38
rs1010661875 Chr.6: 35292583-35292583 on Build GRCh38
rs747679332 Chr.6: 35292760-35292761 on Build GRCh38
rs763169444 Chr.6: 35292607-35292607 on Build GRCh38
rs371253693 Chr.6: 35292685-35292685 on Build GRCh38
rs1557568 Chr.6: 35292753-35292753 on Build GRCh38
rs754691961 Chr.6: 35292640-35292640 on Build GRCh38
rs750830888 Chr.6: 35292589-35292589 on Build GRCh38
rs199659554 Chr.6: 35292762-35292762 on Build GRCh38
rs141883824 Chr.6: 35292575-35292575 on Build GRCh38
rs770353581 Chr.6: 35292603-35292603 on Build GRCh38
rs777084586 Chr.6: 35292555-35292555 on Build GRCh38
rs151113018 Chr.6: 35292707-35292707 on Build GRCh38
rs775304902 Chr.6: 35292829-35292829 on Build GRCh38
rs779414648 Chr.6: 35292876-35292876 on Build GRCh38
rs144616098 Chr.6: 35292880-35292880 on Build GRCh38
rs781648897 Chr.6: 35292527-35292528 on Build GRCh38
rs911838377 Chr.6: 35292649-35292649 on Build GRCh38
rs768869892 Chr.6: 35292699-35292699 on Build GRCh38
rs769705693 Chr.6: 35292823-35292823 on Build GRCh38
rs796120168 Chr.6: 35292627-35292627 on Build GRCh38
rs1929850 Chr.6: 35292500-35292500 on Build GRCh38
rs368273976 Chr.6: 35292596-35292596 on Build GRCh38
rs779138134 Chr.6: 35292782-35292782 on Build GRCh38
rs769863649 Chr.6: 35292450-35292450 on Build GRCh38
rs764929202 Chr.6: 35292725-35292725 on Build GRCh38
rs773000742 Chr.6: 35292715-35292715 on Build GRCh38
rs772236594 Chr.6: 35292531-35292531 on Build GRCh38
rs555930429 Chr.6: 35292798-35292798 on Build GRCh38
rs771529226 Chr.6: 35292766-35292767 on Build GRCh38
rs978731297 Chr.6: 35292871-35292871 on Build GRCh38
rs755013741 Chr.6: 35292854-35292854 on Build GRCh38
rs780031503 Chr.6: 35292520-35292520 on Build GRCh38
rs775980877 Chr.6: 35292604-35292604 on Build GRCh38
rs999684972 Chr.6: 35292835-35292835 on Build GRCh38
rs775847813 Chr.6: 35292790-35292790 on Build GRCh38

View Full Product Details