Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 38106807-38107310 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASH2L
Entrez Gene ID
9070
Gene Name
ASH2 like histone lysine methyltransferase complex subunit
Gene Aliases
ASH2, ASH2L1, ASH2L2, Bre2
UniGene
Hs.521530
Gene Chromosome Location
Chr.8: 38104965-38140080 on Build GRCh38
Transcripts
NM_001105214.2 NM_001261832.1 NM_001282272.1 NM_004674.4 XM_005273682.1 XM_005273683.1 XM_006716412.1 XM_006716413.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1183764 c.377C>T p.A126V Chr.8: 38107142-38107142 on Build GRCh38
1456875 c.388G>T p.G130C Chr.8: 38107153-38107153 on Build GRCh38
3648804 c.401C>T p.S134L Chr.8: 38107166-38107166 on Build GRCh38
1623982 c.300C>A p.G100G Chr.8: 38107065-38107065 on Build GRCh38
3899931 c.322C>T p.R108* Chr.8: 38107087-38107087 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772333911 Chr.8: 38106991-38106991 on Build GRCh38
rs773161690 Chr.8: 38107155-38107155 on Build GRCh38
rs757573888 Chr.8: 38107073-38107073 on Build GRCh38
rs369743312 Chr.8: 38106979-38106979 on Build GRCh38
rs886371335 Chr.8: 38107257-38107257 on Build GRCh38
rs773038287 Chr.8: 38107001-38107001 on Build GRCh38
rs774256468 Chr.8: 38107176-38107176 on Build GRCh38
rs771041637 Chr.8: 38107173-38107173 on Build GRCh38
rs915460255 Chr.8: 38107119-38107119 on Build GRCh38
rs1008962989 Chr.8: 38106912-38106912 on Build GRCh38
rs564644752 Chr.8: 38106910-38106910 on Build GRCh38
rs533558287 Chr.8: 38107253-38107253 on Build GRCh38
rs564457289 Chr.8: 38106946-38106946 on Build GRCh38
rs573302205 Chr.8: 38106966-38106966 on Build GRCh38
rs767317119 Chr.8: 38107049-38107049 on Build GRCh38
rs202092836 Chr.8: 38107004-38107004 on Build GRCh38
rs749648022 Chr.8: 38106978-38106978 on Build GRCh38
rs143179724 Chr.8: 38107120-38107120 on Build GRCh38
rs759241577 Chr.8: 38107044-38107044 on Build GRCh38
rs779036793 Chr.8: 38106983-38106983 on Build GRCh38
rs890557943 Chr.8: 38106858-38106858 on Build GRCh38
rs930456857 Chr.8: 38107246-38107246 on Build GRCh38
rs373182182 Chr.8: 38107206-38107206 on Build GRCh38
rs1023894388 Chr.8: 38106985-38106985 on Build GRCh38
rs950529731 Chr.8: 38107037-38107037 on Build GRCh38
rs562583983 Chr.8: 38107064-38107064 on Build GRCh38
rs766851936 Chr.8: 38107217-38107217 on Build GRCh38
rs747126735 Chr.8: 38106998-38106998 on Build GRCh38
rs557130867 Chr.8: 38106870-38106870 on Build GRCh38
rs542780276 Chr.8: 38106933-38106933 on Build GRCh38
rs745956030 Chr.8: 38106947-38106947 on Build GRCh38
rs199633161 Chr.8: 38106970-38106970 on Build GRCh38
rs769759600 Chr.8: 38107000-38107000 on Build GRCh38
rs775520503 Chr.8: 38106995-38106995 on Build GRCh38
rs754301632 Chr.8: 38107068-38107068 on Build GRCh38
rs145996576 Chr.8: 38107164-38107164 on Build GRCh38
rs774096806 Chr.8: 38107027-38107027 on Build GRCh38
rs771835003 Chr.8: 38107201-38107201 on Build GRCh38
rs775478149 Chr.8: 38107189-38107189 on Build GRCh38
rs1004731312 Chr.8: 38107270-38107270 on Build GRCh38
rs989786690 Chr.8: 38107102-38107102 on Build GRCh38
rs746116048 Chr.8: 38106984-38106984 on Build GRCh38
rs199885385 Chr.8: 38106992-38106992 on Build GRCh38
rs776909484 Chr.8: 38106915-38106915 on Build GRCh38
rs916238831 Chr.8: 38107152-38107152 on Build GRCh38
rs942792158 Chr.8: 38107254-38107254 on Build GRCh38
rs935422809 Chr.8: 38106900-38106900 on Build GRCh38
rs944782148 Chr.8: 38107198-38107198 on Build GRCh38
rs552671842 Chr.8: 38106942-38106942 on Build GRCh38
rs924089852 Chr.8: 38106878-38106878 on Build GRCh38
rs200780652 Chr.8: 38107122-38107122 on Build GRCh38
rs911857078 Chr.8: 38107188-38107188 on Build GRCh38
rs994700700 Chr.8: 38107003-38107003 on Build GRCh38
rs533033225 Chr.8: 38107113-38107113 on Build GRCh38
rs935453800 Chr.8: 38107211-38107211 on Build GRCh38
rs755635084 Chr.8: 38107062-38107062 on Build GRCh38
rs749938935 Chr.8: 38107121-38107121 on Build GRCh38
rs779231573 Chr.8: 38107087-38107087 on Build GRCh38
rs759470548 Chr.8: 38107194-38107194 on Build GRCh38
rs574658403 Chr.8: 38106835-38106835 on Build GRCh38
rs971136710 Chr.8: 38107045-38107045 on Build GRCh38
rs968315407 Chr.8: 38107191-38107191 on Build GRCh38
rs760208693 Chr.8: 38107193-38107194 on Build GRCh38
rs536056551 Chr.8: 38106868-38106868 on Build GRCh38
rs745507632 Chr.8: 38107287-38107287 on Build GRCh38
rs529037543 Chr.8: 38107026-38107026 on Build GRCh38
rs200960407 Chr.8: 38107061-38107061 on Build GRCh38
rs111567903 Chr.8: 38107139-38107139 on Build GRCh38
rs746275587 Chr.8: 38107089-38107089 on Build GRCh38
rs747054614 Chr.8: 38107140-38107140 on Build GRCh38
rs546359163 Chr.8: 38106902-38106902 on Build GRCh38
rs778209374 Chr.8: 38106976-38106976 on Build GRCh38
rs1040138824 Chr.8: 38107276-38107276 on Build GRCh38
rs762675889 Chr.8: 38107237-38107237 on Build GRCh38
rs151151483 Chr.8: 38107202-38107202 on Build GRCh38
rs768624623 Chr.8: 38107143-38107143 on Build GRCh38
rs898483138 Chr.8: 38107278-38107278 on Build GRCh38
rs142524499 Chr.8: 38106957-38106961 on Build GRCh38

View Full Product Details