Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 38114801-38115332 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASH2L
Entrez Gene ID
9070
Gene Name
ASH2 like histone lysine methyltransferase complex subunit
Gene Aliases
ASH2, ASH2L1, ASH2L2, Bre2
UniGene
Hs.521530
Gene Chromosome Location
Chr.8: 38104965-38140080 on Build GRCh38
Transcripts
NM_001105214.2 NM_001261832.1 NM_001282272.1 NM_004674.4 XM_005273682.1 XM_005273683.1 XM_006716412.1 XM_006716413.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
454519 c.718C>A p.P240T Chr.8: 38114941-38114941 on Build GRCh38
750497 c.748G>T p.D250Y Chr.8: 38114971-38114971 on Build GRCh38
5359022 c.777+2T>C p.? Chr.8: 38115002-38115002 on Build GRCh38
1283532 c.763G>T p.G255* Chr.8: 38114986-38114986 on Build GRCh38
338875 c.682-1G>T p.? Chr.8: 38114904-38114904 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs888553165 Chr.8: 38115223-38115223 on Build GRCh38
rs1024937845 Chr.8: 38115305-38115305 on Build GRCh38
rs753958107 Chr.8: 38114875-38114875 on Build GRCh38
rs374558549 Chr.8: 38115045-38115045 on Build GRCh38
rs774159259 Chr.8: 38115045-38115046 on Build GRCh38
rs757356225 Chr.8: 38114867-38114867 on Build GRCh38
rs923407909 Chr.8: 38115150-38115150 on Build GRCh38
rs749240588 Chr.8: 38114898-38114898 on Build GRCh38
rs769047307 Chr.8: 38115103-38115103 on Build GRCh38
rs761182730 Chr.8: 38115009-38115009 on Build GRCh38
rs750520398 Chr.8: 38115042-38115042 on Build GRCh38
rs761762450 Chr.8: 38115026-38115026 on Build GRCh38
rs765382081 Chr.8: 38115040-38115040 on Build GRCh38
rs772750976 Chr.8: 38114981-38114981 on Build GRCh38
rs746406087 Chr.8: 38114964-38114964 on Build GRCh38
rs895785822 Chr.8: 38115284-38115284 on Build GRCh38
rs777459951 Chr.8: 38114929-38114929 on Build GRCh38
rs932850688 Chr.8: 38115187-38115187 on Build GRCh38
rs1041246617 Chr.8: 38115154-38115154 on Build GRCh38
rs532848680 Chr.8: 38114853-38114853 on Build GRCh38
rs1037047557 Chr.8: 38115266-38115266 on Build GRCh38
rs775961865 Chr.8: 38114984-38114984 on Build GRCh38
rs563229105 Chr.8: 38115050-38115050 on Build GRCh38
rs181361379 Chr.8: 38115046-38115046 on Build GRCh38
rs913544735 Chr.8: 38115112-38115112 on Build GRCh38
rs755758771 Chr.8: 38114925-38114925 on Build GRCh38
rs747978296 Chr.8: 38114920-38114920 on Build GRCh38
rs78064147 Chr.8: 38114893-38114893 on Build GRCh38
rs1031852628 Chr.8: 38115060-38115060 on Build GRCh38
rs770477338 Chr.8: 38114943-38114943 on Build GRCh38
rs760953167 Chr.8: 38114863-38114863 on Build GRCh38
rs942014919 Chr.8: 38115275-38115275 on Build GRCh38
rs1013480449 Chr.8: 38114843-38114843 on Build GRCh38
rs537289051 Chr.8: 38115241-38115241 on Build GRCh38
rs530743445 Chr.8: 38115130-38115131 on Build GRCh38
rs779244541 Chr.8: 38114953-38114953 on Build GRCh38
rs957796949 Chr.8: 38115079-38115079 on Build GRCh38
rs932273235 Chr.8: 38115209-38115209 on Build GRCh38
rs757374532 Chr.8: 38114876-38114876 on Build GRCh38
rs369481935 Chr.8: 38114891-38114891 on Build GRCh38
rs547233443 Chr.8: 38115130-38115130 on Build GRCh38
rs938743865 Chr.8: 38115308-38115308 on Build GRCh38
rs1004278501 Chr.8: 38115258-38115258 on Build GRCh38
rs781070656 Chr.8: 38114873-38114894 on Build GRCh38
rs768527842 Chr.8: 38114964-38114966 on Build GRCh38
rs1013513233 Chr.8: 38115294-38115294 on Build GRCh38
rs1048547562 Chr.8: 38115211-38115211 on Build GRCh38
rs768946191 Chr.8: 38115013-38115013 on Build GRCh38
rs776918529 Chr.8: 38115020-38115020 on Build GRCh38
rs921435751 Chr.8: 38115172-38115172 on Build GRCh38
rs987788712 Chr.8: 38115108-38115108 on Build GRCh38
rs764448200 Chr.8: 38114870-38114870 on Build GRCh38
rs961973835 Chr.8: 38114841-38114841 on Build GRCh38
rs897906194 Chr.8: 38115288-38115288 on Build GRCh38
rs766165843 Chr.8: 38114883-38114883 on Build GRCh38

View Full Product Details