Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 78658337-78658834 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZCCHC5
Entrez Gene ID
203430
Gene Name
zinc finger CCHC-type containing 5
Gene Aliases
Mar3, Mart3, ZHC5
UniGene
Hs.134873
Gene Chromosome Location
Chr.X: 78656069-78659328 on Build GRCh38
Transcripts
NM_152694.2

SNP Detail

SNP ID Chromosome Location
rs760859258 Chr.X: 78658433-78658433 on Build GRCh38
rs773367275 Chr.X: 78658421-78658421 on Build GRCh38
rs774069116 Chr.X: 78658412-78658412 on Build GRCh38
rs368928205 Chr.X: 78658437-78658437 on Build GRCh38
rs912814300 Chr.X: 78658601-78658601 on Build GRCh38
rs749219677 Chr.X: 78658395-78658395 on Build GRCh38
rs889874104 Chr.X: 78658776-78658776 on Build GRCh38
rs780068576 Chr.X: 78658394-78658394 on Build GRCh38
rs903548911 Chr.X: 78658392-78658392 on Build GRCh38
rs1016902926 Chr.X: 78658481-78658481 on Build GRCh38
rs998289867 Chr.X: 78658404-78658404 on Build GRCh38
rs181933542 Chr.X: 78658652-78658652 on Build GRCh38
rs1038383002 Chr.X: 78658808-78658808 on Build GRCh38
rs769563595 Chr.X: 78658390-78658390 on Build GRCh38
rs375362383 Chr.X: 78658539-78658539 on Build GRCh38
rs146799453 Chr.X: 78658557-78658557 on Build GRCh38
rs1055237313 Chr.X: 78658717-78658717 on Build GRCh38
rs985483577 Chr.X: 78658595-78658595 on Build GRCh38
rs186946312 Chr.X: 78658614-78658614 on Build GRCh38
rs41311547 Chr.X: 78658653-78658653 on Build GRCh38
rs148985625 Chr.X: 78658366-78658366 on Build GRCh38
rs1036825305 Chr.X: 78658803-78658803 on Build GRCh38
rs181777859 Chr.X: 78658547-78658547 on Build GRCh38
rs1002048498 Chr.X: 78658393-78658393 on Build GRCh38
rs937659257 Chr.X: 78658710-78658710 on Build GRCh38
rs751636354 Chr.X: 78658769-78658769 on Build GRCh38
rs925061263 Chr.X: 78658690-78658690 on Build GRCh38
rs766483176 Chr.X: 78658619-78658619 on Build GRCh38
rs1043469230 Chr.X: 78658668-78658668 on Build GRCh38
rs768472947 Chr.X: 78658399-78658399 on Build GRCh38
rs185152289 Chr.X: 78658712-78658712 on Build GRCh38
rs201025794 Chr.X: 78658457-78658457 on Build GRCh38
rs763793980 Chr.X: 78658425-78658432 on Build GRCh38
rs759524704 Chr.X: 78658740-78658740 on Build GRCh38
rs745391526 Chr.X: 78658374-78658374 on Build GRCh38
rs189458404 Chr.X: 78658618-78658618 on Build GRCh38
rs961417277 Chr.X: 78658500-78658500 on Build GRCh38
rs1051204634 Chr.X: 78658736-78658736 on Build GRCh38
rs934135452 Chr.X: 78658705-78658705 on Build GRCh38
rs1043334988 Chr.X: 78658378-78658378 on Build GRCh38
rs762568615 Chr.X: 78658421-78658421 on Build GRCh38
rs376915913 Chr.X: 78658420-78658420 on Build GRCh38
rs915512573 Chr.X: 78658607-78658607 on Build GRCh38
rs200467390 Chr.X: 78658470-78658470 on Build GRCh38
rs892209848 Chr.X: 78658365-78658365 on Build GRCh38
rs765840391 Chr.X: 78658792-78658792 on Build GRCh38
rs765997941 Chr.X: 78658549-78658549 on Build GRCh38
rs896395847 Chr.X: 78658741-78658741 on Build GRCh38
rs962652949 Chr.X: 78658489-78658489 on Build GRCh38
rs772629844 Chr.X: 78658538-78658538 on Build GRCh38
rs777165609 Chr.X: 78658441-78658441 on Build GRCh38
rs748374091 Chr.X: 78658417-78658417 on Build GRCh38
rs1025480101 Chr.X: 78658514-78658514 on Build GRCh38
rs756964754 Chr.X: 78658360-78658360 on Build GRCh38

View Full Product Details